Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 39025856-39026397 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NDUFS5
Entrez Gene ID
4725
Gene Name
NADH:ubiquinone oxidoreductase subunit S5
Gene Aliases
CI-15k, CI15K
UniGene
Hs.632385
Gene Chromosome Location
Chr.1: 39026295-39034636 on Build GRCh38
Transcripts
NM_001184979.1 NM_004552.2

SNP Detail

SNP ID Chromosome Location
rs960494682 Chr.1: 39026194-39026194 on Build GRCh38
rs753743795 Chr.1: 39026292-39026292 on Build GRCh38
rs773695404 Chr.1: 39026308-39026308 on Build GRCh38
rs74603394 Chr.1: 39026239-39026239 on Build GRCh38
rs979137571 Chr.1: 39026025-39026025 on Build GRCh38
rs1013437713 Chr.1: 39026212-39026212 on Build GRCh38
rs910718590 Chr.1: 39026072-39026072 on Build GRCh38
rs187135619 Chr.1: 39026006-39026006 on Build GRCh38
rs887742509 Chr.1: 39026340-39026340 on Build GRCh38
rs944881587 Chr.1: 39026083-39026083 on Build GRCh38
rs75387701 Chr.1: 39026265-39026265 on Build GRCh38
rs538543326 Chr.1: 39026219-39026219 on Build GRCh38
rs960567456 Chr.1: 39025952-39025952 on Build GRCh38
rs1049649076 Chr.1: 39026329-39026329 on Build GRCh38
rs920387978 Chr.1: 39026250-39026250 on Build GRCh38
rs771534968 Chr.1: 39026156-39026156 on Build GRCh38
rs150783077 Chr.1: 39025968-39025968 on Build GRCh38
rs139153519 Chr.1: 39026136-39026136 on Build GRCh38
rs1049043849 Chr.1: 39026144-39026144 on Build GRCh38
rs887807775 Chr.1: 39026170-39026170 on Build GRCh38
rs1002121657 Chr.1: 39026185-39026185 on Build GRCh38
rs28620880 Chr.1: 39026103-39026103 on Build GRCh38
rs1023458762 Chr.1: 39025939-39025939 on Build GRCh38
rs989211336 Chr.1: 39026056-39026056 on Build GRCh38
rs543987563 Chr.1: 39026334-39026334 on Build GRCh38
rs375280397 Chr.1: 39026367-39026367 on Build GRCh38
rs956032058 Chr.1: 39026036-39026036 on Build GRCh38
rs933546365 Chr.1: 39025886-39025887 on Build GRCh38
rs775244372 Chr.1: 39026150-39026150 on Build GRCh38
rs1037493004 Chr.1: 39025924-39025924 on Build GRCh38
rs1044936258 Chr.1: 39026263-39026263 on Build GRCh38
rs1040405212 Chr.1: 39026125-39026125 on Build GRCh38
rs886066438 Chr.1: 39025923-39025923 on Build GRCh38
rs919453354 Chr.1: 39026320-39026320 on Build GRCh38
rs1021749180 Chr.1: 39026130-39026130 on Build GRCh38
rs760428244 Chr.1: 39026233-39026233 on Build GRCh38
rs1984600 Chr.1: 39026255-39026255 on Build GRCh38
rs929397094 Chr.1: 39026322-39026322 on Build GRCh38
rs974896034 Chr.1: 39026175-39026175 on Build GRCh38
rs922209742 Chr.1: 39025883-39025883 on Build GRCh38
rs193192383 Chr.1: 39026220-39026220 on Build GRCh38
rs1037756878 Chr.1: 39026234-39026234 on Build GRCh38
rs745408141 Chr.1: 39025985-39025985 on Build GRCh38
rs28589353 Chr.1: 39026104-39026104 on Build GRCh38
rs555014861 Chr.1: 39026259-39026259 on Build GRCh38
rs1051151097 Chr.1: 39026068-39026068 on Build GRCh38
rs386630512 Chr.1: 39025936-39025938 on Build GRCh38
rs936253967 Chr.1: 39026052-39026052 on Build GRCh38
rs1040540110 Chr.1: 39026309-39026309 on Build GRCh38
rs910813780 Chr.1: 39026303-39026303 on Build GRCh38
rs10159267 Chr.1: 39025893-39025893 on Build GRCh38
rs1002557754 Chr.1: 39025925-39025925 on Build GRCh38
rs10157617 Chr.1: 39025923-39025923 on Build GRCh38
rs1002624489 Chr.1: 39026363-39026363 on Build GRCh38
rs371600743 Chr.1: 39026192-39026192 on Build GRCh38
rs767214476 Chr.1: 39026041-39026041 on Build GRCh38
rs897928527 Chr.1: 39026131-39026131 on Build GRCh38
rs906460860 Chr.1: 39025942-39025942 on Build GRCh38
rs966468863 Chr.1: 39026228-39026228 on Build GRCh38
rs185196663 Chr.1: 39026364-39026364 on Build GRCh38
rs116390318 Chr.1: 39025938-39025938 on Build GRCh38
rs757307188 Chr.1: 39026362-39026362 on Build GRCh38
rs922081390 Chr.1: 39026178-39026178 on Build GRCh38

View Full Product Details