Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81868375-81868917 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGDIA
Entrez Gene ID
396
Gene Name
Rho GDP dissociation inhibitor alpha
Gene Aliases
GDIA1, HEL-S-47e, NPHS8, RHOGDI, RHOGDI-1
UniGene
Hs.159161
Gene Chromosome Location
Chr.17: 81867719-81871406 on Build GRCh38
Transcripts
NM_001185077.2 NM_001185078.2 NM_001301240.1 NM_001301241.1 NM_001301242.1 NM_001301243.1 NM_004309.5 NR_125441.1 XM_011523574.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4436391 c.597G>A p.K199K Chr.17: 81868894-81868894 on Build GRCh38
6281325 c.614G>A p.*205* Chr.17: 81868877-81868877 on Build GRCh38
3403378 c.587T>G p.L196R Chr.17: 81868904-81868904 on Build GRCh38
4070745 c.600G>T p.K200N Chr.17: 81868891-81868891 on Build GRCh38
1610984 c.598A>G p.K200E Chr.17: 81868893-81868893 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs758009792 Chr.17: 81868548-81868548 on Build GRCh38
rs571770530 Chr.17: 81868840-81868840 on Build GRCh38
rs923534695 Chr.17: 81868423-81868423 on Build GRCh38
rs369699594 Chr.17: 81868841-81868841 on Build GRCh38
rs775598375 Chr.17: 81868456-81868456 on Build GRCh38
rs914312395 Chr.17: 81868586-81868587 on Build GRCh38
rs982131775 Chr.17: 81868584-81868584 on Build GRCh38
rs1003613126 Chr.17: 81868702-81868702 on Build GRCh38
rs750215632 Chr.17: 81868829-81868829 on Build GRCh38
rs934366174 Chr.17: 81868452-81868452 on Build GRCh38
rs369742215 Chr.17: 81868571-81868571 on Build GRCh38
rs955133122 Chr.17: 81868706-81868706 on Build GRCh38
rs753761809 Chr.17: 81868830-81868830 on Build GRCh38
rs779910117 Chr.17: 81868773-81868773 on Build GRCh38
rs1032192506 Chr.17: 81868433-81868433 on Build GRCh38
rs557094400 Chr.17: 81868470-81868470 on Build GRCh38
rs140429915 Chr.17: 81868482-81868482 on Build GRCh38
rs772848070 Chr.17: 81868851-81868851 on Build GRCh38
rs751122756 Chr.17: 81868807-81868807 on Build GRCh38
rs897531590 Chr.17: 81868396-81868396 on Build GRCh38
rs970792413 Chr.17: 81868572-81868572 on Build GRCh38
rs766942075 Chr.17: 81868847-81868847 on Build GRCh38
rs373508832 Chr.17: 81868861-81868861 on Build GRCh38
rs1057282 Chr.17: 81868849-81868849 on Build GRCh38
rs1016323815 Chr.17: 81868672-81868674 on Build GRCh38
rs1038261460 Chr.17: 81868804-81868804 on Build GRCh38
rs1044235684 Chr.17: 81868631-81868631 on Build GRCh38
rs941350334 Chr.17: 81868799-81868799 on Build GRCh38
rs769764976 Chr.17: 81868622-81868622 on Build GRCh38
rs530554967 Chr.17: 81868819-81868819 on Build GRCh38
rs760377906 Chr.17: 81868853-81868853 on Build GRCh38
rs757596677 Chr.17: 81868888-81868888 on Build GRCh38
rs113912799 Chr.17: 81868430-81868430 on Build GRCh38
rs34659851 Chr.17: 81868584-81868584 on Build GRCh38
rs761416911 Chr.17: 81868514-81868514 on Build GRCh38
rs949426713 Chr.17: 81868441-81868441 on Build GRCh38
rs777277511 Chr.17: 81868797-81868797 on Build GRCh38
rs984424896 Chr.17: 81868612-81868612 on Build GRCh38
rs1057284 Chr.17: 81868837-81868837 on Build GRCh38
rs750195456 Chr.17: 81868769-81868769 on Build GRCh38
rs780821323 Chr.17: 81868765-81868765 on Build GRCh38
rs1026307601 Chr.17: 81868484-81868484 on Build GRCh38
rs964875144 Chr.17: 81868537-81868537 on Build GRCh38
rs532896353 Chr.17: 81868659-81868659 on Build GRCh38
rs746580972 Chr.17: 81868843-81868843 on Build GRCh38
rs901315268 Chr.17: 81868658-81868658 on Build GRCh38
rs984534034 Chr.17: 81868731-81868731 on Build GRCh38
rs757001317 Chr.17: 81868564-81868564 on Build GRCh38
rs752292660 Chr.17: 81868540-81868540 on Build GRCh38
rs778825097 Chr.17: 81868831-81868831 on Build GRCh38
rs767150158 Chr.17: 81868814-81868814 on Build GRCh38
rs1020756197 Chr.17: 81868698-81868698 on Build GRCh38
rs1048210857 Chr.17: 81868636-81868636 on Build GRCh38
rs954975497 Chr.17: 81868544-81868544 on Build GRCh38
rs749862467 Chr.17: 81868891-81868893 on Build GRCh38
rs758232372 Chr.17: 81868830-81868836 on Build GRCh38
rs770777679 Chr.17: 81868421-81868421 on Build GRCh38
rs745773792 Chr.17: 81868580-81868580 on Build GRCh38
rs3809896 Chr.17: 81868672-81868672 on Build GRCh38
rs996782121 Chr.17: 81868680-81868680 on Build GRCh38
rs777777300 Chr.17: 81868885-81868885 on Build GRCh38
rs779870102 Chr.17: 81868835-81868836 on Build GRCh38
rs773673054 Chr.17: 81868678-81868678 on Build GRCh38
rs997234113 Chr.17: 81868510-81868510 on Build GRCh38
rs1044737852 Chr.17: 81868476-81868476 on Build GRCh38
rs763492845 Chr.17: 81868801-81868801 on Build GRCh38
rs886337329 Chr.17: 81868821-81868821 on Build GRCh38
rs754784895 Chr.17: 81868500-81868500 on Build GRCh38
rs202026197 Chr.17: 81868833-81868833 on Build GRCh38
rs189865523 Chr.17: 81868633-81868633 on Build GRCh38
rs764015476 Chr.17: 81868768-81868804 on Build GRCh38
rs563157861 Chr.17: 81868697-81868697 on Build GRCh38
rs1042272850 Chr.17: 81868857-81868857 on Build GRCh38
rs11546984 Chr.17: 81868871-81868871 on Build GRCh38
rs755927023 Chr.17: 81868770-81868770 on Build GRCh38
rs773335010 Chr.17: 81868795-81868795 on Build GRCh38
rs1003327596 Chr.17: 81868483-81868483 on Build GRCh38
rs893717708 Chr.17: 81868693-81868693 on Build GRCh38
rs748293793 Chr.17: 81868786-81868786 on Build GRCh38
rs766228744 Chr.17: 81868826-81868826 on Build GRCh38
rs752585650 Chr.17: 81868878-81868878 on Build GRCh38
rs761261929 Chr.17: 81868812-81868812 on Build GRCh38
rs11546989 Chr.17: 81868766-81868766 on Build GRCh38
rs937997362 Chr.17: 81868609-81868609 on Build GRCh38
rs763767452 Chr.17: 81868741-81868741 on Build GRCh38
rs772310030 Chr.17: 81868673-81868673 on Build GRCh38
rs970517789 Chr.17: 81868699-81868699 on Build GRCh38
rs766138328 Chr.17: 81868859-81868859 on Build GRCh38
rs534920169 Chr.17: 81868425-81868425 on Build GRCh38
rs1032657953 Chr.17: 81868517-81868517 on Build GRCh38
rs959440482 Chr.17: 81868713-81868713 on Build GRCh38
rs970320215 Chr.17: 81868735-81868747 on Build GRCh38
rs753882835 Chr.17: 81868793-81868794 on Build GRCh38
rs768717665 Chr.17: 81868774-81868774 on Build GRCh38
rs754757370 Chr.17: 81868862-81868862 on Build GRCh38
rs1030017053 Chr.17: 81868513-81868513 on Build GRCh38
rs766949406 Chr.17: 81868704-81868704 on Build GRCh38
rs530110406 Chr.17: 81868691-81868691 on Build GRCh38
rs982079574 Chr.17: 81868449-81868449 on Build GRCh38
rs879074652 Chr.17: 81868422-81868422 on Build GRCh38
rs546790617 Chr.17: 81868836-81868836 on Build GRCh38
rs763133358 Chr.17: 81868469-81868469 on Build GRCh38
rs777514732 Chr.17: 81868562-81868562 on Build GRCh38
rs940176223 Chr.17: 81868485-81868485 on Build GRCh38
rs573974891 Chr.17: 81868597-81868597 on Build GRCh38
rs780000070 Chr.17: 81868630-81868630 on Build GRCh38
rs948445895 Chr.17: 81868749-81868749 on Build GRCh38
rs943640634 Chr.17: 81868415-81868415 on Build GRCh38
rs567596796 Chr.17: 81868400-81868400 on Build GRCh38
rs201139992 Chr.17: 81868835-81868835 on Build GRCh38
rs778903662 Chr.17: 81868811-81868811 on Build GRCh38
rs897038186 Chr.17: 81868810-81868810 on Build GRCh38
rs1006163200 Chr.17: 81868671-81868671 on Build GRCh38
rs1057283 Chr.17: 81868845-81868845 on Build GRCh38
rs924285282 Chr.17: 81868709-81868709 on Build GRCh38
rs1024936311 Chr.17: 81868542-81868552 on Build GRCh38
rs774369485 Chr.17: 81868668-81868668 on Build GRCh38
rs909344504 Chr.17: 81868414-81868414 on Build GRCh38
rs543025140 Chr.17: 81868494-81868494 on Build GRCh38
rs754414636 Chr.17: 81868708-81868708 on Build GRCh38
rs151190257 Chr.17: 81868455-81868455 on Build GRCh38
rs772376841 Chr.17: 81868791-81868791 on Build GRCh38
rs371644062 Chr.17: 81868767-81868767 on Build GRCh38
rs765171247 Chr.17: 81868868-81868868 on Build GRCh38
rs185290419 Chr.17: 81868563-81868563 on Build GRCh38
rs988211180 Chr.17: 81868437-81868437 on Build GRCh38
rs578069979 Chr.17: 81868436-81868436 on Build GRCh38
rs758434196 Chr.17: 81868882-81868882 on Build GRCh38
rs761068260 Chr.17: 81868796-81868796 on Build GRCh38
rs750057679 Chr.17: 81868818-81868818 on Build GRCh38
rs1009457365 Chr.17: 81868843-81868843 on Build GRCh38
rs938296634 Chr.17: 81868467-81868467 on Build GRCh38
rs750889382 Chr.17: 81868492-81868492 on Build GRCh38
rs774579724 Chr.17: 81868479-81868479 on Build GRCh38
rs1007497861 Chr.17: 81868516-81868516 on Build GRCh38
rs779105538 Chr.17: 81868777-81868777 on Build GRCh38
rs376097951 Chr.17: 81868509-81868509 on Build GRCh38
rs745888774 Chr.17: 81868450-81868450 on Build GRCh38
rs751161946 Chr.17: 81868744-81868744 on Build GRCh38
rs562000594 Chr.17: 81868588-81868588 on Build GRCh38
rs757131763 Chr.17: 81868810-81868811 on Build GRCh38
rs762026141 Chr.17: 81868480-81868480 on Build GRCh38
rs768213538 Chr.17: 81868845-81868848 on Build GRCh38
rs553139378 Chr.17: 81868427-81868427 on Build GRCh38
rs746350097 Chr.17: 81868838-81868838 on Build GRCh38
rs762490731 Chr.17: 81868726-81868726 on Build GRCh38
rs955720841 Chr.17: 81868724-81868724 on Build GRCh38
rs997183097 Chr.17: 81868407-81868407 on Build GRCh38
rs780561517 Chr.17: 81868877-81868885 on Build GRCh38
rs961636919 Chr.17: 81868775-81868775 on Build GRCh38
rs913980090 Chr.17: 81868440-81868440 on Build GRCh38
rs541274253 Chr.17: 81868598-81868598 on Build GRCh38
rs957928529 Chr.17: 81868545-81868545 on Build GRCh38
rs760182464 Chr.17: 81868800-81868800 on Build GRCh38
rs756985934 Chr.17: 81868748-81868748 on Build GRCh38
rs764803182 Chr.17: 81868581-81868581 on Build GRCh38
rs1036155994 Chr.17: 81868488-81868488 on Build GRCh38
rs767433421 Chr.17: 81868635-81868635 on Build GRCh38
rs764816110 Chr.17: 81868525-81868525 on Build GRCh38
rs567655758 Chr.17: 81868834-81868834 on Build GRCh38
rs953330818 Chr.17: 81868426-81868426 on Build GRCh38

View Full Product Details