Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 82036293-82036799 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCXR
Entrez Gene ID
51181
Gene Name
dicarbonyl and L-xylulose reductase
Gene Aliases
DCR, HCR2, HCRII, KIDCR, P34H, PNTSU, SDR20C1, XR
UniGene
Hs.9857
Gene Chromosome Location
Chr.17: 82035881-82037697 on Build GRCh38
Transcripts
NM_001195218.1 NM_016286.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6352058 c.345G>A p.S115S Chr.17: 82036724-82036724 on Build GRCh38
5853816 c.427G>A p.V143I Chr.17: 82036565-82036565 on Build GRCh38
4070794 c.358A>G p.R120G Chr.17: 82036634-82036634 on Build GRCh38
7428764 c.446_448delACT p.Y149delY Chr.17: 82036544-82036546 on Build GRCh38
5638934 c.418C>T p.Q140* Chr.17: 82036574-82036574 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs972000005 Chr.17: 82036525-82036525 on Build GRCh38
rs757440269 Chr.17: 82036365-82036365 on Build GRCh38
rs754208020 Chr.17: 82036550-82036550 on Build GRCh38
rs747057271 Chr.17: 82036670-82036670 on Build GRCh38
rs202081300 Chr.17: 82036471-82036471 on Build GRCh38
rs9780 Chr.17: 82036724-82036724 on Build GRCh38
rs940473032 Chr.17: 82036327-82036327 on Build GRCh38
rs751641743 Chr.17: 82036362-82036362 on Build GRCh38
rs376209981 Chr.17: 82036379-82036379 on Build GRCh38
rs372987720 Chr.17: 82036364-82036364 on Build GRCh38
rs775400332 Chr.17: 82036434-82036434 on Build GRCh38
rs764999220 Chr.17: 82036431-82036431 on Build GRCh38
rs753192836 Chr.17: 82036560-82036560 on Build GRCh38
rs370221792 Chr.17: 82036771-82036771 on Build GRCh38
rs557076879 Chr.17: 82036743-82036743 on Build GRCh38
rs749872041 Chr.17: 82036388-82036388 on Build GRCh38
rs925457760 Chr.17: 82036481-82036481 on Build GRCh38
rs781290247 Chr.17: 82036373-82036373 on Build GRCh38
rs773110073 Chr.17: 82036612-82036612 on Build GRCh38
rs777550591 Chr.17: 82036733-82036733 on Build GRCh38
rs765937746 Chr.17: 82036703-82036703 on Build GRCh38
rs770569294 Chr.17: 82036414-82036414 on Build GRCh38
rs763406253 Chr.17: 82036687-82036687 on Build GRCh38
rs369458736 Chr.17: 82036739-82036739 on Build GRCh38
rs372696285 Chr.17: 82036443-82036443 on Build GRCh38
rs779587311 Chr.17: 82036478-82036478 on Build GRCh38
rs547679682 Chr.17: 82036374-82036374 on Build GRCh38
rs765462896 Chr.17: 82036622-82036626 on Build GRCh38
rs759764247 Chr.17: 82036680-82036680 on Build GRCh38
rs112902290 Chr.17: 82036542-82036542 on Build GRCh38
rs762809374 Chr.17: 82036438-82036438 on Build GRCh38
rs368246980 Chr.17: 82036629-82036629 on Build GRCh38
rs371698202 Chr.17: 82036548-82036548 on Build GRCh38
rs369595969 Chr.17: 82036673-82036673 on Build GRCh38
rs373494830 Chr.17: 82036769-82036769 on Build GRCh38
rs533137522 Chr.17: 82036469-82036469 on Build GRCh38
rs774831272 Chr.17: 82036772-82036772 on Build GRCh38
rs371674068 Chr.17: 82036659-82036659 on Build GRCh38
rs76055808 Chr.17: 82036685-82036685 on Build GRCh38
rs536701155 Chr.17: 82036524-82036524 on Build GRCh38
rs769865642 Chr.17: 82036744-82036744 on Build GRCh38
rs377194919 Chr.17: 82036652-82036652 on Build GRCh38
rs770388738 Chr.17: 82036491-82036491 on Build GRCh38
rs746517171 Chr.17: 82036401-82036401 on Build GRCh38
rs200134744 Chr.17: 82036543-82036543 on Build GRCh38
rs760707101 Chr.17: 82036453-82036453 on Build GRCh38
rs372804928 Chr.17: 82036677-82036677 on Build GRCh38
rs774329370 Chr.17: 82036576-82036576 on Build GRCh38
rs1006126343 Chr.17: 82036408-82036408 on Build GRCh38
rs1034480967 Chr.17: 82036720-82036720 on Build GRCh38
rs776205242 Chr.17: 82036389-82036390 on Build GRCh38
rs374496366 Chr.17: 82036452-82036452 on Build GRCh38
rs760474627 Chr.17: 82036335-82036335 on Build GRCh38
rs377118987 Chr.17: 82036317-82036317 on Build GRCh38
rs753109310 Chr.17: 82036643-82036643 on Build GRCh38
rs779532057 Chr.17: 82036556-82036556 on Build GRCh38
rs764030303 Chr.17: 82036440-82036440 on Build GRCh38
rs142512551 Chr.17: 82036396-82036396 on Build GRCh38
rs776431707 Chr.17: 82036338-82036338 on Build GRCh38
rs374021893 Chr.17: 82036497-82036497 on Build GRCh38
rs200246714 Chr.17: 82036697-82036697 on Build GRCh38
rs752672219 Chr.17: 82036355-82036355 on Build GRCh38
rs772055207 Chr.17: 82036599-82036599 on Build GRCh38
rs766670193 Chr.17: 82036623-82036623 on Build GRCh38
rs199558783 Chr.17: 82036419-82036419 on Build GRCh38
rs201800605 Chr.17: 82036773-82036773 on Build GRCh38
rs1034161197 Chr.17: 82036383-82036383 on Build GRCh38
rs755603658 Chr.17: 82036698-82036698 on Build GRCh38
rs776828939 Chr.17: 82036502-82036503 on Build GRCh38
rs780500485 Chr.17: 82036565-82036565 on Build GRCh38
rs370292802 Chr.17: 82036494-82036494 on Build GRCh38
rs371112705 Chr.17: 82036763-82036763 on Build GRCh38
rs750656443 Chr.17: 82036458-82036458 on Build GRCh38
rs765181801 Chr.17: 82036351-82036351 on Build GRCh38
rs749049318 Chr.17: 82036571-82036571 on Build GRCh38
rs772988865 Chr.17: 82036330-82036330 on Build GRCh38
rs375268755 Chr.17: 82036662-82036662 on Build GRCh38
rs201185898 Chr.17: 82036476-82036476 on Build GRCh38
rs776179930 Chr.17: 82036415-82036415 on Build GRCh38
rs778509874 Chr.17: 82036660-82036660 on Build GRCh38
rs150760136 Chr.17: 82036706-82036706 on Build GRCh38
rs769386254 Chr.17: 82036570-82036570 on Build GRCh38
rs745394831 Chr.17: 82036567-82036567 on Build GRCh38
rs201745238 Chr.17: 82036632-82036632 on Build GRCh38
rs755658533 Chr.17: 82036651-82036651 on Build GRCh38
rs757310330 Chr.17: 82036445-82036445 on Build GRCh38
rs769112869 Chr.17: 82036500-82036505 on Build GRCh38
rs763022554 Chr.17: 82036723-82036724 on Build GRCh38
rs775207320 Chr.17: 82036498-82036498 on Build GRCh38
rs765510515 Chr.17: 82036683-82036683 on Build GRCh38
rs761727634 Chr.17: 82036593-82036593 on Build GRCh38
rs1013576157 Chr.17: 82036344-82036344 on Build GRCh38
rs746489718 Chr.17: 82036486-82036486 on Build GRCh38
rs755427002 Chr.17: 82036555-82036555 on Build GRCh38
rs771131569 Chr.17: 82036740-82036740 on Build GRCh38
rs771858493 Chr.17: 82036663-82036663 on Build GRCh38
rs985887100 Chr.17: 82036748-82036748 on Build GRCh38
rs747859957 Chr.17: 82036713-82036713 on Build GRCh38
rs377435527 Chr.17: 82036770-82036770 on Build GRCh38
rs376885972 Chr.17: 82036667-82036667 on Build GRCh38
rs758772890 Chr.17: 82036644-82036644 on Build GRCh38
rs759273434 Chr.17: 82036346-82036346 on Build GRCh38
rs750584259 Chr.17: 82036657-82036664 on Build GRCh38
rs780688197 Chr.17: 82036387-82036387 on Build GRCh38
rs1032542088 Chr.17: 82036752-82036752 on Build GRCh38
rs768538061 Chr.17: 82036500-82036500 on Build GRCh38
rs371130365 Chr.17: 82036389-82036389 on Build GRCh38
rs756492160 Chr.17: 82036459-82036459 on Build GRCh38
rs775615109 Chr.17: 82036325-82036325 on Build GRCh38
rs529196400 Chr.17: 82036316-82036316 on Build GRCh38
rs748339072 Chr.17: 82036568-82036568 on Build GRCh38
rs372927639 Chr.17: 82036315-82036315 on Build GRCh38
rs375839857 Chr.17: 82036616-82036616 on Build GRCh38
rs761927545 Chr.17: 82036514-82036514 on Build GRCh38
rs375837859 Chr.17: 82036618-82036618 on Build GRCh38
rs367865246 Chr.17: 82036515-82036515 on Build GRCh38
rs370503205 Chr.17: 82036561-82036561 on Build GRCh38
rs761071627 Chr.17: 82036485-82036486 on Build GRCh38
rs1030418190 Chr.17: 82036462-82036462 on Build GRCh38
rs750656230 Chr.17: 82036536-82036536 on Build GRCh38
rs776981111 Chr.17: 82036679-82036679 on Build GRCh38
rs1044015276 Chr.17: 82036681-82036681 on Build GRCh38
rs369621092 Chr.17: 82036337-82036337 on Build GRCh38
rs774308885 Chr.17: 82036513-82036513 on Build GRCh38
rs753355351 Chr.17: 82036704-82036704 on Build GRCh38
rs754570980 Chr.17: 82036705-82036705 on Build GRCh38
rs764224937 Chr.17: 82036361-82036361 on Build GRCh38
rs773234858 Chr.17: 82036666-82036666 on Build GRCh38
rs367769179 Chr.17: 82036657-82036657 on Build GRCh38
rs758459371 Chr.17: 82036356-82036356 on Build GRCh38

View Full Product Details