Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 82391300-82391827 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OGFOD3
Entrez Gene ID
79701
Gene Name
2-oxoglutarate and iron dependent oxygenase domain containing 3
Gene Aliases
C17orf101
UniGene
Hs.187505
Gene Chromosome Location
Chr.17: 82389210-82418637 on Build GRCh38
Transcripts
NM_024648.2 NM_175902.4 NR_033265.1

SNP Detail

SNP ID Chromosome Location
rs72857414 Chr.17: 82391761-82391761 on Build GRCh38
rs943671193 Chr.17: 82391798-82391798 on Build GRCh38
rs182965211 Chr.17: 82391670-82391670 on Build GRCh38
rs552385841 Chr.17: 82391707-82391707 on Build GRCh38
rs968755751 Chr.17: 82391736-82391736 on Build GRCh38
rs749738400 Chr.17: 82391729-82391729 on Build GRCh38
rs908244231 Chr.17: 82391796-82391796 on Build GRCh38
rs968723194 Chr.17: 82391452-82391452 on Build GRCh38
rs992466557 Chr.17: 82391558-82391558 on Build GRCh38
rs986084004 Chr.17: 82391336-82391338 on Build GRCh38
rs754015118 Chr.17: 82391373-82391373 on Build GRCh38
rs964540234 Chr.17: 82391396-82391396 on Build GRCh38
rs1031700580 Chr.17: 82391753-82391753 on Build GRCh38
rs960017323 Chr.17: 82391756-82391756 on Build GRCh38
rs866672722 Chr.17: 82391708-82391708 on Build GRCh38
rs980982031 Chr.17: 82391780-82391780 on Build GRCh38
rs949019015 Chr.17: 82391573-82391573 on Build GRCh38
rs990696159 Chr.17: 82391785-82391785 on Build GRCh38
rs543559361 Chr.17: 82391325-82391325 on Build GRCh38
rs1043123493 Chr.17: 82391459-82391459 on Build GRCh38
rs756215476 Chr.17: 82391587-82391587 on Build GRCh38
rs1044840549 Chr.17: 82391611-82391611 on Build GRCh38
rs553092259 Chr.17: 82391763-82391763 on Build GRCh38
rs968696352 Chr.17: 82391750-82391750 on Build GRCh38
rs923862199 Chr.17: 82391499-82391499 on Build GRCh38
rs151081594 Chr.17: 82391432-82391432 on Build GRCh38
rs566511219 Chr.17: 82391572-82391572 on Build GRCh38
rs190515887 Chr.17: 82391411-82391411 on Build GRCh38
rs956648275 Chr.17: 82391535-82391535 on Build GRCh38
rs1036671207 Chr.17: 82391580-82391580 on Build GRCh38
rs531405566 Chr.17: 82391324-82391324 on Build GRCh38
rs748277775 Chr.17: 82391445-82391445 on Build GRCh38
rs868759077 Chr.17: 82391406-82391406 on Build GRCh38
rs931102025 Chr.17: 82391793-82391793 on Build GRCh38
rs185846081 Chr.17: 82391768-82391768 on Build GRCh38
rs937892425 Chr.17: 82391503-82391503 on Build GRCh38
rs919754458 Chr.17: 82391792-82391792 on Build GRCh38
rs965688306 Chr.17: 82391706-82391706 on Build GRCh38
rs28517555 Chr.17: 82391749-82391749 on Build GRCh38
rs965758313 Chr.17: 82391359-82391359 on Build GRCh38
rs142650870 Chr.17: 82391803-82391803 on Build GRCh38
rs947851918 Chr.17: 82391610-82391610 on Build GRCh38
rs930990772 Chr.17: 82391613-82391613 on Build GRCh38
rs1007100836 Chr.17: 82391696-82391696 on Build GRCh38
rs866229032 Chr.17: 82391612-82391612 on Build GRCh38
rs755284515 Chr.17: 82391376-82391376 on Build GRCh38
rs771339291 Chr.17: 82391735-82391735 on Build GRCh38
rs1008501828 Chr.17: 82391693-82391693 on Build GRCh38
rs764262002 Chr.17: 82391329-82391329 on Build GRCh38
rs900010614 Chr.17: 82391704-82391704 on Build GRCh38
rs547314604 Chr.17: 82391438-82391438 on Build GRCh38
rs761330339 Chr.17: 82391800-82391800 on Build GRCh38
rs757587448 Chr.17: 82391697-82391697 on Build GRCh38
rs912849814 Chr.17: 82391337-82391337 on Build GRCh38
rs988797563 Chr.17: 82391400-82391400 on Build GRCh38
rs778437702 Chr.17: 82391596-82391596 on Build GRCh38
rs977380018 Chr.17: 82391467-82391467 on Build GRCh38
rs925071642 Chr.17: 82391475-82391475 on Build GRCh38
rs536135291 Chr.17: 82391794-82391794 on Build GRCh38
rs556682972 Chr.17: 82391762-82391762 on Build GRCh38
rs1046752072 Chr.17: 82391650-82391650 on Build GRCh38
rs368228942 Chr.17: 82391802-82391802 on Build GRCh38
rs913246799 Chr.17: 82391401-82391401 on Build GRCh38

View Full Product Details