Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 5291626-5292129 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZBTB14
Entrez Gene ID
7541
Gene Name
zinc finger and BTB domain containing 14
Gene Aliases
ZF5, ZFP-161, ZFP-5, ZFP161, ZNF478
UniGene
Hs.592340
Gene Chromosome Location
Chr.18: 5289019-5297053 on Build GRCh38
Transcripts
NM_001143823.2 NM_001243702.1 NM_001243704.1 NM_003409.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
989029 c.89G>A p.R30H Chr.18: 5292119-5292119 on Build GRCh38
316675 c.203_206delTTGA p.L68fs*8 Chr.18: 5292002-5292005 on Build GRCh38
4897062 c.195C>T p.F65F Chr.18: 5292013-5292013 on Build GRCh38
4923168 c.274T>A p.Y92N Chr.18: 5291934-5291934 on Build GRCh38
6585342 c.141C>G p.F47L Chr.18: 5292067-5292067 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370055289 Chr.18: 5292044-5292044 on Build GRCh38
rs374388980 Chr.18: 5291800-5291800 on Build GRCh38
rs138010445 Chr.18: 5291695-5291695 on Build GRCh38
rs751432738 Chr.18: 5291763-5291763 on Build GRCh38
rs899193748 Chr.18: 5291795-5291795 on Build GRCh38
rs767418961 Chr.18: 5291925-5291925 on Build GRCh38
rs762478865 Chr.18: 5291758-5291758 on Build GRCh38
rs746655476 Chr.18: 5291944-5291944 on Build GRCh38
rs757007838 Chr.18: 5291685-5291685 on Build GRCh38
rs775006811 Chr.18: 5292041-5292043 on Build GRCh38
rs748719359 Chr.18: 5291818-5291818 on Build GRCh38
rs775171169 Chr.18: 5291873-5291873 on Build GRCh38
rs201423565 Chr.18: 5291911-5291911 on Build GRCh38
rs141599897 Chr.18: 5291650-5291650 on Build GRCh38
rs142776648 Chr.18: 5292061-5292061 on Build GRCh38
rs773755916 Chr.18: 5291832-5291832 on Build GRCh38
rs1034501304 Chr.18: 5291744-5291744 on Build GRCh38
rs620685 Chr.18: 5292014-5292014 on Build GRCh38
rs534567842 Chr.18: 5292091-5292091 on Build GRCh38
rs368255676 Chr.18: 5291808-5291808 on Build GRCh38
rs200323387 Chr.18: 5291647-5291647 on Build GRCh38
rs750188289 Chr.18: 5291799-5291799 on Build GRCh38
rs779573117 Chr.18: 5291810-5291810 on Build GRCh38
rs761081643 Chr.18: 5291854-5291854 on Build GRCh38
rs962156159 Chr.18: 5291964-5291964 on Build GRCh38
rs34590676 Chr.18: 5292066-5292066 on Build GRCh38
rs763970791 Chr.18: 5291759-5291759 on Build GRCh38
rs76196850 Chr.18: 5291709-5291709 on Build GRCh38
rs1040739964 Chr.18: 5291697-5291697 on Build GRCh38
rs762375909 Chr.18: 5291908-5291908 on Build GRCh38
rs751381362 Chr.18: 5291681-5291681 on Build GRCh38
rs761806426 Chr.18: 5291924-5291924 on Build GRCh38
rs368181021 Chr.18: 5291659-5291659 on Build GRCh38
rs755895516 Chr.18: 5291940-5291940 on Build GRCh38
rs780273813 Chr.18: 5291806-5291806 on Build GRCh38
rs762217065 Chr.18: 5291646-5291646 on Build GRCh38
rs144136246 Chr.18: 5291855-5291855 on Build GRCh38
rs371540884 Chr.18: 5292058-5292058 on Build GRCh38
rs146246023 Chr.18: 5291677-5291677 on Build GRCh38
rs753471496 Chr.18: 5291649-5291649 on Build GRCh38
rs748212245 Chr.18: 5292043-5292043 on Build GRCh38
rs746033772 Chr.18: 5291702-5291702 on Build GRCh38
rs7235420 Chr.18: 5291792-5291792 on Build GRCh38
rs778378352 Chr.18: 5292054-5292054 on Build GRCh38
rs371564765 Chr.18: 5291797-5291797 on Build GRCh38
rs535631234 Chr.18: 5291959-5291959 on Build GRCh38
rs926276947 Chr.18: 5292095-5292095 on Build GRCh38
rs764915518 Chr.18: 5291750-5291750 on Build GRCh38
rs764708261 Chr.18: 5291653-5291653 on Build GRCh38
rs775346339 Chr.18: 5291811-5291811 on Build GRCh38
rs762270017 Chr.18: 5291730-5291730 on Build GRCh38
rs1010277012 Chr.18: 5291706-5291706 on Build GRCh38
rs759333646 Chr.18: 5291734-5291734 on Build GRCh38
rs768712260 Chr.18: 5291809-5291809 on Build GRCh38
rs375725731 Chr.18: 5291926-5291926 on Build GRCh38
rs620652 Chr.18: 5292031-5292031 on Build GRCh38
rs149549029 Chr.18: 5291731-5291731 on Build GRCh38
rs555237306 Chr.18: 5291949-5291949 on Build GRCh38
rs879239857 Chr.18: 5291670-5291670 on Build GRCh38
rs769064199 Chr.18: 5291721-5291721 on Build GRCh38
rs770050145 Chr.18: 5291704-5291704 on Build GRCh38
rs752201154 Chr.18: 5291751-5291751 on Build GRCh38
rs772698050 Chr.18: 5291821-5291821 on Build GRCh38
rs892625778 Chr.18: 5291657-5291657 on Build GRCh38
rs7235740 Chr.18: 5291978-5291978 on Build GRCh38
rs755138426 Chr.18: 5291986-5291986 on Build GRCh38
rs758310921 Chr.18: 5291660-5291660 on Build GRCh38
rs761432696 Chr.18: 5291836-5291837 on Build GRCh38
rs532980484 Chr.18: 5291860-5291860 on Build GRCh38
rs762596527 Chr.18: 5291885-5291887 on Build GRCh38
rs1024867648 Chr.18: 5292055-5292055 on Build GRCh38
rs757058661 Chr.18: 5291765-5291765 on Build GRCh38
rs749364948 Chr.18: 5291710-5291710 on Build GRCh38
rs774878982 Chr.18: 5291725-5291725 on Build GRCh38

View Full Product Details