Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 5969301-5969798 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
L3MBTL4
Entrez Gene ID
91133
Gene Name
l(3)mbt-like 4 (Drosophila)
Gene Aliases
HsT1031
UniGene
Hs.128279
Gene Chromosome Location
Chr.18: 5954703-6414911 on Build GRCh38
Transcripts
NM_173464.3 XM_005258166.1 XM_011525757.1 XM_011525758.1 XM_011525759.1 XM_011525760.1 XM_011525761.1 XM_011525767.2 XM_017026074.1 XM_017026075.1 XM_017026076.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3526908 c.1547G>A p.R516Q Chr.18: 5969487-5969487 on Build GRCh38
1719342 c.1558C>T p.L520F Chr.18: 5969476-5969476 on Build GRCh38
3937773 c.1494G>T p.Q498H Chr.18: 5969540-5969540 on Build GRCh38
223920 c.1517C>T p.S506F Chr.18: 5969517-5969517 on Build GRCh38
3821634 c.1565G>A p.R522K Chr.18: 5969469-5969469 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs909504537 Chr.18: 5969502-5969502 on Build GRCh38
rs200212638 Chr.18: 5969561-5969561 on Build GRCh38
rs959197827 Chr.18: 5969729-5969729 on Build GRCh38
rs745638592 Chr.18: 5969576-5969576 on Build GRCh38
rs759713659 Chr.18: 5969519-5969519 on Build GRCh38
rs779809285 Chr.18: 5969499-5969499 on Build GRCh38
rs769217376 Chr.18: 5969355-5969355 on Build GRCh38
rs895286750 Chr.18: 5969733-5969733 on Build GRCh38
rs754000455 Chr.18: 5969425-5969425 on Build GRCh38
rs948156329 Chr.18: 5969770-5969770 on Build GRCh38
rs775594346 Chr.18: 5969525-5969525 on Build GRCh38
rs750618760 Chr.18: 5969410-5969410 on Build GRCh38
rs372269355 Chr.18: 5969556-5969556 on Build GRCh38
rs761779632 Chr.18: 5969461-5969461 on Build GRCh38
rs772993353 Chr.18: 5969513-5969513 on Build GRCh38
rs943549954 Chr.18: 5969639-5969639 on Build GRCh38
rs373197727 Chr.18: 5969473-5969473 on Build GRCh38
rs530335558 Chr.18: 5969441-5969441 on Build GRCh38
rs371317063 Chr.18: 5969532-5969532 on Build GRCh38
rs779463960 Chr.18: 5969433-5969433 on Build GRCh38
rs898090328 Chr.18: 5969428-5969428 on Build GRCh38
rs1034033082 Chr.18: 5969714-5969714 on Build GRCh38
rs762517126 Chr.18: 5969469-5969469 on Build GRCh38
rs950073320 Chr.18: 5969384-5969384 on Build GRCh38
rs61756685 Chr.18: 5969583-5969583 on Build GRCh38
rs377549116 Chr.18: 5969439-5969439 on Build GRCh38
rs770789924 Chr.18: 5969569-5969569 on Build GRCh38
rs767710721 Chr.18: 5969538-5969538 on Build GRCh38
rs575718738 Chr.18: 5969710-5969710 on Build GRCh38
rs778689011 Chr.18: 5969552-5969552 on Build GRCh38
rs369372793 Chr.18: 5969362-5969362 on Build GRCh38
rs373941763 Chr.18: 5969508-5969508 on Build GRCh38
rs755021468 Chr.18: 5969427-5969427 on Build GRCh38
rs748870398 Chr.18: 5969453-5969453 on Build GRCh38
rs751992714 Chr.18: 5969535-5969535 on Build GRCh38
rs367551777 Chr.18: 5969581-5969581 on Build GRCh38
rs779539304 Chr.18: 5969444-5969444 on Build GRCh38
rs762527333 Chr.18: 5969388-5969388 on Build GRCh38
rs926491181 Chr.18: 5969335-5969335 on Build GRCh38
rs745457257 Chr.18: 5969354-5969354 on Build GRCh38
rs774923684 Chr.18: 5969582-5969582 on Build GRCh38
rs147435588 Chr.18: 5969409-5969409 on Build GRCh38
rs778529742 Chr.18: 5969507-5969507 on Build GRCh38
rs973038857 Chr.18: 5969695-5969695 on Build GRCh38
rs775682221 Chr.18: 5969577-5969577 on Build GRCh38
rs1056560868 Chr.18: 5969728-5969728 on Build GRCh38
rs201894188 Chr.18: 5969543-5969543 on Build GRCh38
rs773014368 Chr.18: 5969467-5969467 on Build GRCh38
rs8097452 Chr.18: 5969656-5969656 on Build GRCh38
rs558633711 Chr.18: 5969708-5969708 on Build GRCh38
rs770619603 Chr.18: 5969516-5969516 on Build GRCh38
rs753234283 Chr.18: 5969483-5969483 on Build GRCh38
rs373461607 Chr.18: 5969413-5969413 on Build GRCh38
rs752280912 Chr.18: 5969613-5969613 on Build GRCh38
rs187077179 Chr.18: 5969696-5969696 on Build GRCh38
rs368389000 Chr.18: 5969517-5969517 on Build GRCh38
rs201367172 Chr.18: 5969544-5969544 on Build GRCh38
rs558304288 Chr.18: 5969635-5969635 on Build GRCh38
rs377084361 Chr.18: 5969401-5969401 on Build GRCh38
rs139803585 Chr.18: 5969742-5969742 on Build GRCh38
rs374219031 Chr.18: 5969536-5969536 on Build GRCh38
rs202182210 Chr.18: 5969457-5969457 on Build GRCh38
rs748888746 Chr.18: 5969500-5969500 on Build GRCh38
rs757556292 Chr.18: 5969488-5969488 on Build GRCh38
rs909567276 Chr.18: 5969680-5969680 on Build GRCh38
rs768290365 Chr.18: 5969506-5969506 on Build GRCh38
rs917229606 Chr.18: 5969377-5969377 on Build GRCh38
rs370465816 Chr.18: 5969487-5969487 on Build GRCh38
rs201350966 Chr.18: 5969440-5969440 on Build GRCh38
rs182439193 Chr.18: 5969685-5969685 on Build GRCh38
rs967956699 Chr.18: 5969509-5969509 on Build GRCh38
rs901587010 Chr.18: 5969674-5969674 on Build GRCh38
rs757164290 Chr.18: 5969636-5969636 on Build GRCh38
rs756520091 Chr.18: 5969605-5969605 on Build GRCh38
rs763149251 Chr.18: 5969578-5969578 on Build GRCh38
rs761754785 Chr.18: 5969404-5969404 on Build GRCh38
rs757435198 Chr.18: 5969640-5969640 on Build GRCh38
rs765321604 Chr.18: 5969520-5969520 on Build GRCh38
rs537703926 Chr.18: 5969549-5969549 on Build GRCh38
rs759219730 Chr.18: 5969707-5969707 on Build GRCh38
rs373702353 Chr.18: 5969608-5969608 on Build GRCh38
rs937424283 Chr.18: 5969338-5969338 on Build GRCh38
rs762907322 Chr.18: 5969528-5969528 on Build GRCh38
rs375581509 Chr.18: 5969555-5969555 on Build GRCh38
rs776360548 Chr.18: 5969574-5969574 on Build GRCh38
rs746864758 Chr.18: 5969565-5969565 on Build GRCh38
rs199776978 Chr.18: 5969593-5969593 on Build GRCh38
rs760586941 Chr.18: 5969514-5969514 on Build GRCh38
rs765233692 Chr.18: 5969482-5969482 on Build GRCh38
rs201731549 Chr.18: 5969485-5969485 on Build GRCh38
rs771797079 Chr.18: 5969462-5969462 on Build GRCh38
rs369240470 Chr.18: 5969602-5969602 on Build GRCh38
rs372242423 Chr.18: 5969474-5969474 on Build GRCh38
rs373517552 Chr.18: 5969399-5969399 on Build GRCh38
rs375609427 Chr.18: 5969477-5969477 on Build GRCh38
rs776659783 Chr.18: 5969583-5969587 on Build GRCh38

View Full Product Details