Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 11852488-11853002 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHMP1B
Entrez Gene ID
57132
Gene Name
charged multivesicular body protein 1B
Gene Aliases
C10orf2, C18-ORF2, C18orf2, CHMP1.5, Vps46-2, Vps46B, hVps46-2
UniGene
Hs.656244
Gene Chromosome Location
Chr.18: 11851390-11854449 on Build GRCh38
Transcripts
NM_020412.4 NM_001142339.2 NM_001261443.1 NM_182978.3

Gene Symbol
GNAL
Entrez Gene ID
2774
Gene Name
G protein subunit alpha L
Gene Aliases
DYT25
UniGene
Hs.136295
Gene Chromosome Location
Chr.18: 11688956-11885685 on Build GRCh38
Transcripts
NM_020412.4 NM_001142339.2 NM_001261443.1 NM_182978.3

SNP Detail

SNP ID Chromosome Location
rs1056305935 Chr.18: 11852806-11852806 on Build GRCh38
rs1007728488 Chr.18: 11852733-11852734 on Build GRCh38
rs984844149 Chr.18: 11852781-11852781 on Build GRCh38
rs1022369070 Chr.18: 11852689-11852689 on Build GRCh38
rs756419024 Chr.18: 11852836-11852836 on Build GRCh38
rs1009948217 Chr.18: 11852770-11852770 on Build GRCh38
rs987576882 Chr.18: 11852554-11852554 on Build GRCh38
rs778869754 Chr.18: 11852614-11852617 on Build GRCh38
rs965286732 Chr.18: 11852940-11852940 on Build GRCh38
rs756872862 Chr.18: 11852875-11852876 on Build GRCh38
rs1049514462 Chr.18: 11852597-11852597 on Build GRCh38
rs188714622 Chr.18: 11852737-11852737 on Build GRCh38
rs553823573 Chr.18: 11852570-11852570 on Build GRCh38
rs1017841468 Chr.18: 11852892-11852892 on Build GRCh38
rs113786915 Chr.18: 11852803-11852803 on Build GRCh38
rs1017710305 Chr.18: 11852909-11852909 on Build GRCh38
rs536357745 Chr.18: 11852930-11852930 on Build GRCh38
rs905002176 Chr.18: 11852619-11852619 on Build GRCh38
rs1009651297 Chr.18: 11852513-11852513 on Build GRCh38
rs371563566 Chr.18: 11852974-11852974 on Build GRCh38
rs964918523 Chr.18: 11852586-11852586 on Build GRCh38
rs916964680 Chr.18: 11852691-11852691 on Build GRCh38
rs997681211 Chr.18: 11852953-11852953 on Build GRCh38
rs1054818893 Chr.18: 11852752-11852752 on Build GRCh38
rs568567690 Chr.18: 11852812-11852815 on Build GRCh38
rs372421623 Chr.18: 11852725-11852725 on Build GRCh38
rs892189295 Chr.18: 11852683-11852685 on Build GRCh38
rs950860653 Chr.18: 11852771-11852771 on Build GRCh38
rs143323198 Chr.18: 11852517-11852517 on Build GRCh38
rs941267886 Chr.18: 11852617-11852617 on Build GRCh38
rs902085598 Chr.18: 11852945-11852945 on Build GRCh38
rs368920612 Chr.18: 11852788-11852788 on Build GRCh38
rs867794493 Chr.18: 11852686-11852686 on Build GRCh38
rs939095927 Chr.18: 11852646-11852646 on Build GRCh38
rs1017743186 Chr.18: 11852577-11852577 on Build GRCh38
rs74815075 Chr.18: 11852624-11852624 on Build GRCh38
rs141499690 Chr.18: 11852625-11852627 on Build GRCh38
rs114918245 Chr.18: 11852744-11852744 on Build GRCh38
rs767611648 Chr.18: 11852616-11852617 on Build GRCh38
rs980655073 Chr.18: 11852800-11852800 on Build GRCh38
rs536555807 Chr.18: 11852706-11852706 on Build GRCh38
rs893799835 Chr.18: 11852760-11852760 on Build GRCh38
rs983147647 Chr.18: 11852665-11852665 on Build GRCh38
rs762465286 Chr.18: 11852601-11852601 on Build GRCh38
rs77078320 Chr.18: 11852625-11852625 on Build GRCh38
rs1044929812 Chr.18: 11852618-11852618 on Build GRCh38
rs538233539 Chr.18: 11852535-11852535 on Build GRCh38
rs903494629 Chr.18: 11852871-11852871 on Build GRCh38
rs929000579 Chr.18: 11852660-11852660 on Build GRCh38
rs954799589 Chr.18: 11852541-11852541 on Build GRCh38
rs1007779292 Chr.18: 11852890-11852890 on Build GRCh38
rs945084465 Chr.18: 11852842-11852842 on Build GRCh38
rs376716959 Chr.18: 11852784-11852784 on Build GRCh38
rs578065718 Chr.18: 11852754-11852754 on Build GRCh38
rs1026892745 Chr.18: 11852958-11852958 on Build GRCh38
rs527932871 Chr.18: 11852932-11852932 on Build GRCh38
rs560125252 Chr.18: 11852926-11852926 on Build GRCh38
rs974087229 Chr.18: 11852588-11852588 on Build GRCh38
rs544967754 Chr.18: 11852801-11852801 on Build GRCh38
rs1044221874 Chr.18: 11852741-11852741 on Build GRCh38
rs199858817 Chr.18: 11852613-11852613 on Build GRCh38
rs750357952 Chr.18: 11852591-11852592 on Build GRCh38
rs879017184 Chr.18: 11852615-11852616 on Build GRCh38
rs774928325 Chr.18: 11852598-11852598 on Build GRCh38
rs59761845 Chr.18: 11852626-11852627 on Build GRCh38
rs201893984 Chr.18: 11852615-11852615 on Build GRCh38
rs183254172 Chr.18: 11852527-11852527 on Build GRCh38
rs556362341 Chr.18: 11852750-11852750 on Build GRCh38
rs756038955 Chr.18: 11852615-11852617 on Build GRCh38
rs939190806 Chr.18: 11852803-11852805 on Build GRCh38
rs1043400628 Chr.18: 11852860-11852860 on Build GRCh38
rs565943555 Chr.18: 11852606-11852606 on Build GRCh38
rs145799015 Chr.18: 11852614-11852615 on Build GRCh38

View Full Product Details