Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 23524052-23524553 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C18orf8
Entrez Gene ID
29919
Gene Name
chromosome 18 open reading frame 8
Gene Aliases
HsT2591, MIC1, Mic-1
UniGene
Hs.464779
Gene Chromosome Location
Chr.18: 23503470-23531807 on Build GRCh38
Transcripts
NM_001276342.1 NM_001318707.1 NM_001318708.1 NM_001318709.1 NM_013326.4 XM_011525939.1 XM_017025717.1 XM_005258277.1 XM_005258279.2 XM_006722479.3 XM_011526015.1 XM_017025787.1

Gene Symbol
NPC1
Entrez Gene ID
4864
Gene Name
NPC intracellular cholesterol transporter 1
Gene Aliases
NPC
UniGene
Hs.464779
Gene Chromosome Location
Chr.18: 23506184-23586617 on Build GRCh38
Transcripts
NM_001276342.1 NM_001318707.1 NM_001318708.1 NM_001318709.1 NM_013326.4 XM_011525939.1 XM_017025717.1 XM_005258277.1 XM_005258279.2 XM_006722479.3 XM_011526015.1 XM_017025787.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4429186 c.1041C>T p.I347I Chr.18: 23524463-23524463 on Build GRCh38
5387884 c.986C>T p.S329F Chr.18: 23524154-23524154 on Build GRCh38
4931272 c.1014A>G p.S338S Chr.18: 23524436-23524436 on Build GRCh38
987079 c.1050C>T p.S350S Chr.18: 23524472-23524472 on Build GRCh38
987078 c.1047C>T p.I349I Chr.18: 23524469-23524469 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760432686 Chr.18: 23524505-23524505 on Build GRCh38
rs911347192 Chr.18: 23524249-23524249 on Build GRCh38
rs576588447 Chr.18: 23524275-23524275 on Build GRCh38
rs115130680 Chr.18: 23524270-23524270 on Build GRCh38
rs748824696 Chr.18: 23524177-23524177 on Build GRCh38
rs374093556 Chr.18: 23524367-23524367 on Build GRCh38
rs148467554 Chr.18: 23524186-23524186 on Build GRCh38
rs150540565 Chr.18: 23524357-23524357 on Build GRCh38
rs774042832 Chr.18: 23524179-23524179 on Build GRCh38
rs938103624 Chr.18: 23524202-23524202 on Build GRCh38
rs753032782 Chr.18: 23524215-23524215 on Build GRCh38
rs766817459 Chr.18: 23524197-23524197 on Build GRCh38
rs764249235 Chr.18: 23524499-23524499 on Build GRCh38
rs771637617 Chr.18: 23524417-23524417 on Build GRCh38
rs747367515 Chr.18: 23524424-23524424 on Build GRCh38
rs763089805 Chr.18: 23524219-23524219 on Build GRCh38
rs8084285 Chr.18: 23524335-23524335 on Build GRCh38
rs778395007 Chr.18: 23524389-23524389 on Build GRCh38
rs911401933 Chr.18: 23524474-23524474 on Build GRCh38
rs1053868734 Chr.18: 23524321-23524321 on Build GRCh38
rs144205739 Chr.18: 23524506-23524506 on Build GRCh38
rs750204096 Chr.18: 23524366-23524366 on Build GRCh38
rs531155366 Chr.18: 23524101-23524101 on Build GRCh38
rs917990007 Chr.18: 23524460-23524460 on Build GRCh38
rs759077540 Chr.18: 23524423-23524424 on Build GRCh38
rs140268783 Chr.18: 23524391-23524391 on Build GRCh38
rs959378304 Chr.18: 23524406-23524406 on Build GRCh38
rs780844044 Chr.18: 23524140-23524140 on Build GRCh38
rs777435331 Chr.18: 23524423-23524423 on Build GRCh38
rs201899092 Chr.18: 23524199-23524199 on Build GRCh38
rs747787855 Chr.18: 23524180-23524180 on Build GRCh38
rs201465238 Chr.18: 23524492-23524492 on Build GRCh38
rs780004220 Chr.18: 23524376-23524376 on Build GRCh38
rs371188093 Chr.18: 23524143-23524143 on Build GRCh38
rs949206251 Chr.18: 23524272-23524272 on Build GRCh38
rs147982431 Chr.18: 23524461-23524461 on Build GRCh38
rs754543959 Chr.18: 23524387-23524387 on Build GRCh38
rs569720427 Chr.18: 23524501-23524501 on Build GRCh38
rs141623982 Chr.18: 23524472-23524472 on Build GRCh38
rs891385 Chr.18: 23524187-23524187 on Build GRCh38
rs1030061975 Chr.18: 23524329-23524329 on Build GRCh38
rs76115546 Chr.18: 23524514-23524514 on Build GRCh38
rs1008392023 Chr.18: 23524339-23524339 on Build GRCh38
rs766843454 Chr.18: 23524085-23524085 on Build GRCh38
rs979727154 Chr.18: 23524082-23524082 on Build GRCh38
rs775724739 Chr.18: 23524478-23524478 on Build GRCh38
rs753755942 Chr.18: 23524381-23524381 on Build GRCh38
rs752944530 Chr.18: 23524112-23524112 on Build GRCh38
rs377625484 Chr.18: 23524331-23524331 on Build GRCh38
rs768120449 Chr.18: 23524178-23524178 on Build GRCh38
rs973383045 Chr.18: 23524448-23524448 on Build GRCh38
rs554160472 Chr.18: 23524121-23524121 on Build GRCh38
rs774004980 Chr.18: 23524227-23524228 on Build GRCh38
rs771296956 Chr.18: 23524426-23524426 on Build GRCh38
rs377544931 Chr.18: 23524356-23524356 on Build GRCh38
rs375981319 Chr.18: 23524099-23524099 on Build GRCh38
rs3745022 Chr.18: 23524319-23524319 on Build GRCh38
rs560591661 Chr.18: 23524361-23524361 on Build GRCh38
rs200566715 Chr.18: 23524453-23524453 on Build GRCh38
rs1042317243 Chr.18: 23524282-23524282 on Build GRCh38
rs765236943 Chr.18: 23524311-23524311 on Build GRCh38
rs766981646 Chr.18: 23524503-23524505 on Build GRCh38
rs996344205 Chr.18: 23524328-23524328 on Build GRCh38
rs761042685 Chr.18: 23524195-23524195 on Build GRCh38
rs765392851 Chr.18: 23524200-23524200 on Build GRCh38
rs572406475 Chr.18: 23524322-23524322 on Build GRCh38
rs755061519 Chr.18: 23524102-23524102 on Build GRCh38
rs902432633 Chr.18: 23524310-23524310 on Build GRCh38
rs370329557 Chr.18: 23524109-23524109 on Build GRCh38
rs143803533 Chr.18: 23524141-23524141 on Build GRCh38
rs753841760 Chr.18: 23524511-23524511 on Build GRCh38
rs777238935 Chr.18: 23524198-23524198 on Build GRCh38
rs759000250 Chr.18: 23524254-23524254 on Build GRCh38
rs779988837 Chr.18: 23524144-23524144 on Build GRCh38
rs765954710 Chr.18: 23524116-23524117 on Build GRCh38

View Full Product Details