Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 31645163-31645694 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B4GALT6
Entrez Gene ID
9331
Gene Name
beta-1,4-galactosyltransferase 6
Gene Aliases
B4Gal-T6, beta4Gal-T6
UniGene
Hs.591063
Gene Chromosome Location
Chr.18: 31622246-31724670 on Build GRCh38
Transcripts
NM_004775.3 XM_005258387.4 XM_006722578.3 XM_006722579.3 XM_017026088.1 XM_017026089.1 XM_017026090.1 XM_017026091.1 XM_017026092.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4153946 c.388G>T p.E130* Chr.18: 31645438-31645438 on Build GRCh38
6465371 c.394C>T p.H132Y Chr.18: 31645432-31645432 on Build GRCh38
220171 c.372C>G p.S124R Chr.18: 31645454-31645454 on Build GRCh38
220147 c.386A>T p.D129V Chr.18: 31645440-31645440 on Build GRCh38
401762 c.372C>A p.S124R Chr.18: 31645454-31645454 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746472355 Chr.18: 31645454-31645454 on Build GRCh38
rs771517633 Chr.18: 31645458-31645458 on Build GRCh38
rs771159523 Chr.18: 31645601-31645602 on Build GRCh38
rs939192644 Chr.18: 31645385-31645385 on Build GRCh38
rs976869639 Chr.18: 31645227-31645227 on Build GRCh38
rs1029791702 Chr.18: 31645556-31645556 on Build GRCh38
rs980751782 Chr.18: 31645657-31645657 on Build GRCh38
rs199941208 Chr.18: 31645408-31645408 on Build GRCh38
rs146313105 Chr.18: 31645317-31645317 on Build GRCh38
rs751697628 Chr.18: 31645501-31645501 on Build GRCh38
rs547780069 Chr.18: 31645627-31645627 on Build GRCh38
rs370200838 Chr.18: 31645488-31645488 on Build GRCh38
rs779821972 Chr.18: 31645419-31645419 on Build GRCh38
rs557412068 Chr.18: 31645513-31645514 on Build GRCh38
rs769009688 Chr.18: 31645557-31645557 on Build GRCh38
rs998341121 Chr.18: 31645550-31645550 on Build GRCh38
rs752963055 Chr.18: 31645376-31645376 on Build GRCh38
rs760012180 Chr.18: 31645467-31645467 on Build GRCh38
rs926517033 Chr.18: 31645384-31645384 on Build GRCh38
rs944346632 Chr.18: 31645275-31645275 on Build GRCh38
rs988651291 Chr.18: 31645616-31645616 on Build GRCh38
rs771596136 Chr.18: 31645439-31645439 on Build GRCh38
rs1056309124 Chr.18: 31645399-31645399 on Build GRCh38
rs369396189 Chr.18: 31645366-31645366 on Build GRCh38
rs922727567 Chr.18: 31645246-31645246 on Build GRCh38
rs117060609 Chr.18: 31645620-31645620 on Build GRCh38
rs956777746 Chr.18: 31645278-31645278 on Build GRCh38
rs201760528 Chr.18: 31645503-31645503 on Build GRCh38
rs535745238 Chr.18: 31645661-31645661 on Build GRCh38
rs145403140 Chr.18: 31645355-31645355 on Build GRCh38
rs956854005 Chr.18: 31645605-31645605 on Build GRCh38
rs763129897 Chr.18: 31645389-31645389 on Build GRCh38
rs983121851 Chr.18: 31645193-31645193 on Build GRCh38
rs911648800 Chr.18: 31645244-31645244 on Build GRCh38
rs753712194 Chr.18: 31645421-31645421 on Build GRCh38
rs778544529 Chr.18: 31645427-31645427 on Build GRCh38
rs915228103 Chr.18: 31645340-31645340 on Build GRCh38
rs771232882 Chr.18: 31645342-31645342 on Build GRCh38
rs540377067 Chr.18: 31645346-31645346 on Build GRCh38
rs1045401991 Chr.18: 31645373-31645373 on Build GRCh38
rs756035811 Chr.18: 31645412-31645412 on Build GRCh38
rs562425613 Chr.18: 31645548-31645548 on Build GRCh38
rs776990971 Chr.18: 31645461-31645461 on Build GRCh38
rs781424223 Chr.18: 31645611-31645611 on Build GRCh38
rs764209448 Chr.18: 31645398-31645398 on Build GRCh38
rs747742960 Chr.18: 31645436-31645436 on Build GRCh38
rs114543734 Chr.18: 31645424-31645424 on Build GRCh38
rs373090639 Chr.18: 31645453-31645453 on Build GRCh38
rs141485306 Chr.18: 31645650-31645650 on Build GRCh38
rs921640460 Chr.18: 31645336-31645336 on Build GRCh38
rs139156163 Chr.18: 31645489-31645489 on Build GRCh38
rs763216864 Chr.18: 31645493-31645493 on Build GRCh38
rs764297278 Chr.18: 31645497-31645497 on Build GRCh38
rs746879761 Chr.18: 31645370-31645371 on Build GRCh38
rs962778985 Chr.18: 31645537-31645537 on Build GRCh38

View Full Product Details