Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 63640792-63641300 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SERPINB4
Entrez Gene ID
6318
Gene Name
serpin family B member 4
Gene Aliases
LEUPIN, PI11, SCCA-2, SCCA1, SCCA2
UniGene
Hs.123035
Gene Chromosome Location
Chr.18: 63637259-63644319 on Build GRCh38
Transcripts
NM_002974.3 NM_175041.1 XM_011526138.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3959330 c.469+5G>C p.? Chr.18: 63640869-63640869 on Build GRCh38
7301139 c.367A>G p.I123V Chr.18: 63640976-63640976 on Build GRCh38
6211202 c.364G>T p.A122S Chr.18: 63640979-63640979 on Build GRCh38
473980 c.366C>T p.A122A Chr.18: 63640977-63640977 on Build GRCh38
6298309 c.469+10C>A p.? Chr.18: 63640864-63640864 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146103700 Chr.18: 63641141-63641141 on Build GRCh38
rs754077340 Chr.18: 63641033-63641034 on Build GRCh38
rs762497510 Chr.18: 63640992-63640992 on Build GRCh38
rs958385111 Chr.18: 63641268-63641268 on Build GRCh38
rs780407516 Chr.18: 63640916-63640916 on Build GRCh38
rs768559162 Chr.18: 63640835-63640835 on Build GRCh38
rs754186109 Chr.18: 63641004-63641004 on Build GRCh38
rs34183895 Chr.18: 63641061-63641062 on Build GRCh38
rs375182220 Chr.18: 63641114-63641114 on Build GRCh38
rs773616722 Chr.18: 63640867-63640867 on Build GRCh38
rs775341950 Chr.18: 63640990-63640990 on Build GRCh38
rs967298435 Chr.18: 63641068-63641068 on Build GRCh38
rs201629395 Chr.18: 63640912-63640912 on Build GRCh38
rs139267790 Chr.18: 63640999-63640999 on Build GRCh38
rs571728294 Chr.18: 63640925-63640925 on Build GRCh38
rs879433035 Chr.18: 63641121-63641121 on Build GRCh38
rs769759872 Chr.18: 63640985-63640985 on Build GRCh38
rs773105953 Chr.18: 63640817-63640817 on Build GRCh38
rs912249361 Chr.18: 63641108-63641108 on Build GRCh38
rs778102288 Chr.18: 63640956-63640956 on Build GRCh38
rs772083334 Chr.18: 63640934-63640934 on Build GRCh38
rs931467117 Chr.18: 63641161-63641161 on Build GRCh38
rs759318436 Chr.18: 63640982-63640982 on Build GRCh38
rs113035765 Chr.18: 63641178-63641178 on Build GRCh38
rs748242356 Chr.18: 63640846-63640846 on Build GRCh38
rs7234633 Chr.18: 63641061-63641061 on Build GRCh38
rs780495304 Chr.18: 63640828-63640828 on Build GRCh38
rs575335103 Chr.18: 63641148-63641148 on Build GRCh38
rs764588249 Chr.18: 63640888-63640888 on Build GRCh38
rs772324861 Chr.18: 63640854-63640854 on Build GRCh38
rs200851651 Chr.18: 63640875-63640875 on Build GRCh38
rs770192003 Chr.18: 63641069-63641069 on Build GRCh38
rs191670636 Chr.18: 63640822-63640822 on Build GRCh38
rs766309263 Chr.18: 63640870-63640870 on Build GRCh38
rs767493208 Chr.18: 63640813-63640813 on Build GRCh38
rs1040255038 Chr.18: 63640857-63640857 on Build GRCh38
rs776662515 Chr.18: 63640874-63640874 on Build GRCh38
rs748237263 Chr.18: 63640930-63640930 on Build GRCh38
rs374647211 Chr.18: 63640878-63640878 on Build GRCh38
rs755345143 Chr.18: 63641017-63641017 on Build GRCh38
rs760824171 Chr.18: 63640819-63640819 on Build GRCh38
rs1039463889 Chr.18: 63641147-63641147 on Build GRCh38
rs374969261 Chr.18: 63640840-63640840 on Build GRCh38
rs755296444 Chr.18: 63641246-63641246 on Build GRCh38
rs529909736 Chr.18: 63641222-63641222 on Build GRCh38
rs1013863439 Chr.18: 63641191-63641191 on Build GRCh38
rs150896887 Chr.18: 63640868-63640868 on Build GRCh38
rs749188625 Chr.18: 63640904-63640906 on Build GRCh38
rs980880247 Chr.18: 63641002-63641002 on Build GRCh38
rs778014417 Chr.18: 63641033-63641033 on Build GRCh38
rs113197355 Chr.18: 63641212-63641212 on Build GRCh38
rs542629069 Chr.18: 63641258-63641258 on Build GRCh38
rs776574301 Chr.18: 63640967-63640967 on Build GRCh38
rs373177927 Chr.18: 63641039-63641039 on Build GRCh38
rs149979278 Chr.18: 63641024-63641024 on Build GRCh38
rs140278212 Chr.18: 63640899-63640899 on Build GRCh38
rs536169238 Chr.18: 63641208-63641208 on Build GRCh38
rs778129627 Chr.18: 63641167-63641167 on Build GRCh38
rs763611615 Chr.18: 63640996-63640996 on Build GRCh38
rs201970247 Chr.18: 63640921-63640921 on Build GRCh38
rs199998776 Chr.18: 63640894-63640894 on Build GRCh38
rs1025893643 Chr.18: 63640881-63640881 on Build GRCh38
rs779444600 Chr.18: 63641018-63641018 on Build GRCh38
rs145373357 Chr.18: 63640913-63640913 on Build GRCh38
rs531514914 Chr.18: 63641134-63641134 on Build GRCh38
rs140074412 Chr.18: 63641190-63641190 on Build GRCh38
rs188855628 Chr.18: 63641213-63641213 on Build GRCh38
rs761456422 Chr.18: 63640998-63640998 on Build GRCh38
rs752250571 Chr.18: 63640893-63640893 on Build GRCh38
rs753176216 Chr.18: 63641019-63641019 on Build GRCh38
rs887328034 Chr.18: 63641171-63641171 on Build GRCh38
rs573006804 Chr.18: 63641106-63641106 on Build GRCh38
rs1024993179 Chr.18: 63640919-63640919 on Build GRCh38
rs771276876 Chr.18: 63641041-63641041 on Build GRCh38
rs537798593 Chr.18: 63640964-63640964 on Build GRCh38
rs779344580 Chr.18: 63640926-63640926 on Build GRCh38
rs779713662 Chr.18: 63640876-63640905 on Build GRCh38
rs183407670 Chr.18: 63640997-63640997 on Build GRCh38
rs202067086 Chr.18: 63640838-63640838 on Build GRCh38
rs749814353 Chr.18: 63640829-63640829 on Build GRCh38
rs747257812 Chr.18: 63640963-63640963 on Build GRCh38

View Full Product Details