Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 74155063-74155573 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TIMM21
Entrez Gene ID
29090
Gene Name
translocase of inner mitochondrial membrane 21
Gene Aliases
C18orf55, HSPC154, TIM21
UniGene
Hs.532835
Gene Chromosome Location
Chr.18: 74148511-74158969 on Build GRCh38
Transcripts
NM_014177.2 XM_005266689.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6364270 c.401C>G p.S134C Chr.18: 74155342-74155342 on Build GRCh38
4073210 c.447C>T p.C149C Chr.18: 74155388-74155388 on Build GRCh38
989862 c.343C>A p.L115I Chr.18: 74155186-74155186 on Build GRCh38
4073209 c.424T>C p.Y142H Chr.18: 74155365-74155365 on Build GRCh38
4073208 c.318A>C p.R106S Chr.18: 74155161-74155161 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755184007 Chr.18: 74155207-74155207 on Build GRCh38
rs762857996 Chr.18: 74155089-74155089 on Build GRCh38
rs776792558 Chr.18: 74155289-74155289 on Build GRCh38
rs115074506 Chr.18: 74155459-74155459 on Build GRCh38
rs1055844264 Chr.18: 74155084-74155084 on Build GRCh38
rs200031543 Chr.18: 74155309-74155309 on Build GRCh38
rs768815142 Chr.18: 74155177-74155178 on Build GRCh38
rs772533136 Chr.18: 74155277-74155277 on Build GRCh38
rs183718529 Chr.18: 74155109-74155109 on Build GRCh38
rs567722365 Chr.18: 74155200-74155200 on Build GRCh38
rs762034862 Chr.18: 74155300-74155300 on Build GRCh38
rs754001060 Chr.18: 74155205-74155205 on Build GRCh38
rs976532811 Chr.18: 74155441-74155441 on Build GRCh38
rs538663883 Chr.18: 74155519-74155519 on Build GRCh38
rs764240675 Chr.18: 74155445-74155445 on Build GRCh38
rs374248144 Chr.18: 74155444-74155444 on Build GRCh38
rs764016839 Chr.18: 74155332-74155332 on Build GRCh38
rs781015726 Chr.18: 74155363-74155363 on Build GRCh38
rs769390261 Chr.18: 74155267-74155267 on Build GRCh38
rs745591348 Chr.18: 74155124-74155124 on Build GRCh38
rs928622740 Chr.18: 74155537-74155537 on Build GRCh38
rs763928775 Chr.18: 74155099-74155099 on Build GRCh38
rs755629945 Chr.18: 74155398-74155398 on Build GRCh38
rs755658553 Chr.18: 74155367-74155367 on Build GRCh38
rs751319359 Chr.18: 74155344-74155344 on Build GRCh38
rs968687839 Chr.18: 74155393-74155393 on Build GRCh38
rs982436865 Chr.18: 74155465-74155465 on Build GRCh38
rs373918903 Chr.18: 74155116-74155116 on Build GRCh38
rs761310400 Chr.18: 74155282-74155282 on Build GRCh38
rs202045787 Chr.18: 74155296-74155296 on Build GRCh38
rs147896042 Chr.18: 74155237-74155237 on Build GRCh38
rs767969197 Chr.18: 74155156-74155156 on Build GRCh38
rs148956058 Chr.18: 74155345-74155345 on Build GRCh38
rs751324829 Chr.18: 74155446-74155446 on Build GRCh38
rs774643346 Chr.18: 74155130-74155130 on Build GRCh38
rs942333359 Chr.18: 74155528-74155528 on Build GRCh38
rs200969509 Chr.18: 74155285-74155285 on Build GRCh38
rs569672012 Chr.18: 74155102-74155102 on Build GRCh38
rs773574473 Chr.18: 74155278-74155278 on Build GRCh38
rs756718833 Chr.18: 74155231-74155231 on Build GRCh38
rs769445468 Chr.18: 74155125-74155125 on Build GRCh38
rs748994093 Chr.18: 74155380-74155380 on Build GRCh38
rs200901555 Chr.18: 74155114-74155114 on Build GRCh38
rs774540480 Chr.18: 74155212-74155213 on Build GRCh38
rs1057239314 Chr.18: 74155186-74155186 on Build GRCh38
rs747347327 Chr.18: 74155389-74155389 on Build GRCh38
rs187799098 Chr.18: 74155418-74155418 on Build GRCh38
rs1028107078 Chr.18: 74155378-74155378 on Build GRCh38
rs775851315 Chr.18: 74155442-74155442 on Build GRCh38
rs371261111 Chr.18: 74155218-74155218 on Build GRCh38
rs373827582 Chr.18: 74155340-74155340 on Build GRCh38
rs571658975 Chr.18: 74155544-74155544 on Build GRCh38
rs569527724 Chr.18: 74155470-74155470 on Build GRCh38
rs758310130 Chr.18: 74155223-74155223 on Build GRCh38
rs561143789 Chr.18: 74155409-74155409 on Build GRCh38
rs549469399 Chr.18: 74155101-74155101 on Build GRCh38
rs745399899 Chr.18: 74155257-74155257 on Build GRCh38
rs775158256 Chr.18: 74155271-74155271 on Build GRCh38
rs780690796 Chr.18: 74155234-74155234 on Build GRCh38
rs762191215 Chr.18: 74155150-74155150 on Build GRCh38
rs778132407 Chr.18: 74155386-74155386 on Build GRCh38
rs544136857 Chr.18: 74155226-74155226 on Build GRCh38
rs756895979 Chr.18: 74155162-74155162 on Build GRCh38
rs201930254 Chr.18: 74155117-74155117 on Build GRCh38
rs747025822 Chr.18: 74155227-74155227 on Build GRCh38
rs199965974 Chr.18: 74155372-74155372 on Build GRCh38
rs528588522 Chr.18: 74155366-74155366 on Build GRCh38
rs760645079 Chr.18: 74155187-74155187 on Build GRCh38
rs558712228 Chr.18: 74155110-74155110 on Build GRCh38
rs748636635 Chr.18: 74155422-74155422 on Build GRCh38
rs116581614 Chr.18: 74155531-74155531 on Build GRCh38
rs772316169 Chr.18: 74155382-74155382 on Build GRCh38
rs1003848535 Chr.18: 74155306-74155307 on Build GRCh38
rs758874883 Chr.18: 74155485-74155485 on Build GRCh38
rs145075415 Chr.18: 74155321-74155321 on Build GRCh38
rs747959111 Chr.18: 74155113-74155113 on Build GRCh38
rs762828541 Chr.18: 74155330-74155330 on Build GRCh38
rs879936654 Chr.18: 74155532-74155532 on Build GRCh38
rs145254538 Chr.18: 74155123-74155123 on Build GRCh38
rs34544053 Chr.18: 74155206-74155207 on Build GRCh38
rs371309985 Chr.18: 74155184-74155184 on Build GRCh38
rs1020281964 Chr.18: 74155346-74155346 on Build GRCh38
rs3737513 Chr.18: 74155086-74155086 on Build GRCh38
rs1019701975 Chr.18: 74155312-74155312 on Build GRCh38

View Full Product Details