Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 42459883-42460417 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPCS
Entrez Gene ID
79717
Gene Name
phosphopantothenoylcysteine synthetase
Gene Aliases
-
UniGene
Hs.706662
Gene Chromosome Location
Chr.1: 42456341-42473381 on Build GRCh38
Transcripts
NM_001077447.2 NM_001287506.1 NM_001287507.1 NM_001287508.1 NM_001287509.1 NM_001287510.1 NM_001287511.1 NM_024664.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6753261 c.918T>C p.F306F Chr.1: 42459908-42459908 on Build GRCh38
5010539 c.933C>G p.N311K Chr.1: 42459923-42459923 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1025506440 Chr.1: 42460362-42460362 on Build GRCh38
rs753360321 Chr.1: 42459925-42459925 on Build GRCh38
rs936617755 Chr.1: 42460045-42460045 on Build GRCh38
rs139273454 Chr.1: 42459997-42459997 on Build GRCh38
rs960008686 Chr.1: 42460014-42460014 on Build GRCh38
rs1007973171 Chr.1: 42460188-42460188 on Build GRCh38
rs1035910938 Chr.1: 42459996-42459996 on Build GRCh38
rs1006431686 Chr.1: 42460285-42460285 on Build GRCh38
rs1011252955 Chr.1: 42460250-42460250 on Build GRCh38
rs536121182 Chr.1: 42460362-42460362 on Build GRCh38
rs746509252 Chr.1: 42459938-42459939 on Build GRCh38
rs555182356 Chr.1: 42460067-42460067 on Build GRCh38
rs760575381 Chr.1: 42459917-42459917 on Build GRCh38
rs541949803 Chr.1: 42460059-42460059 on Build GRCh38
rs368483468 Chr.1: 42459919-42459919 on Build GRCh38
rs187060100 Chr.1: 42460121-42460121 on Build GRCh38
rs745779231 Chr.1: 42460106-42460106 on Build GRCh38
rs952234267 Chr.1: 42459924-42459924 on Build GRCh38
rs762935673 Chr.1: 42459994-42459994 on Build GRCh38
rs199853153 Chr.1: 42459986-42459987 on Build GRCh38
rs564373566 Chr.1: 42460241-42460241 on Build GRCh38
rs1011369397 Chr.1: 42459985-42459985 on Build GRCh38
rs952373140 Chr.1: 42460273-42460273 on Build GRCh38
rs9787162 Chr.1: 42460017-42460017 on Build GRCh38
rs1008876005 Chr.1: 42459938-42459938 on Build GRCh38
rs72950592 Chr.1: 42460056-42460056 on Build GRCh38
rs544101357 Chr.1: 42460225-42460225 on Build GRCh38
rs1039975212 Chr.1: 42460237-42460237 on Build GRCh38
rs945409791 Chr.1: 42460007-42460007 on Build GRCh38
rs145650298 Chr.1: 42460328-42460328 on Build GRCh38
rs902956193 Chr.1: 42460294-42460294 on Build GRCh38
rs533291250 Chr.1: 42460242-42460242 on Build GRCh38
rs779561295 Chr.1: 42459936-42459936 on Build GRCh38
rs920019331 Chr.1: 42460144-42460144 on Build GRCh38
rs1042718413 Chr.1: 42460283-42460283 on Build GRCh38
rs775424687 Chr.1: 42459906-42459906 on Build GRCh38
rs1040855091 Chr.1: 42460208-42460208 on Build GRCh38
rs879216175 Chr.1: 42460094-42460094 on Build GRCh38
rs992213398 Chr.1: 42459995-42459995 on Build GRCh38
rs889630824 Chr.1: 42460159-42460159 on Build GRCh38
rs925161690 Chr.1: 42460030-42460030 on Build GRCh38
rs552577157 Chr.1: 42460026-42460026 on Build GRCh38
rs756781299 Chr.1: 42459937-42459937 on Build GRCh38
rs149975288 Chr.1: 42460151-42460151 on Build GRCh38
rs974005715 Chr.1: 42460101-42460101 on Build GRCh38
rs764998252 Chr.1: 42459964-42459964 on Build GRCh38
rs960138389 Chr.1: 42460335-42460335 on Build GRCh38
rs1012876628 Chr.1: 42460339-42460339 on Build GRCh38
rs750031368 Chr.1: 42459970-42459970 on Build GRCh38
rs899490031 Chr.1: 42459979-42459979 on Build GRCh38
rs557605002 Chr.1: 42459978-42459978 on Build GRCh38
rs896799668 Chr.1: 42460220-42460220 on Build GRCh38
rs527394053 Chr.1: 42460333-42460333 on Build GRCh38
rs944337569 Chr.1: 42460236-42460236 on Build GRCh38
rs1033777712 Chr.1: 42460292-42460292 on Build GRCh38

View Full Product Details