Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 405365-405876 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2CD4C
Entrez Gene ID
126567
Gene Name
C2 calcium dependent domain containing 4C
Gene Aliases
FAM148C, KIAA1957, NLF3
UniGene
Hs.223770
Gene Chromosome Location
Chr.19: 405435-409170 on Build GRCh38
Transcripts
NM_001136263.1 XM_011527694.1

SNP Detail

SNP ID Chromosome Location
rs1032145401 Chr.19: 405703-405703 on Build GRCh38
rs891243283 Chr.19: 405792-405792 on Build GRCh38
rs925140023 Chr.19: 405627-405627 on Build GRCh38
rs979701885 Chr.19: 405624-405624 on Build GRCh38
rs536509277 Chr.19: 405636-405636 on Build GRCh38
rs537994894 Chr.19: 405535-405535 on Build GRCh38
rs998618703 Chr.19: 405580-405580 on Build GRCh38
rs937766953 Chr.19: 405628-405628 on Build GRCh38
rs949777333 Chr.19: 405737-405737 on Build GRCh38
rs1014588493 Chr.19: 405442-405442 on Build GRCh38
rs866222472 Chr.19: 405691-405691 on Build GRCh38
rs144201691 Chr.19: 405609-405609 on Build GRCh38
rs374851541 Chr.19: 405498-405498 on Build GRCh38
rs543838094 Chr.19: 405472-405472 on Build GRCh38
rs1020940355 Chr.19: 405681-405681 on Build GRCh38
rs748183605 Chr.19: 405705-405705 on Build GRCh38
rs554060150 Chr.19: 405602-405602 on Build GRCh38
rs1000752322 Chr.19: 405690-405690 on Build GRCh38
rs1016736215 Chr.19: 405632-405632 on Build GRCh38
rs902660521 Chr.19: 405578-405578 on Build GRCh38
rs767268450 Chr.19: 405443-405443 on Build GRCh38
rs765646445 Chr.19: 405573-405573 on Build GRCh38
rs531717315 Chr.19: 405606-405606 on Build GRCh38
rs774152642 Chr.19: 405749-405749 on Build GRCh38
rs770961345 Chr.19: 405722-405722 on Build GRCh38
rs1049774452 Chr.19: 405657-405657 on Build GRCh38
rs945172937 Chr.19: 405507-405507 on Build GRCh38
rs1030190929 Chr.19: 405791-405791 on Build GRCh38
rs986746854 Chr.19: 405613-405613 on Build GRCh38
rs925045759 Chr.19: 405525-405525 on Build GRCh38
rs979220924 Chr.19: 405516-405516 on Build GRCh38
rs967061395 Chr.19: 405689-405689 on Build GRCh38
rs555937903 Chr.19: 405477-405477 on Build GRCh38
rs996940996 Chr.19: 405649-405649 on Build GRCh38
rs1061151 Chr.19: 405568-405568 on Build GRCh38
rs1021265560 Chr.19: 405617-405617 on Build GRCh38
rs567242121 Chr.19: 405510-405510 on Build GRCh38
rs970490357 Chr.19: 405724-405724 on Build GRCh38
rs1005399300 Chr.19: 405630-405630 on Build GRCh38
rs562817132 Chr.19: 405488-405488 on Build GRCh38
rs143740799 Chr.19: 405764-405764 on Build GRCh38
rs1016766924 Chr.19: 405779-405779 on Build GRCh38
rs1049811907 Chr.19: 405666-405666 on Build GRCh38
rs975441309 Chr.19: 405474-405474 on Build GRCh38
rs747260022 Chr.19: 405612-405612 on Build GRCh38
rs1001205985 Chr.19: 405810-405810 on Build GRCh38
rs920104161 Chr.19: 405785-405785 on Build GRCh38
rs1047372865 Chr.19: 405635-405635 on Build GRCh38
rs572533983 Chr.19: 405845-405845 on Build GRCh38
rs566544252 Chr.19: 405710-405710 on Build GRCh38
rs922600158 Chr.19: 405503-405503 on Build GRCh38
rs775224071 Chr.19: 405816-405816 on Build GRCh38
rs937747484 Chr.19: 405530-405530 on Build GRCh38
rs745715371 Chr.19: 405788-405788 on Build GRCh38
rs946970807 Chr.19: 405673-405673 on Build GRCh38
rs940268940 Chr.19: 405759-405759 on Build GRCh38
rs545708130 Chr.19: 405441-405441 on Build GRCh38
rs750862513 Chr.19: 405574-405574 on Build GRCh38
rs983905688 Chr.19: 405450-405450 on Build GRCh38
rs990550167 Chr.19: 405720-405720 on Build GRCh38
rs552381966 Chr.19: 405592-405592 on Build GRCh38
rs558351417 Chr.19: 405721-405721 on Build GRCh38
rs760271535 Chr.19: 405501-405501 on Build GRCh38
rs896117196 Chr.19: 405558-405559 on Build GRCh38
rs985467421 Chr.19: 405665-405665 on Build GRCh38
rs1031361720 Chr.19: 405406-405406 on Build GRCh38
rs542129190 Chr.19: 405562-405562 on Build GRCh38
rs1050471849 Chr.19: 405560-405560 on Build GRCh38
rs970329285 Chr.19: 405447-405447 on Build GRCh38
rs1038981905 Chr.19: 405638-405638 on Build GRCh38
rs986676237 Chr.19: 405500-405500 on Build GRCh38
rs894766891 Chr.19: 405421-405421 on Build GRCh38
rs7251812 Chr.19: 405634-405634 on Build GRCh38
rs189043677 Chr.19: 405637-405637 on Build GRCh38
rs558664149 Chr.19: 405405-405405 on Build GRCh38
rs911473283 Chr.19: 405564-405564 on Build GRCh38
rs34282976 Chr.19: 405541-405541 on Build GRCh38
rs574100367 Chr.19: 405795-405795 on Build GRCh38
rs898625216 Chr.19: 405639-405639 on Build GRCh38
rs897035051 Chr.19: 405787-405787 on Build GRCh38
rs911487570 Chr.19: 405668-405668 on Build GRCh38
rs1014458932 Chr.19: 405440-405440 on Build GRCh38
rs534086555 Chr.19: 405393-405393 on Build GRCh38
rs7255344 Chr.19: 405805-405805 on Build GRCh38
rs917598712 Chr.19: 405631-405631 on Build GRCh38
rs1005431890 Chr.19: 405773-405773 on Build GRCh38
rs576555255 Chr.19: 405734-405734 on Build GRCh38
rs1042786138 Chr.19: 405796-405796 on Build GRCh38
rs560436352 Chr.19: 405563-405563 on Build GRCh38
rs763623677 Chr.19: 405529-405529 on Build GRCh38
rs565089844 Chr.19: 405449-405449 on Build GRCh38
rs140083457 Chr.19: 405711-405711 on Build GRCh38
rs1061150 Chr.19: 405593-405593 on Build GRCh38
rs372453903 Chr.19: 405748-405748 on Build GRCh38
rs932780859 Chr.19: 405786-405786 on Build GRCh38
rs375629725 Chr.19: 405828-405828 on Build GRCh38
rs1052814257 Chr.19: 405586-405586 on Build GRCh38
rs1050947654 Chr.19: 405392-405392 on Build GRCh38
rs966632808 Chr.19: 405618-405618 on Build GRCh38
rs955595404 Chr.19: 405641-405641 on Build GRCh38

View Full Product Details