Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 406421-406923 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2CD4C
Entrez Gene ID
126567
Gene Name
C2 calcium dependent domain containing 4C
Gene Aliases
FAM148C, KIAA1957, NLF3
UniGene
Hs.223770
Gene Chromosome Location
Chr.19: 405435-409170 on Build GRCh38
Transcripts
NM_001136263.1 XM_011527694.1

SNP Detail

SNP ID Chromosome Location
rs1032025240 Chr.19: 406819-406819 on Build GRCh38
rs116465746 Chr.19: 406566-406566 on Build GRCh38
rs561563229 Chr.19: 406757-406757 on Build GRCh38
rs539102296 Chr.19: 406795-406795 on Build GRCh38
rs117685610 Chr.19: 406783-406783 on Build GRCh38
rs72988457 Chr.19: 406655-406655 on Build GRCh38
rs949438762 Chr.19: 406460-406460 on Build GRCh38
rs151015660 Chr.19: 406592-406592 on Build GRCh38
rs376240883 Chr.19: 406853-406854 on Build GRCh38
rs899380174 Chr.19: 406488-406488 on Build GRCh38
rs969484079 Chr.19: 406614-406614 on Build GRCh38
rs568517733 Chr.19: 406853-406853 on Build GRCh38
rs1011110948 Chr.19: 406706-406706 on Build GRCh38
rs929320171 Chr.19: 406861-406861 on Build GRCh38
rs555875217 Chr.19: 406608-406608 on Build GRCh38
rs962331727 Chr.19: 406626-406626 on Build GRCh38
rs929566607 Chr.19: 406770-406770 on Build GRCh38
rs1016431780 Chr.19: 406486-406486 on Build GRCh38
rs759976274 Chr.19: 406821-406821 on Build GRCh38
rs1019357731 Chr.19: 406672-406672 on Build GRCh38
rs965973985 Chr.19: 406871-406871 on Build GRCh38
rs1032285754 Chr.19: 406701-406701 on Build GRCh38
rs993685343 Chr.19: 406650-406650 on Build GRCh38
rs951948261 Chr.19: 406737-406737 on Build GRCh38
rs993543785 Chr.19: 406831-406831 on Build GRCh38
rs367857669 Chr.19: 406473-406473 on Build GRCh38
rs535117188 Chr.19: 406885-406885 on Build GRCh38
rs912133352 Chr.19: 406881-406881 on Build GRCh38
rs570388609 Chr.19: 406824-406824 on Build GRCh38
rs986536554 Chr.19: 406515-406515 on Build GRCh38
rs920596766 Chr.19: 406855-406855 on Build GRCh38
rs549871520 Chr.19: 406514-406514 on Build GRCh38
rs113250867 Chr.19: 406713-406713 on Build GRCh38
rs1033750080 Chr.19: 406844-406844 on Build GRCh38
rs934341903 Chr.19: 406682-406682 on Build GRCh38
rs996576325 Chr.19: 406818-406818 on Build GRCh38
rs1020870259 Chr.19: 406836-406836 on Build GRCh38
rs544062749 Chr.19: 406594-406594 on Build GRCh38
rs768361667 Chr.19: 406629-406629 on Build GRCh38
rs1037953287 Chr.19: 406630-406630 on Build GRCh38
rs1009062470 Chr.19: 406565-406565 on Build GRCh38
rs920539801 Chr.19: 406640-406640 on Build GRCh38
rs12974729 Chr.19: 406739-406739 on Build GRCh38
rs368808800 Chr.19: 406796-406796 on Build GRCh38
rs1037682703 Chr.19: 406746-406746 on Build GRCh38
rs892533326 Chr.19: 406448-406448 on Build GRCh38
rs1047934500 Chr.19: 406777-406777 on Build GRCh38
rs537760763 Chr.19: 406850-406850 on Build GRCh38
rs371939491 Chr.19: 406661-406661 on Build GRCh38
rs1046442137 Chr.19: 406609-406609 on Build GRCh38
rs981955920 Chr.19: 406843-406843 on Build GRCh38
rs963739584 Chr.19: 406676-406676 on Build GRCh38
rs946977655 Chr.19: 406792-406792 on Build GRCh38
rs999796388 Chr.19: 406591-406591 on Build GRCh38
rs1054034677 Chr.19: 406694-406694 on Build GRCh38
rs1008396037 Chr.19: 406657-406657 on Build GRCh38
rs149668702 Chr.19: 406709-406709 on Build GRCh38
rs561458787 Chr.19: 406730-406730 on Build GRCh38
rs113387360 Chr.19: 406852-406853 on Build GRCh38
rs910401812 Chr.19: 406744-406745 on Build GRCh38
rs944516342 Chr.19: 406764-406764 on Build GRCh38
rs899412758 Chr.19: 406636-406636 on Build GRCh38
rs555065687 Chr.19: 406855-406855 on Build GRCh38
rs913733337 Chr.19: 406712-406712 on Build GRCh38
rs372117619 Chr.19: 406781-406781 on Build GRCh38
rs145480797 Chr.19: 406879-406879 on Build GRCh38
rs905426705 Chr.19: 406814-406814 on Build GRCh38
rs140857937 Chr.19: 406707-406707 on Build GRCh38
rs570824372 Chr.19: 406848-406848 on Build GRCh38
rs1027654939 Chr.19: 406734-406735 on Build GRCh38
rs892764107 Chr.19: 406791-406791 on Build GRCh38
rs7247159 Chr.19: 406724-406724 on Build GRCh38
rs1019703760 Chr.19: 406538-406538 on Build GRCh38
rs959460679 Chr.19: 406848-406848 on Build GRCh38
rs920565684 Chr.19: 406769-406769 on Build GRCh38
rs890657679 Chr.19: 406779-406779 on Build GRCh38
rs1025414624 Chr.19: 406506-406506 on Build GRCh38
rs552086733 Chr.19: 406820-406820 on Build GRCh38
rs558533874 Chr.19: 406584-406584 on Build GRCh38
rs926622088 Chr.19: 406621-406621 on Build GRCh38
rs183908020 Chr.19: 406761-406761 on Build GRCh38
rs897944077 Chr.19: 406825-406825 on Build GRCh38
rs987955494 Chr.19: 406593-406593 on Build GRCh38
rs111590454 Chr.19: 406851-406851 on Build GRCh38
rs1002418320 Chr.19: 406480-406480 on Build GRCh38
rs1050240462 Chr.19: 406555-406555 on Build GRCh38
rs373847849 Chr.19: 406561-406561 on Build GRCh38
rs927993656 Chr.19: 406624-406624 on Build GRCh38
rs538595960 Chr.19: 406656-406656 on Build GRCh38
rs971004012 Chr.19: 406842-406842 on Build GRCh38
rs1046852312 Chr.19: 406461-406461 on Build GRCh38
rs565666684 Chr.19: 406788-406788 on Build GRCh38
rs549450895 Chr.19: 406467-406467 on Build GRCh38
rs187155860 Chr.19: 406901-406901 on Build GRCh38
rs541179276 Chr.19: 406680-406680 on Build GRCh38
rs2310958 Chr.19: 406497-406497 on Build GRCh38

View Full Product Details