Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1009776-1010271 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRIN3B
Entrez Gene ID
116444
Gene Name
glutamate ionotropic receptor NMDA type subunit 3B
Gene Aliases
GluN3B, NR3B
UniGene
Hs.660378
Gene Chromosome Location
Chr.19: 1000419-1009814 on Build GRCh38
Transcripts
XM_017026243.1 NM_001033026.1 NM_033420.3 XM_005259675.2 XM_005259676.2 XM_005259677.2 XM_005259678.2 XM_017027457.1

Gene Symbol
TMEM259
Entrez Gene ID
91304
Gene Name
transmembrane protein 259
Gene Aliases
ASBABP1, C19orf6, MBRL, MEMBRALIN, R32184_3
UniGene
Hs.515003
Gene Chromosome Location
Chr.19: 1009651-1021169 on Build GRCh38
Transcripts
XM_017026243.1 NM_001033026.1 NM_033420.3 XM_005259675.2 XM_005259676.2 XM_005259677.2 XM_005259678.2 XM_017027457.1

SNP Detail

SNP ID Chromosome Location
rs1056938648 Chr.19: 1010085-1010085 on Build GRCh38
rs916376498 Chr.19: 1009911-1009911 on Build GRCh38
rs1024301983 Chr.19: 1010248-1010248 on Build GRCh38
rs890206661 Chr.19: 1010130-1010130 on Build GRCh38
rs559120926 Chr.19: 1009997-1009997 on Build GRCh38
rs1058603 Chr.19: 1009884-1009884 on Build GRCh38
rs758768716 Chr.19: 1010095-1010095 on Build GRCh38
rs1028547238 Chr.19: 1010167-1010167 on Build GRCh38
rs77532719 Chr.19: 1009805-1009805 on Build GRCh38
rs903376334 Chr.19: 1010190-1010190 on Build GRCh38
rs1053827833 Chr.19: 1009881-1009881 on Build GRCh38
rs1058792 Chr.19: 1009864-1009864 on Build GRCh38
rs759606693 Chr.19: 1010238-1010238 on Build GRCh38
rs764389226 Chr.19: 1009973-1009973 on Build GRCh38
rs538161277 Chr.19: 1009856-1009856 on Build GRCh38
rs1056989383 Chr.19: 1010241-1010241 on Build GRCh38
rs11540363 Chr.19: 1009796-1009796 on Build GRCh38
rs114997271 Chr.19: 1009821-1009821 on Build GRCh38
rs1015977053 Chr.19: 1009838-1009838 on Build GRCh38
rs746504731 Chr.19: 1010105-1010105 on Build GRCh38
rs976954984 Chr.19: 1010013-1010013 on Build GRCh38
rs1042042996 Chr.19: 1009900-1009900 on Build GRCh38
rs952498278 Chr.19: 1009981-1009981 on Build GRCh38
rs148752826 Chr.19: 1010104-1010104 on Build GRCh38
rs945471331 Chr.19: 1010206-1010206 on Build GRCh38
rs534162598 Chr.19: 1009939-1009939 on Build GRCh38
rs578003248 Chr.19: 1009846-1009846 on Build GRCh38
rs541944315 Chr.19: 1010056-1010056 on Build GRCh38
rs996304626 Chr.19: 1009968-1009968 on Build GRCh38
rs566817080 Chr.19: 1010237-1010237 on Build GRCh38
rs1026700522 Chr.19: 1009979-1009979 on Build GRCh38
rs977005921 Chr.19: 1010155-1010155 on Build GRCh38
rs1004515385 Chr.19: 1009830-1009830 on Build GRCh38
rs1004315136 Chr.19: 1010135-1010135 on Build GRCh38
rs958923034 Chr.19: 1010228-1010228 on Build GRCh38
rs183246851 Chr.19: 1010040-1010040 on Build GRCh38
rs1050558990 Chr.19: 1010117-1010117 on Build GRCh38
rs991612773 Chr.19: 1009891-1009891 on Build GRCh38
rs1037766951 Chr.19: 1009944-1009944 on Build GRCh38
rs115454056 Chr.19: 1010184-1010184 on Build GRCh38
rs952372949 Chr.19: 1009804-1009804 on Build GRCh38
rs1012621520 Chr.19: 1009977-1009977 on Build GRCh38
rs967332205 Chr.19: 1009872-1009872 on Build GRCh38
rs116247697 Chr.19: 1010116-1010116 on Build GRCh38
rs1002688173 Chr.19: 1009917-1009917 on Build GRCh38
rs938597827 Chr.19: 1010076-1010076 on Build GRCh38
rs1046316587 Chr.19: 1009812-1009812 on Build GRCh38
rs796729687 Chr.19: 1010121-1010121 on Build GRCh38
rs560020484 Chr.19: 1010060-1010060 on Build GRCh38
rs568906861 Chr.19: 1010173-1010173 on Build GRCh38
rs926008557 Chr.19: 1009886-1009886 on Build GRCh38
rs780680909 Chr.19: 1010099-1010099 on Build GRCh38
rs77386938 Chr.19: 1010028-1010028 on Build GRCh38
rs1015253572 Chr.19: 1010136-1010136 on Build GRCh38
rs954210424 Chr.19: 1010172-1010172 on Build GRCh38
rs1055359483 Chr.19: 1009797-1009797 on Build GRCh38
rs750815427 Chr.19: 1010061-1010061 on Build GRCh38
rs966866033 Chr.19: 1010144-1010144 on Build GRCh38
rs11540364 Chr.19: 1009871-1009871 on Build GRCh38
rs945391705 Chr.19: 1010048-1010048 on Build GRCh38
rs577440562 Chr.19: 1010022-1010022 on Build GRCh38
rs766352941 Chr.19: 1009801-1009801 on Build GRCh38
rs898853716 Chr.19: 1009962-1009962 on Build GRCh38
rs1000481993 Chr.19: 1009991-1009991 on Build GRCh38
rs112912381 Chr.19: 1009982-1009982 on Build GRCh38
rs944223003 Chr.19: 1009843-1009843 on Build GRCh38
rs780789535 Chr.19: 1010223-1010223 on Build GRCh38
rs13160 Chr.19: 1009915-1009915 on Build GRCh38
rs550511896 Chr.19: 1009861-1009861 on Build GRCh38
rs927988890 Chr.19: 1010224-1010224 on Build GRCh38
rs554689012 Chr.19: 1009904-1009904 on Build GRCh38
rs116971600 Chr.19: 1010210-1010210 on Build GRCh38
rs915337659 Chr.19: 1010193-1010193 on Build GRCh38
rs1065363 Chr.19: 1009920-1009920 on Build GRCh38
rs1058764 Chr.19: 1009946-1009946 on Build GRCh38
rs534322397 Chr.19: 1010244-1010244 on Build GRCh38
rs917342097 Chr.19: 1010025-1010025 on Build GRCh38
rs1008862781 Chr.19: 1009987-1009987 on Build GRCh38
rs1035479606 Chr.19: 1009933-1009933 on Build GRCh38
rs1016283389 Chr.19: 1009992-1009992 on Build GRCh38
rs549702255 Chr.19: 1010154-1010154 on Build GRCh38
rs531462585 Chr.19: 1010138-1010138 on Build GRCh38
rs376133714 Chr.19: 1010108-1010108 on Build GRCh38
rs937410249 Chr.19: 1009888-1009888 on Build GRCh38
rs940591757 Chr.19: 1010090-1010090 on Build GRCh38
rs945366715 Chr.19: 1009897-1009897 on Build GRCh38
rs370000938 Chr.19: 1010190-1010190 on Build GRCh38
rs568918724 Chr.19: 1009863-1009863 on Build GRCh38
rs1008684734 Chr.19: 1010168-1010168 on Build GRCh38
rs11540365 Chr.19: 1009798-1009798 on Build GRCh38
rs1037640920 Chr.19: 1010100-1010100 on Build GRCh38
rs13627 Chr.19: 1010023-1010023 on Build GRCh38
rs940653767 Chr.19: 1009941-1009941 on Build GRCh38
rs907801780 Chr.19: 1009934-1009934 on Build GRCh38

View Full Product Details