Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1010349-1010889 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM259
Entrez Gene ID
91304
Gene Name
transmembrane protein 259
Gene Aliases
ASBABP1, C19orf6, MBRL, MEMBRALIN, R32184_3
UniGene
Hs.515003
Gene Chromosome Location
Chr.19: 1009651-1021169 on Build GRCh38
Transcripts
NM_001033026.1 NM_033420.3 XM_005259675.2 XM_005259676.2 XM_005259677.2 XM_005259678.2 XM_017027457.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6381161 c.1622T>A p.M541K Chr.19: 1010591-1010591 on Build GRCh38
6374663 c.1577C>A p.A526E Chr.19: 1010636-1010636 on Build GRCh38
6381722 c.1550C>T p.A517V Chr.19: 1010663-1010663 on Build GRCh38
4073699 c.1348G>A p.E450K Chr.19: 1010865-1010865 on Build GRCh38
4131129 c.1521T>C p.P507P Chr.19: 1010692-1010692 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775848544 Chr.19: 1010599-1010599 on Build GRCh38
rs775510259 Chr.19: 1010850-1010850 on Build GRCh38
rs1015334131 Chr.19: 1010814-1010814 on Build GRCh38
rs764178778 Chr.19: 1010413-1010413 on Build GRCh38
rs928835377 Chr.19: 1010782-1010782 on Build GRCh38
rs553445332 Chr.19: 1010776-1010776 on Build GRCh38
rs375152820 Chr.19: 1010371-1010371 on Build GRCh38
rs778370526 Chr.19: 1010533-1010556 on Build GRCh38
rs750517552 Chr.19: 1010639-1010639 on Build GRCh38
rs1011159058 Chr.19: 1010519-1010519 on Build GRCh38
rs758058478 Chr.19: 1010660-1010660 on Build GRCh38
rs752963465 Chr.19: 1010388-1010388 on Build GRCh38
rs764231714 Chr.19: 1010567-1010567 on Build GRCh38
rs762153382 Chr.19: 1010635-1010635 on Build GRCh38
rs756897645 Chr.19: 1010725-1010725 on Build GRCh38
rs760855478 Chr.19: 1010695-1010695 on Build GRCh38
rs767581676 Chr.19: 1010636-1010636 on Build GRCh38
rs748974414 Chr.19: 1010434-1010434 on Build GRCh38
rs760806208 Chr.19: 1010498-1010498 on Build GRCh38
rs375149925 Chr.19: 1010773-1010773 on Build GRCh38
rs779786855 Chr.19: 1010508-1010508 on Build GRCh38
rs760663235 Chr.19: 1010644-1010644 on Build GRCh38
rs1058810 Chr.19: 1010808-1010808 on Build GRCh38
rs758661782 Chr.19: 1010395-1010395 on Build GRCh38
rs973258332 Chr.19: 1010435-1010435 on Build GRCh38
rs543346420 Chr.19: 1010547-1010547 on Build GRCh38
rs762823507 Chr.19: 1010851-1010851 on Build GRCh38
rs962029917 Chr.19: 1010823-1010823 on Build GRCh38
rs779554733 Chr.19: 1010747-1010747 on Build GRCh38
rs765217467 Chr.19: 1010786-1010786 on Build GRCh38
rs751810702 Chr.19: 1010493-1010493 on Build GRCh38
rs758895356 Chr.19: 1010825-1010825 on Build GRCh38
rs754750585 Chr.19: 1010532-1010532 on Build GRCh38
rs771672096 Chr.19: 1010562-1010562 on Build GRCh38
rs749847410 Chr.19: 1010682-1010682 on Build GRCh38
rs370807950 Chr.19: 1010755-1010755 on Build GRCh38
rs1054624626 Chr.19: 1010509-1010509 on Build GRCh38
rs751530483 Chr.19: 1010610-1010610 on Build GRCh38
rs756093650 Chr.19: 1010507-1010507 on Build GRCh38
rs751322999 Chr.19: 1010797-1010797 on Build GRCh38
rs775383878 Chr.19: 1010788-1010788 on Build GRCh38
rs2240159 Chr.19: 1010692-1010692 on Build GRCh38
rs764346419 Chr.19: 1010396-1010396 on Build GRCh38
rs747710292 Chr.19: 1010458-1010458 on Build GRCh38
rs746587856 Chr.19: 1010669-1010669 on Build GRCh38
rs757457166 Chr.19: 1010494-1010494 on Build GRCh38
rs1027470607 Chr.19: 1010575-1010575 on Build GRCh38
rs907176892 Chr.19: 1010533-1010533 on Build GRCh38
rs771547295 Chr.19: 1010465-1010465 on Build GRCh38
rs77868901 Chr.19: 1010397-1010397 on Build GRCh38
rs752097768 Chr.19: 1010838-1010838 on Build GRCh38
rs753693052 Chr.19: 1010653-1010653 on Build GRCh38
rs759545655 Chr.19: 1010785-1010785 on Build GRCh38
rs761244317 Chr.19: 1010374-1010374 on Build GRCh38
rs750887241 Chr.19: 1010417-1010417 on Build GRCh38
rs756650314 Chr.19: 1010455-1010456 on Build GRCh38
rs567001833 Chr.19: 1010746-1010746 on Build GRCh38
rs543289243 Chr.19: 1010651-1010651 on Build GRCh38
rs568738514 Chr.19: 1010729-1010729 on Build GRCh38
rs766934371 Chr.19: 1010377-1010377 on Build GRCh38
rs773984176 Chr.19: 1010447-1010447 on Build GRCh38
rs994885629 Chr.19: 1010692-1010708 on Build GRCh38
rs769836141 Chr.19: 1010846-1010846 on Build GRCh38
rs1023696312 Chr.19: 1010791-1010791 on Build GRCh38
rs777106663 Chr.19: 1010381-1010381 on Build GRCh38
rs762847651 Chr.19: 1010792-1010792 on Build GRCh38
rs776828471 Chr.19: 1010839-1010839 on Build GRCh38
rs952103278 Chr.19: 1010593-1010593 on Build GRCh38
rs556881874 Chr.19: 1010718-1010718 on Build GRCh38
rs746439530 Chr.19: 1010569-1010569 on Build GRCh38
rs185964545 Chr.19: 1010523-1010523 on Build GRCh38
rs566862093 Chr.19: 1010687-1010687 on Build GRCh38
rs750423204 Chr.19: 1010505-1010505 on Build GRCh38
rs755550740 Chr.19: 1010740-1010740 on Build GRCh38
rs774820642 Chr.19: 1010629-1010629 on Build GRCh38
rs747604535 Chr.19: 1010639-1010662 on Build GRCh38
rs922430115 Chr.19: 1010727-1010727 on Build GRCh38
rs561681748 Chr.19: 1010659-1010659 on Build GRCh38
rs1048746416 Chr.19: 1010456-1010456 on Build GRCh38
rs564800791 Chr.19: 1010783-1010783 on Build GRCh38
rs763859039 Chr.19: 1010710-1010710 on Build GRCh38
rs764468869 Chr.19: 1010482-1010482 on Build GRCh38
rs902798970 Chr.19: 1010758-1010758 on Build GRCh38
rs10401738 Chr.19: 1010756-1010756 on Build GRCh38
rs777044051 Chr.19: 1010467-1010467 on Build GRCh38
rs778709761 Chr.19: 1010657-1010657 on Build GRCh38
rs868786236 Chr.19: 1010455-1010455 on Build GRCh38
rs770768939 Chr.19: 1010426-1010426 on Build GRCh38
rs767740453 Chr.19: 1010504-1010504 on Build GRCh38
rs746784027 Chr.19: 1010758-1010760 on Build GRCh38
rs564114518 Chr.19: 1010566-1010566 on Build GRCh38
rs961578826 Chr.19: 1010713-1010713 on Build GRCh38
rs768298504 Chr.19: 1010445-1010445 on Build GRCh38
rs749003927 Chr.19: 1010370-1010370 on Build GRCh38
rs775929855 Chr.19: 1010478-1010478 on Build GRCh38
rs375738820 Chr.19: 1010522-1010522 on Build GRCh38
rs865868288 Chr.19: 1010406-1010406 on Build GRCh38
rs62131162 Chr.19: 1010407-1010407 on Build GRCh38
rs202006160 Chr.19: 1010837-1010837 on Build GRCh38
rs760118582 Chr.19: 1010470-1010470 on Build GRCh38
rs527744500 Chr.19: 1010691-1010691 on Build GRCh38
rs972546106 Chr.19: 1010637-1010637 on Build GRCh38
rs889461539 Chr.19: 1010658-1010658 on Build GRCh38
rs199857013 Chr.19: 1010548-1010548 on Build GRCh38
rs541213100 Chr.19: 1010700-1010700 on Build GRCh38
rs1026409321 Chr.19: 1010781-1010810 on Build GRCh38
rs866593137 Chr.19: 1010414-1010414 on Build GRCh38
rs1050284409 Chr.19: 1010704-1010705 on Build GRCh38
rs183061118 Chr.19: 1010702-1010702 on Build GRCh38
rs762810824 Chr.19: 1010424-1010424 on Build GRCh38
rs201119201 Chr.19: 1010810-1010810 on Build GRCh38
rs200818449 Chr.19: 1010488-1010488 on Build GRCh38
rs745319187 Chr.19: 1010737-1010737 on Build GRCh38
rs557860304 Chr.19: 1010813-1010813 on Build GRCh38
rs986069458 Chr.19: 1010459-1010459 on Build GRCh38
rs765100951 Chr.19: 1010701-1010701 on Build GRCh38
rs753995364 Chr.19: 1010581-1010581 on Build GRCh38
rs745997144 Chr.19: 1010841-1010841 on Build GRCh38
rs763461027 Chr.19: 1010601-1010601 on Build GRCh38
rs757400179 Chr.19: 1010399-1010399 on Build GRCh38
rs769114027 Chr.19: 1010611-1010611 on Build GRCh38
rs11540362 Chr.19: 1010674-1010674 on Build GRCh38
rs753874054 Chr.19: 1010696-1010696 on Build GRCh38
rs764037047 Chr.19: 1010863-1010863 on Build GRCh38
rs754805969 Chr.19: 1010656-1010656 on Build GRCh38
rs779607928 Chr.19: 1010812-1010812 on Build GRCh38
rs1044001211 Chr.19: 1010673-1010673 on Build GRCh38
rs910938492 Chr.19: 1010752-1010752 on Build GRCh38
rs767216885 Chr.19: 1010806-1010806 on Build GRCh38
rs924387988 Chr.19: 1010487-1010487 on Build GRCh38
rs1008352619 Chr.19: 1010849-1010849 on Build GRCh38
rs765611899 Chr.19: 1010477-1010477 on Build GRCh38
rs201676598 Chr.19: 1010683-1010683 on Build GRCh38
rs780652243 Chr.19: 1010681-1010681 on Build GRCh38
rs1000078217 Chr.19: 1010539-1010539 on Build GRCh38
rs763827608 Chr.19: 1010796-1010796 on Build GRCh38
rs745790852 Chr.19: 1010420-1010420 on Build GRCh38
rs778582961 Chr.19: 1010544-1010544 on Build GRCh38
rs765602985 Chr.19: 1010387-1010387 on Build GRCh38
rs372851056 Chr.19: 1010479-1010479 on Build GRCh38
rs531446822 Chr.19: 1010641-1010641 on Build GRCh38
rs954263233 Chr.19: 1010379-1010379 on Build GRCh38
rs747226405 Chr.19: 1010781-1010781 on Build GRCh38
rs909721339 Chr.19: 1010463-1010463 on Build GRCh38
rs1016879025 Chr.19: 1010712-1010712 on Build GRCh38
rs151265454 Chr.19: 1010433-1010433 on Build GRCh38

View Full Product Details