Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1440214-1440749 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPS15
Entrez Gene ID
6209
Gene Name
ribosomal protein S15
Gene Aliases
RIG, S15
UniGene
Hs.406683
Gene Chromosome Location
Chr.19: 1438364-1440497 on Build GRCh38
Transcripts
NM_001018.4 NM_001308226.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4340347 c.391C>T p.P131S Chr.19: 1440415-1440415 on Build GRCh38
991851 c.384T>C p.H128H Chr.19: 1440408-1440408 on Build GRCh38
4401416 c.417C>T p.S139S Chr.19: 1440441-1440441 on Build GRCh38
4486886 c.310C>T p.Q104* Chr.19: 1440239-1440239 on Build GRCh38
4542313 c.315G>A p.V105V Chr.19: 1440244-1440244 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1026168282 Chr.19: 1440691-1440691 on Build GRCh38
rs932561987 Chr.19: 1440675-1440675 on Build GRCh38
rs147232734 Chr.19: 1440360-1440360 on Build GRCh38
rs562792008 Chr.19: 1440593-1440593 on Build GRCh38
rs770007962 Chr.19: 1440281-1440281 on Build GRCh38
rs762052044 Chr.19: 1440508-1440508 on Build GRCh38
rs530205134 Chr.19: 1440643-1440643 on Build GRCh38
rs766478125 Chr.19: 1440328-1440328 on Build GRCh38
rs577500033 Chr.19: 1440256-1440256 on Build GRCh38
rs779270514 Chr.19: 1440438-1440438 on Build GRCh38
rs769561997 Chr.19: 1440463-1440463 on Build GRCh38
rs751682265 Chr.19: 1440332-1440332 on Build GRCh38
rs559642808 Chr.19: 1440522-1440522 on Build GRCh38
rs893625094 Chr.19: 1440689-1440689 on Build GRCh38
rs866149891 Chr.19: 1440482-1440482 on Build GRCh38
rs770812659 Chr.19: 1440481-1440481 on Build GRCh38
rs919008473 Chr.19: 1440308-1440308 on Build GRCh38
rs1026073507 Chr.19: 1440526-1440526 on Build GRCh38
rs749066783 Chr.19: 1440477-1440477 on Build GRCh38
rs372356339 Chr.19: 1440316-1440316 on Build GRCh38
rs148819921 Chr.19: 1440665-1440665 on Build GRCh38
rs530285210 Chr.19: 1440536-1440536 on Build GRCh38
rs189976721 Chr.19: 1440523-1440523 on Build GRCh38
rs757758099 Chr.19: 1440345-1440345 on Build GRCh38
rs150004865 Chr.19: 1440486-1440486 on Build GRCh38
rs779437588 Chr.19: 1440351-1440351 on Build GRCh38
rs760172624 Chr.19: 1440303-1440303 on Build GRCh38
rs762922130 Chr.19: 1440327-1440327 on Build GRCh38
rs751073871 Chr.19: 1440529-1440529 on Build GRCh38
rs992166950 Chr.19: 1440558-1440558 on Build GRCh38
rs751698438 Chr.19: 1440272-1440272 on Build GRCh38
rs1011203319 Chr.19: 1440657-1440670 on Build GRCh38
rs767487024 Chr.19: 1440307-1440307 on Build GRCh38
rs182407332 Chr.19: 1440651-1440651 on Build GRCh38
rs755211240 Chr.19: 1440275-1440275 on Build GRCh38
rs750545902 Chr.19: 1440633-1440633 on Build GRCh38
rs947523940 Chr.19: 1440637-1440637 on Build GRCh38
rs373220983 Chr.19: 1440447-1440447 on Build GRCh38
rs958954016 Chr.19: 1440542-1440542 on Build GRCh38
rs750337911 Chr.19: 1440261-1440261 on Build GRCh38
rs866904785 Chr.19: 1440541-1440541 on Build GRCh38
rs765197829 Chr.19: 1440257-1440257 on Build GRCh38
rs774109942 Chr.19: 1440399-1440399 on Build GRCh38
rs943682984 Chr.19: 1440634-1440634 on Build GRCh38
rs112773659 Chr.19: 1440494-1440494 on Build GRCh38
rs139332437 Chr.19: 1440375-1440375 on Build GRCh38
rs993349549 Chr.19: 1440686-1440686 on Build GRCh38
rs1053300491 Chr.19: 1440615-1440615 on Build GRCh38
rs766385001 Chr.19: 1440395-1440395 on Build GRCh38
rs748525817 Chr.19: 1440279-1440279 on Build GRCh38
rs966370590 Chr.19: 1440311-1440311 on Build GRCh38
rs749732610 Chr.19: 1440286-1440286 on Build GRCh38
rs775429612 Chr.19: 1440495-1440495 on Build GRCh38
rs377297587 Chr.19: 1440340-1440340 on Build GRCh38
rs763886990 Chr.19: 1440539-1440539 on Build GRCh38
rs1002867968 Chr.19: 1440450-1440450 on Build GRCh38
rs908327272 Chr.19: 1440607-1440607 on Build GRCh38
rs934902715 Chr.19: 1440600-1440600 on Build GRCh38
rs758343372 Chr.19: 1440687-1440687 on Build GRCh38
rs149421216 Chr.19: 1440417-1440417 on Build GRCh38
rs951816132 Chr.19: 1440534-1440534 on Build GRCh38
rs535301473 Chr.19: 1440527-1440527 on Build GRCh38
rs967342574 Chr.19: 1440519-1440519 on Build GRCh38
rs566852534 Chr.19: 1440652-1440652 on Build GRCh38
rs1054136171 Chr.19: 1440685-1440685 on Build GRCh38
rs767434466 Chr.19: 1440408-1440408 on Build GRCh38
rs747748171 Chr.19: 1440452-1440452 on Build GRCh38
rs367755975 Chr.19: 1440285-1440285 on Build GRCh38
rs757669754 Chr.19: 1440432-1440432 on Build GRCh38
rs773035799 Chr.19: 1440466-1440466 on Build GRCh38
rs1027132908 Chr.19: 1440586-1440586 on Build GRCh38
rs758949305 Chr.19: 1440444-1440444 on Build GRCh38
rs755050534 Chr.19: 1440334-1440334 on Build GRCh38
rs756367394 Chr.19: 1440342-1440342 on Build GRCh38
rs754008361 Chr.19: 1440505-1440505 on Build GRCh38
rs766229763 Chr.19: 1440275-1440284 on Build GRCh38
rs1035661148 Chr.19: 1440668-1440668 on Build GRCh38
rs767548807 Chr.19: 1440490-1440490 on Build GRCh38
rs201240131 Chr.19: 1440510-1440510 on Build GRCh38
rs953016018 Chr.19: 1440592-1440592 on Build GRCh38
rs559707888 Chr.19: 1440485-1440485 on Build GRCh38
rs559322630 Chr.19: 1440253-1440253 on Build GRCh38
rs764367919 Chr.19: 1440428-1440428 on Build GRCh38
rs762905661 Chr.19: 1440381-1440381 on Build GRCh38
rs774968325 Chr.19: 1440294-1440294 on Build GRCh38
rs910287902 Chr.19: 1440562-1440562 on Build GRCh38
rs371659834 Chr.19: 1440288-1440288 on Build GRCh38
rs780213150 Chr.19: 1440268-1440268 on Build GRCh38
rs896405285 Chr.19: 1440669-1440669 on Build GRCh38
rs758441363 Chr.19: 1440265-1440265 on Build GRCh38
rs764280436 Chr.19: 1440497-1440497 on Build GRCh38
rs943070655 Chr.19: 1440563-1440563 on Build GRCh38
rs776236475 Chr.19: 1440367-1440367 on Build GRCh38
rs200097056 Chr.19: 1440456-1440456 on Build GRCh38
rs761547420 Chr.19: 1440310-1440310 on Build GRCh38
rs746319104 Chr.19: 1440442-1440442 on Build GRCh38
rs199705036 Chr.19: 1440411-1440411 on Build GRCh38
rs747688084 Chr.19: 1440373-1440373 on Build GRCh38
rs754173493 Chr.19: 1440429-1440429 on Build GRCh38
rs778055553 Chr.19: 1440344-1440344 on Build GRCh38
rs945242191 Chr.19: 1440706-1440706 on Build GRCh38
rs781333206 Chr.19: 1440277-1440277 on Build GRCh38
rs773123698 Chr.19: 1440379-1440379 on Build GRCh38
rs775367066 Chr.19: 1440284-1440285 on Build GRCh38
rs375011460 Chr.19: 1440480-1440480 on Build GRCh38
rs931050968 Chr.19: 1440384-1440384 on Build GRCh38

View Full Product Details