Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 2514315-2514822 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNG7
Entrez Gene ID
2788
Gene Name
G protein subunit gamma 7
Gene Aliases
-
UniGene
Hs.515544
Gene Chromosome Location
Chr.19: 2511219-2702748 on Build GRCh38
Transcripts
NM_052847.2 XM_017026606.1

SNP Detail

SNP ID Chromosome Location
rs982177209 Chr.19: 2514730-2514730 on Build GRCh38
rs183962368 Chr.19: 2514729-2514729 on Build GRCh38
rs916444125 Chr.19: 2514708-2514708 on Build GRCh38
rs540311572 Chr.19: 2514751-2514751 on Build GRCh38
rs1036500302 Chr.19: 2514381-2514381 on Build GRCh38
rs958117718 Chr.19: 2514540-2514540 on Build GRCh38
rs545460068 Chr.19: 2514792-2514792 on Build GRCh38
rs186498120 Chr.19: 2514334-2514334 on Build GRCh38
rs561994398 Chr.19: 2514656-2514656 on Build GRCh38
rs922658573 Chr.19: 2514412-2514412 on Build GRCh38
rs187278797 Chr.19: 2514609-2514609 on Build GRCh38
rs371504205 Chr.19: 2514617-2514617 on Build GRCh38
rs754739487 Chr.19: 2514673-2514673 on Build GRCh38
rs549704757 Chr.19: 2514366-2514366 on Build GRCh38
rs995225619 Chr.19: 2514417-2514417 on Build GRCh38
rs535194164 Chr.19: 2514745-2514745 on Build GRCh38
rs1017514080 Chr.19: 2514376-2514376 on Build GRCh38
rs917414 Chr.19: 2514362-2514362 on Build GRCh38
rs988106121 Chr.19: 2514647-2514647 on Build GRCh38
rs147103992 Chr.19: 2514427-2514427 on Build GRCh38
rs929047718 Chr.19: 2514703-2514703 on Build GRCh38
rs896170166 Chr.19: 2514520-2514521 on Build GRCh38
rs975886407 Chr.19: 2514393-2514393 on Build GRCh38
rs574001596 Chr.19: 2514682-2514682 on Build GRCh38
rs191020194 Chr.19: 2514651-2514651 on Build GRCh38
rs920828667 Chr.19: 2514750-2514750 on Build GRCh38
rs978433888 Chr.19: 2514505-2514505 on Build GRCh38
rs189948071 Chr.19: 2514350-2514350 on Build GRCh38
rs867375975 Chr.19: 2514681-2514681 on Build GRCh38
rs946639433 Chr.19: 2514657-2514657 on Build GRCh38
rs574320870 Chr.19: 2514340-2514340 on Build GRCh38
rs751118893 Chr.19: 2514658-2514658 on Build GRCh38
rs1047428397 Chr.19: 2514716-2514716 on Build GRCh38
rs1005757816 Chr.19: 2514573-2514573 on Build GRCh38
rs1026729474 Chr.19: 2514423-2514423 on Build GRCh38
rs995412875 Chr.19: 2514625-2514625 on Build GRCh38
rs878855842 Chr.19: 2514483-2514483 on Build GRCh38
rs937817474 Chr.19: 2514680-2514680 on Build GRCh38
rs143964441 Chr.19: 2514584-2514584 on Build GRCh38
rs951549244 Chr.19: 2514367-2514367 on Build GRCh38
rs138544273 Chr.19: 2514436-2514436 on Build GRCh38
rs1037022769 Chr.19: 2514611-2514611 on Build GRCh38
rs182901784 Chr.19: 2514519-2514519 on Build GRCh38
rs538259575 Chr.19: 2514624-2514624 on Build GRCh38
rs1046367218 Chr.19: 2514608-2514608 on Build GRCh38
rs999416213 Chr.19: 2514674-2514674 on Build GRCh38
rs917415 Chr.19: 2514375-2514375 on Build GRCh38
rs148360397 Chr.19: 2514755-2514755 on Build GRCh38
rs541780278 Chr.19: 2514378-2514378 on Build GRCh38
rs112808296 Chr.19: 2514639-2514639 on Build GRCh38
rs940927493 Chr.19: 2514610-2514610 on Build GRCh38
rs17755118 Chr.19: 2514734-2514734 on Build GRCh38
rs942388014 Chr.19: 2514793-2514793 on Build GRCh38
rs758637649 Chr.19: 2514571-2514571 on Build GRCh38
rs549246483 Chr.19: 2514452-2514452 on Build GRCh38
rs1046420474 Chr.19: 2514349-2514349 on Build GRCh38
rs905005905 Chr.19: 2514472-2514472 on Build GRCh38
rs1043619492 Chr.19: 2514457-2514457 on Build GRCh38
rs1050608733 Chr.19: 2514634-2514634 on Build GRCh38
rs948162681 Chr.19: 2514607-2514607 on Build GRCh38
rs76433508 Chr.19: 2514677-2514677 on Build GRCh38
rs1019322029 Chr.19: 2514446-2514446 on Build GRCh38
rs765769994 Chr.19: 2514549-2514549 on Build GRCh38
rs989255260 Chr.19: 2514697-2514697 on Build GRCh38
rs112351144 Chr.19: 2514735-2514735 on Build GRCh38
rs3752174 Chr.19: 2514567-2514567 on Build GRCh38
rs112501431 Chr.19: 2514390-2514390 on Build GRCh38
rs913781741 Chr.19: 2514445-2514445 on Build GRCh38
rs993083435 Chr.19: 2514521-2514521 on Build GRCh38

View Full Product Details