Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 6307074-6307498 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACER1
Entrez Gene ID
125981
Gene Name
alkaline ceramidase 1
Gene Aliases
ALKCDase1, ASAH3
UniGene
Hs.352609
Gene Chromosome Location
Chr.19: 6306142-6360359 on Build GRCh38
Transcripts
NM_133492.2 XM_011527673.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1002866 c.596G>T p.R199M Chr.19: 6307183-6307183 on Build GRCh38
6152468 c.624C>G p.I208M Chr.19: 6307155-6307155 on Build GRCh38
2727388 c.572G>A p.R191H Chr.19: 6307207-6307207 on Build GRCh38
3540885 c.516G>A p.L172L Chr.19: 6307263-6307263 on Build GRCh38
1397013 c.561G>A p.W187* Chr.19: 6307218-6307218 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs182136574 Chr.19: 6307405-6307405 on Build GRCh38
rs748802326 Chr.19: 6307316-6307316 on Build GRCh38
rs369764190 Chr.19: 6307331-6307331 on Build GRCh38
rs905285931 Chr.19: 6307418-6307418 on Build GRCh38
rs759800494 Chr.19: 6307325-6307325 on Build GRCh38
rs763956739 Chr.19: 6307101-6307101 on Build GRCh38
rs770707530 Chr.19: 6307118-6307118 on Build GRCh38
rs990519814 Chr.19: 6307454-6307454 on Build GRCh38
rs758340987 Chr.19: 6307266-6307266 on Build GRCh38
rs769333173 Chr.19: 6307234-6307234 on Build GRCh38
rs75161094 Chr.19: 6307337-6307337 on Build GRCh38
rs768150338 Chr.19: 6307208-6307208 on Build GRCh38
rs185571387 Chr.19: 6307429-6307429 on Build GRCh38
rs774946174 Chr.19: 6307324-6307324 on Build GRCh38
rs780068847 Chr.19: 6307270-6307270 on Build GRCh38
rs769279708 Chr.19: 6307306-6307306 on Build GRCh38
rs779026098 Chr.19: 6307122-6307122 on Build GRCh38
rs1006850744 Chr.19: 6307179-6307179 on Build GRCh38
rs745613358 Chr.19: 6307129-6307129 on Build GRCh38
rs752679766 Chr.19: 6307183-6307183 on Build GRCh38
rs540211951 Chr.19: 6307335-6307335 on Build GRCh38
rs200048395 Chr.19: 6307238-6307238 on Build GRCh38
rs751767455 Chr.19: 6307103-6307103 on Build GRCh38
rs1036601997 Chr.19: 6307384-6307384 on Build GRCh38
rs374922368 Chr.19: 6307145-6307145 on Build GRCh38
rs755080094 Chr.19: 6307107-6307107 on Build GRCh38
rs756289024 Chr.19: 6307194-6307194 on Build GRCh38
rs750375155 Chr.19: 6307264-6307264 on Build GRCh38
rs376515305 Chr.19: 6307251-6307251 on Build GRCh38
rs147053887 Chr.19: 6307235-6307235 on Build GRCh38
rs151165925 Chr.19: 6307207-6307207 on Build GRCh38
rs374124977 Chr.19: 6307248-6307248 on Build GRCh38
rs201541653 Chr.19: 6307116-6307116 on Build GRCh38
rs371234159 Chr.19: 6307429-6307429 on Build GRCh38
rs879325220 Chr.19: 6307444-6307444 on Build GRCh38
rs756235967 Chr.19: 6307257-6307257 on Build GRCh38
rs73559815 Chr.19: 6307336-6307336 on Build GRCh38
rs912741247 Chr.19: 6307100-6307100 on Build GRCh38
rs573281259 Chr.19: 6307096-6307096 on Build GRCh38
rs754848053 Chr.19: 6307273-6307273 on Build GRCh38
rs760623156 Chr.19: 6307094-6307094 on Build GRCh38
rs531533771 Chr.19: 6307131-6307131 on Build GRCh38
rs780678028 Chr.19: 6307282-6307282 on Build GRCh38
rs376645109 Chr.19: 6307461-6307461 on Build GRCh38
rs551946361 Chr.19: 6307368-6307368 on Build GRCh38
rs775254145 Chr.19: 6307133-6307133 on Build GRCh38
rs913132646 Chr.19: 6307313-6307313 on Build GRCh38
rs778976899 Chr.19: 6307200-6307200 on Build GRCh38
rs562608344 Chr.19: 6307382-6307382 on Build GRCh38
rs755099727 Chr.19: 6307170-6307170 on Build GRCh38
rs142427159 Chr.19: 6307168-6307168 on Build GRCh38
rs978389478 Chr.19: 6307143-6307143 on Build GRCh38
rs766200696 Chr.19: 6307339-6307339 on Build GRCh38
rs763271446 Chr.19: 6307148-6307148 on Build GRCh38
rs766795322 Chr.19: 6307164-6307164 on Build GRCh38
rs747862175 Chr.19: 6307305-6307305 on Build GRCh38
rs531375810 Chr.19: 6307391-6307391 on Build GRCh38
rs538794923 Chr.19: 6307135-6307135 on Build GRCh38
rs139590144 Chr.19: 6307221-6307221 on Build GRCh38
rs747588366 Chr.19: 6307232-6307232 on Build GRCh38
rs376784758 Chr.19: 6307242-6307242 on Build GRCh38
rs750977772 Chr.19: 6307147-6307147 on Build GRCh38
rs867415948 Chr.19: 6307277-6307277 on Build GRCh38
rs746728081 Chr.19: 6307271-6307271 on Build GRCh38
rs201079840 Chr.19: 6307218-6307218 on Build GRCh38
rs761412692 Chr.19: 6307142-6307142 on Build GRCh38
rs543930967 Chr.19: 6307374-6307374 on Build GRCh38
rs1046510002 Chr.19: 6307406-6307406 on Build GRCh38
rs898496401 Chr.19: 6307294-6307294 on Build GRCh38
rs767968355 Chr.19: 6307329-6307329 on Build GRCh38
rs771387752 Chr.19: 6307320-6307320 on Build GRCh38
rs754916001 Chr.19: 6307236-6307236 on Build GRCh38
rs781187898 Chr.19: 6307114-6307114 on Build GRCh38
rs200172124 Chr.19: 6307203-6307203 on Build GRCh38
rs781481589 Chr.19: 6307173-6307173 on Build GRCh38
rs763507699 Chr.19: 6307237-6307237 on Build GRCh38
rs147699720 Chr.19: 6307250-6307250 on Build GRCh38
rs772652134 Chr.19: 6307312-6307312 on Build GRCh38
rs764338165 Chr.19: 6307261-6307261 on Build GRCh38
rs754609481 Chr.19: 6307313-6307313 on Build GRCh38
rs753015512 Chr.19: 6307254-6307254 on Build GRCh38
rs775934550 Chr.19: 6307140-6307140 on Build GRCh38
rs771992138 Chr.19: 6307205-6307205 on Build GRCh38

View Full Product Details