Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 6454243-6454744 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC25A23
Entrez Gene ID
79085
Gene Name
solute carrier family 25 member 23
Gene Aliases
APC2, MCSC2, SCaMC-3
UniGene
Hs.356231
Gene Chromosome Location
Chr.19: 6436081-6460221 on Build GRCh38
Transcripts
NM_024103.2 XM_011528274.1 XM_011528275.1 XM_011528276.1 XM_011528277.1 XM_011528278.1 XM_011528279.1 XM_011528280.1 XM_011528281.1 XM_011528282.1 XM_011528283.1 XM_011528284.1 XM_011528285.1 XM_017027285.1 XM_017027286.1 XM_017027287.1 XM_017027288.1 XM_017027289.1 XM_017027290.1 XM_017027291.1 XM_017027292.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6780593 c.689G>A p.R230Q Chr.19: 6454429-6454429 on Build GRCh38
3540915 c.674T>G p.I225S Chr.19: 6454444-6454444 on Build GRCh38
5057052 c.545C>T p.T182M Chr.19: 6454656-6454656 on Build GRCh38
1397368 c.520G>A p.E174K Chr.19: 6454681-6454681 on Build GRCh38
4726818 c.556T>C p.W186R Chr.19: 6454645-6454645 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768164990 Chr.19: 6454320-6454320 on Build GRCh38
rs761183980 Chr.19: 6454335-6454335 on Build GRCh38
rs747828312 Chr.19: 6454419-6454419 on Build GRCh38
rs764031191 Chr.19: 6454677-6454677 on Build GRCh38
rs956751686 Chr.19: 6454534-6454534 on Build GRCh38
rs148091799 Chr.19: 6454298-6454298 on Build GRCh38
rs200108265 Chr.19: 6454451-6454451 on Build GRCh38
rs760929756 Chr.19: 6454510-6454510 on Build GRCh38
rs755851001 Chr.19: 6454554-6454554 on Build GRCh38
rs144673485 Chr.19: 6454429-6454429 on Build GRCh38
rs767359266 Chr.19: 6454664-6454664 on Build GRCh38
rs746413062 Chr.19: 6454581-6454581 on Build GRCh38
rs141513592 Chr.19: 6454637-6454637 on Build GRCh38
rs751505416 Chr.19: 6454536-6454536 on Build GRCh38
rs761827163 Chr.19: 6454522-6454522 on Build GRCh38
rs1007416649 Chr.19: 6454473-6454473 on Build GRCh38
rs757861732 Chr.19: 6454712-6454712 on Build GRCh38
rs553187254 Chr.19: 6454610-6454610 on Build GRCh38
rs998781995 Chr.19: 6454485-6454485 on Build GRCh38
rs939480943 Chr.19: 6454600-6454600 on Build GRCh38
rs776240701 Chr.19: 6454325-6454325 on Build GRCh38
rs752552389 Chr.19: 6454613-6454613 on Build GRCh38
rs760982670 Chr.19: 6454593-6454593 on Build GRCh38
rs760405277 Chr.19: 6454474-6454474 on Build GRCh38
rs555365380 Chr.19: 6454406-6454406 on Build GRCh38
rs901681788 Chr.19: 6454642-6454642 on Build GRCh38
rs767785527 Chr.19: 6454439-6454439 on Build GRCh38
rs758854413 Chr.19: 6454629-6454629 on Build GRCh38
rs546358775 Chr.19: 6454361-6454361 on Build GRCh38
rs1027777028 Chr.19: 6454272-6454272 on Build GRCh38
rs752929163 Chr.19: 6454440-6454440 on Build GRCh38
rs767585978 Chr.19: 6454371-6454371 on Build GRCh38
rs994722404 Chr.19: 6454577-6454577 on Build GRCh38
rs143828533 Chr.19: 6454390-6454390 on Build GRCh38
rs769063053 Chr.19: 6454646-6454646 on Build GRCh38
rs138621235 Chr.19: 6454609-6454609 on Build GRCh38
rs755905898 Chr.19: 6454614-6454614 on Build GRCh38
rs753939864 Chr.19: 6454512-6454512 on Build GRCh38
rs535462610 Chr.19: 6454326-6454326 on Build GRCh38
rs763029573 Chr.19: 6454659-6454659 on Build GRCh38
rs965987191 Chr.19: 6454476-6454476 on Build GRCh38
rs775634908 Chr.19: 6454667-6454667 on Build GRCh38
rs764897151 Chr.19: 6454685-6454685 on Build GRCh38
rs748811382 Chr.19: 6454571-6454571 on Build GRCh38
rs759761443 Chr.19: 6454438-6454438 on Build GRCh38
rs775275908 Chr.19: 6454460-6454460 on Build GRCh38
rs919481136 Chr.19: 6454607-6454607 on Build GRCh38
rs202184533 Chr.19: 6454627-6454627 on Build GRCh38
rs753656506 Chr.19: 6454681-6454681 on Build GRCh38
rs374394051 Chr.19: 6454625-6454625 on Build GRCh38
rs773185504 Chr.19: 6454656-6454656 on Build GRCh38
rs777706742 Chr.19: 6454623-6454623 on Build GRCh38
rs557355336 Chr.19: 6454495-6454495 on Build GRCh38
rs772824474 Chr.19: 6454421-6454421 on Build GRCh38
rs371501314 Chr.19: 6454430-6454430 on Build GRCh38
rs747800728 Chr.19: 6454463-6454463 on Build GRCh38
rs61729399 Chr.19: 6454655-6454655 on Build GRCh38
rs965933867 Chr.19: 6454310-6454310 on Build GRCh38
rs756317236 Chr.19: 6454441-6454441 on Build GRCh38
rs374180459 Chr.19: 6454711-6454711 on Build GRCh38
rs200801447 Chr.19: 6454454-6454454 on Build GRCh38
rs369392953 Chr.19: 6454498-6454498 on Build GRCh38
rs779809798 Chr.19: 6454453-6454453 on Build GRCh38
rs192489049 Chr.19: 6454338-6454338 on Build GRCh38
rs372272220 Chr.19: 6454289-6454289 on Build GRCh38
rs148999239 Chr.19: 6454389-6454389 on Build GRCh38
rs768883309 Chr.19: 6454601-6454601 on Build GRCh38
rs377109434 Chr.19: 6454592-6454592 on Build GRCh38
rs748356618 Chr.19: 6454365-6454365 on Build GRCh38
rs200436209 Chr.19: 6454274-6454274 on Build GRCh38
rs777508730 Chr.19: 6454570-6454570 on Build GRCh38
rs368733973 Chr.19: 6454709-6454709 on Build GRCh38
rs771577567 Chr.19: 6454489-6454489 on Build GRCh38
rs531076203 Chr.19: 6454715-6454715 on Build GRCh38
rs749918479 Chr.19: 6454698-6454698 on Build GRCh38
rs776075903 Chr.19: 6454418-6454418 on Build GRCh38
rs755271159 Chr.19: 6454275-6454275 on Build GRCh38
rs764208908 Chr.19: 6454446-6454446 on Build GRCh38
rs776771269 Chr.19: 6454602-6454602 on Build GRCh38
rs561256810 Chr.19: 6454694-6454694 on Build GRCh38
rs200876868 Chr.19: 6454624-6454624 on Build GRCh38
rs763593363 Chr.19: 6454426-6454426 on Build GRCh38
rs747559236 Chr.19: 6454721-6454721 on Build GRCh38
rs752835589 Chr.19: 6454379-6454379 on Build GRCh38
rs764263928 Chr.19: 6454511-6454511 on Build GRCh38
rs781024193 Chr.19: 6454458-6454458 on Build GRCh38
rs1031555067 Chr.19: 6454523-6454523 on Build GRCh38
rs912988896 Chr.19: 6454432-6454432 on Build GRCh38
rs151079430 Chr.19: 6454402-6454402 on Build GRCh38
rs763536951 Chr.19: 6454341-6454341 on Build GRCh38
rs868089702 Chr.19: 6454470-6454470 on Build GRCh38
rs775903458 Chr.19: 6454509-6454509 on Build GRCh38
rs780886775 Chr.19: 6454541-6454541 on Build GRCh38
rs765279074 Chr.19: 6454605-6454605 on Build GRCh38
rs765351263 Chr.19: 6454525-6454525 on Build GRCh38
rs778473826 Chr.19: 6454579-6454579 on Build GRCh38
rs748355158 Chr.19: 6454648-6454648 on Build GRCh38
rs753886444 Chr.19: 6454447-6454447 on Build GRCh38
rs756892783 Chr.19: 6454682-6454682 on Build GRCh38
rs367621690 Chr.19: 6454301-6454301 on Build GRCh38
rs746727077 Chr.19: 6454312-6454312 on Build GRCh38
rs10402833 Chr.19: 6454356-6454356 on Build GRCh38
rs370370463 Chr.19: 6454628-6454628 on Build GRCh38
rs8103745 Chr.19: 6454640-6454640 on Build GRCh38
rs773919182 Chr.19: 6454340-6454340 on Build GRCh38
rs73920670 Chr.19: 6454295-6454295 on Build GRCh38
rs772379566 Chr.19: 6454503-6454503 on Build GRCh38
rs369833901 Chr.19: 6454603-6454603 on Build GRCh38
rs766896211 Chr.19: 6454355-6454355 on Build GRCh38
rs374018344 Chr.19: 6454551-6454551 on Build GRCh38
rs754917974 Chr.19: 6454539-6454539 on Build GRCh38
rs760677113 Chr.19: 6454671-6454671 on Build GRCh38
rs141353364 Chr.19: 6454403-6454403 on Build GRCh38
rs573934447 Chr.19: 6454578-6454578 on Build GRCh38
rs758311961 Chr.19: 6454391-6454391 on Build GRCh38
rs772602498 Chr.19: 6454587-6454587 on Build GRCh38
rs201308330 Chr.19: 6454469-6454469 on Build GRCh38
rs756254075 Chr.19: 6454388-6454388 on Build GRCh38

View Full Product Details