Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12132885-12133399 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF20
Entrez Gene ID
7568
Gene Name
zinc finger protein 20
Gene Aliases
KOX13
UniGene
Hs.512823
Gene Chromosome Location
Chr.19: 12131350-12140407 on Build GRCh38
Transcripts
NM_001203250.1 NM_021143.3 NR_037802.1

Gene Symbol
ZNF625-ZNF20
Entrez Gene ID
100529855
Gene Name
ZNF625-ZNF20 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.512823
Gene Chromosome Location
Chr.19: 12131350-12156731 on Build GRCh38
Transcripts
NM_001203250.1 NM_021143.3 NR_037802.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1630703 c.1217A>G p.Y406C Chr.19: 12132969-12132969 on Build GRCh38
6279869 c.1020A>G p.E340E Chr.19: 12133166-12133166 on Build GRCh38
1480576 c.1044C>T p.F348F Chr.19: 12133142-12133142 on Build GRCh38
991163 c.924G>A p.E308E Chr.19: 12133262-12133262 on Build GRCh38
1325492 c.852G>A p.E284E Chr.19: 12133334-12133334 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs188768884 Chr.19: 12133055-12133055 on Build GRCh38
rs767730766 Chr.19: 12133211-12133211 on Build GRCh38
rs770762512 Chr.19: 12132976-12132976 on Build GRCh38
rs761046677 Chr.19: 12132957-12132960 on Build GRCh38
rs1018172490 Chr.19: 12133232-12133232 on Build GRCh38
rs747280225 Chr.19: 12132934-12132934 on Build GRCh38
rs751822976 Chr.19: 12133082-12133082 on Build GRCh38
rs752912328 Chr.19: 12133212-12133212 on Build GRCh38
rs528783213 Chr.19: 12133078-12133078 on Build GRCh38
rs770707135 Chr.19: 12133124-12133124 on Build GRCh38
rs973390509 Chr.19: 12133300-12133300 on Build GRCh38
rs771467949 Chr.19: 12133001-12133001 on Build GRCh38
rs758147372 Chr.19: 12133315-12133315 on Build GRCh38
rs371813663 Chr.19: 12132916-12132916 on Build GRCh38
rs922581532 Chr.19: 12133015-12133015 on Build GRCh38
rs371234628 Chr.19: 12133199-12133199 on Build GRCh38
rs144638502 Chr.19: 12133085-12133085 on Build GRCh38
rs879183996 Chr.19: 12132978-12132978 on Build GRCh38
rs879100964 Chr.19: 12132919-12132919 on Build GRCh38
rs758019766 Chr.19: 12133147-12133148 on Build GRCh38
rs777721501 Chr.19: 12132954-12132954 on Build GRCh38
rs867497505 Chr.19: 12132926-12132926 on Build GRCh38
rs200241660 Chr.19: 12133284-12133284 on Build GRCh38
rs777895698 Chr.19: 12133256-12133256 on Build GRCh38
rs200153045 Chr.19: 12133116-12133116 on Build GRCh38
rs748193345 Chr.19: 12132947-12132947 on Build GRCh38
rs375525666 Chr.19: 12132955-12132955 on Build GRCh38
rs879083289 Chr.19: 12133202-12133202 on Build GRCh38
rs756156006 Chr.19: 12133240-12133240 on Build GRCh38
rs776227779 Chr.19: 12133341-12133341 on Build GRCh38
rs756171875 Chr.19: 12133101-12133101 on Build GRCh38
rs879112712 Chr.19: 12133017-12133017 on Build GRCh38
rs763384806 Chr.19: 12133056-12133056 on Build GRCh38
rs983595812 Chr.19: 12133337-12133337 on Build GRCh38
rs570195962 Chr.19: 12133245-12133245 on Build GRCh38
rs377501935 Chr.19: 12133090-12133090 on Build GRCh38
rs201539246 Chr.19: 12133170-12133170 on Build GRCh38
rs754395783 Chr.19: 12133101-12133102 on Build GRCh38
rs751887439 Chr.19: 12133186-12133186 on Build GRCh38
rs878864244 Chr.19: 12132965-12132965 on Build GRCh38
rs902275502 Chr.19: 12133140-12133140 on Build GRCh38
rs866613076 Chr.19: 12133262-12133262 on Build GRCh38
rs755172434 Chr.19: 12133084-12133084 on Build GRCh38
rs878961126 Chr.19: 12133241-12133241 on Build GRCh38
rs771528125 Chr.19: 12133353-12133353 on Build GRCh38
rs776889368 Chr.19: 12132938-12132938 on Build GRCh38
rs892889821 Chr.19: 12133157-12133157 on Build GRCh38
rs769709427 Chr.19: 12132951-12132951 on Build GRCh38
rs202013937 Chr.19: 12133130-12133130 on Build GRCh38
rs758861838 Chr.19: 12132923-12132923 on Build GRCh38
rs534107900 Chr.19: 12133294-12133294 on Build GRCh38
rs766856178 Chr.19: 12133030-12133030 on Build GRCh38
rs778738258 Chr.19: 12133125-12133125 on Build GRCh38
rs929449020 Chr.19: 12132971-12132971 on Build GRCh38
rs985770790 Chr.19: 12133215-12133215 on Build GRCh38
rs61745731 Chr.19: 12133180-12133180 on Build GRCh38
rs964660998 Chr.19: 12133268-12133268 on Build GRCh38
rs763673077 Chr.19: 12133185-12133186 on Build GRCh38
rs763616103 Chr.19: 12133182-12133182 on Build GRCh38
rs773961571 Chr.19: 12132989-12132989 on Build GRCh38
rs879156001 Chr.19: 12133198-12133198 on Build GRCh38
rs779729546 Chr.19: 12133325-12133325 on Build GRCh38
rs747668608 Chr.19: 12133345-12133345 on Build GRCh38
rs199910328 Chr.19: 12133023-12133023 on Build GRCh38
rs879039309 Chr.19: 12133197-12133197 on Build GRCh38
rs759299619 Chr.19: 12132997-12132997 on Build GRCh38
rs1052849080 Chr.19: 12133151-12133151 on Build GRCh38
rs574468226 Chr.19: 12133336-12133336 on Build GRCh38
rs745920069 Chr.19: 12133022-12133022 on Build GRCh38
rs774744338 Chr.19: 12133354-12133354 on Build GRCh38
rs140674699 Chr.19: 12132932-12132932 on Build GRCh38
rs908621372 Chr.19: 12133047-12133047 on Build GRCh38
rs377612416 Chr.19: 12132922-12132922 on Build GRCh38
rs955271175 Chr.19: 12133194-12133194 on Build GRCh38
rs781395520 Chr.19: 12132939-12132939 on Build GRCh38
rs574560050 Chr.19: 12132921-12132921 on Build GRCh38
rs183515721 Chr.19: 12133031-12133031 on Build GRCh38
rs765796983 Chr.19: 12133036-12133036 on Build GRCh38
rs767010680 Chr.19: 12133183-12133183 on Build GRCh38
rs749264919 Chr.19: 12133118-12133118 on Build GRCh38
rs192722077 Chr.19: 12133310-12133310 on Build GRCh38
rs374093683 Chr.19: 12133145-12133145 on Build GRCh38
rs746676870 Chr.19: 12133332-12133332 on Build GRCh38
rs145847204 Chr.19: 12133174-12133174 on Build GRCh38
rs775032921 Chr.19: 12133162-12133162 on Build GRCh38
rs757126330 Chr.19: 12133293-12133293 on Build GRCh38
rs373414827 Chr.19: 12133089-12133089 on Build GRCh38

View Full Product Details