Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 13137189-13137684 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NACC1
Entrez Gene ID
112939
Gene Name
nucleus accumbens associated 1
Gene Aliases
BEND8, BTBD14B, BTBD30, NAC-1, NAC1
UniGene
Hs.531614
Gene Chromosome Location
Chr.19: 13116848-13141147 on Build GRCh38
Transcripts
NM_052876.3 XM_005259721.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6721588 c.1198C>T p.R400W Chr.19: 13137348-13137348 on Build GRCh38
6003515 c.1315G>A p.A439T Chr.19: 13137566-13137566 on Build GRCh38
438658 c.1176C>T p.G392G Chr.19: 13137326-13137326 on Build GRCh38
5490883 c.1226+2T>A p.? Chr.19: 13137378-13137378 on Build GRCh38
5495215 c.1227-2A>C p.? Chr.19: 13137476-13137476 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768549552 Chr.19: 13137314-13137314 on Build GRCh38
rs200569039 Chr.19: 13137254-13137254 on Build GRCh38
rs202081230 Chr.19: 13137231-13137231 on Build GRCh38
rs372461691 Chr.19: 13137461-13137461 on Build GRCh38
rs763704757 Chr.19: 13137383-13137383 on Build GRCh38
rs369073854 Chr.19: 13137435-13137435 on Build GRCh38
rs909700658 Chr.19: 13137656-13137656 on Build GRCh38
rs959857370 Chr.19: 13137215-13137215 on Build GRCh38
rs200105986 Chr.19: 13137408-13137408 on Build GRCh38
rs772718447 Chr.19: 13137597-13137597 on Build GRCh38
rs900306187 Chr.19: 13137657-13137657 on Build GRCh38
rs766034729 Chr.19: 13137401-13137401 on Build GRCh38
rs777947610 Chr.19: 13137392-13137392 on Build GRCh38
rs773370523 Chr.19: 13137232-13137232 on Build GRCh38
rs778872897 Chr.19: 13137315-13137315 on Build GRCh38
rs140114730 Chr.19: 13137499-13137499 on Build GRCh38
rs375331902 Chr.19: 13137431-13137431 on Build GRCh38
rs575090870 Chr.19: 13137255-13137255 on Build GRCh38
rs768635406 Chr.19: 13137227-13137227 on Build GRCh38
rs368795274 Chr.19: 13137365-13137365 on Build GRCh38
rs146131893 Chr.19: 13137330-13137330 on Build GRCh38
rs989245791 Chr.19: 13137623-13137623 on Build GRCh38
rs900414550 Chr.19: 13137450-13137450 on Build GRCh38
rs774609733 Chr.19: 13137533-13137533 on Build GRCh38
rs779749676 Chr.19: 13137284-13137284 on Build GRCh38
rs773923594 Chr.19: 13137602-13137602 on Build GRCh38
rs747944755 Chr.19: 13137414-13137414 on Build GRCh38
rs756964764 Chr.19: 13137405-13137410 on Build GRCh38
rs745459411 Chr.19: 13137426-13137426 on Build GRCh38
rs765947525 Chr.19: 13137436-13137436 on Build GRCh38
rs764977943 Chr.19: 13137447-13137447 on Build GRCh38
rs201169854 Chr.19: 13137448-13137448 on Build GRCh38
rs1038881615 Chr.19: 13137654-13137654 on Build GRCh38
rs759780892 Chr.19: 13137351-13137351 on Build GRCh38
rs377712026 Chr.19: 13137627-13137627 on Build GRCh38
rs752068577 Chr.19: 13137375-13137375 on Build GRCh38
rs367990402 Chr.19: 13137432-13137432 on Build GRCh38
rs960473471 Chr.19: 13137550-13137550 on Build GRCh38
rs749518870 Chr.19: 13137523-13137523 on Build GRCh38
rs138422589 Chr.19: 13137614-13137614 on Build GRCh38
rs770998331 Chr.19: 13137426-13137426 on Build GRCh38
rs983996364 Chr.19: 13137651-13137654 on Build GRCh38
rs764645348 Chr.19: 13137371-13137371 on Build GRCh38
rs531044891 Chr.19: 13137367-13137367 on Build GRCh38
rs375919115 Chr.19: 13137320-13137320 on Build GRCh38
rs189998121 Chr.19: 13137474-13137474 on Build GRCh38
rs930007970 Chr.19: 13137635-13137635 on Build GRCh38
rs780213965 Chr.19: 13137505-13137505 on Build GRCh38
rs1018301095 Chr.19: 13137610-13137610 on Build GRCh38
rs374545548 Chr.19: 13137329-13137329 on Build GRCh38
rs925941464 Chr.19: 13137405-13137405 on Build GRCh38
rs765643095 Chr.19: 13137257-13137257 on Build GRCh38
rs376552799 Chr.19: 13137406-13137406 on Build GRCh38
rs568103073 Chr.19: 13137467-13137467 on Build GRCh38
rs761889887 Chr.19: 13137229-13137229 on Build GRCh38
rs769014877 Chr.19: 13137525-13137525 on Build GRCh38
rs753401854 Chr.19: 13137439-13137439 on Build GRCh38
rs544079071 Chr.19: 13137278-13137278 on Build GRCh38
rs563515047 Chr.19: 13137341-13137341 on Build GRCh38
rs766612303 Chr.19: 13137240-13137240 on Build GRCh38
rs149246627 Chr.19: 13137618-13137618 on Build GRCh38
rs201961947 Chr.19: 13137430-13137430 on Build GRCh38
rs765549322 Chr.19: 13137356-13137356 on Build GRCh38
rs557681256 Chr.19: 13137451-13137451 on Build GRCh38
rs755614345 Chr.19: 13137388-13137388 on Build GRCh38
rs372781005 Chr.19: 13137556-13137556 on Build GRCh38
rs780504447 Chr.19: 13137267-13137267 on Build GRCh38
rs372175440 Chr.19: 13137440-13137440 on Build GRCh38
rs763308699 Chr.19: 13137368-13137368 on Build GRCh38
rs758694300 Chr.19: 13137328-13137328 on Build GRCh38
rs748984953 Chr.19: 13137296-13137296 on Build GRCh38
rs762248857 Chr.19: 13137434-13137434 on Build GRCh38
rs780836889 Chr.19: 13137425-13137426 on Build GRCh38
rs760752426 Chr.19: 13137236-13137236 on Build GRCh38
rs746929045 Chr.19: 13137462-13137462 on Build GRCh38
rs570819174 Chr.19: 13137628-13137628 on Build GRCh38
rs749074942 Chr.19: 13137226-13137226 on Build GRCh38
rs370917305 Chr.19: 13137321-13137321 on Build GRCh38
rs181258080 Chr.19: 13137603-13137603 on Build GRCh38
rs758196083 Chr.19: 13137419-13137419 on Build GRCh38
rs755422378 Chr.19: 13137255-13137255 on Build GRCh38
rs916113827 Chr.19: 13137557-13137557 on Build GRCh38
rs751066940 Chr.19: 13137404-13137404 on Build GRCh38
rs1004975661 Chr.19: 13137508-13137508 on Build GRCh38
rs375357951 Chr.19: 13137471-13137471 on Build GRCh38
rs377339552 Chr.19: 13137574-13137574 on Build GRCh38
rs867273051 Chr.19: 13137534-13137534 on Build GRCh38
rs560977787 Chr.19: 13137232-13137232 on Build GRCh38
rs755970142 Chr.19: 13137450-13137451 on Build GRCh38
rs367710907 Chr.19: 13137260-13137260 on Build GRCh38
rs777735101 Chr.19: 13137420-13137420 on Build GRCh38
rs749960513 Chr.19: 13137277-13137277 on Build GRCh38
rs1034748037 Chr.19: 13137526-13137526 on Build GRCh38
rs758938722 Chr.19: 13137266-13137266 on Build GRCh38
rs149167452 Chr.19: 13137226-13137227 on Build GRCh38
rs1043442263 Chr.19: 13137390-13137390 on Build GRCh38
rs929808453 Chr.19: 13137422-13137422 on Build GRCh38

View Full Product Details