Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 43544951-43545451 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTPRF
Entrez Gene ID
5792
Gene Name
protein tyrosine phosphatase, receptor type F
Gene Aliases
BNAH2, LAR
UniGene
Hs.272062
Gene Chromosome Location
Chr.1: 43522238-43623672 on Build GRCh38
Transcripts
XM_005271079.3 XM_005271080.3 XM_005271081.3 XM_005271082.3 XM_006710795.3 XM_006710796.3 XM_006710797.3 XM_006710798.3 XM_006710799.3 XM_006710800.3 XM_006710801.3 XM_011541871.2 XM_011541872.2 XM_011541873.2 XM_017001943.1 XM_017001944.1 XM_017001945.1 XM_017001946.1 XM_017001947.1 XM_017001948.1 XM_017001949.1 XM_017001950.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7386242 c.79G>T p.A27S Chr.1: 43545154-43545154 on Build GRCh38
4808568 c.66T>C p.G22G Chr.1: 43545141-43545141 on Build GRCh38
6126679 c.21A>T p.P7P Chr.1: 43545096-43545096 on Build GRCh38
4650913 c.34G>A p.V12M Chr.1: 43545109-43545109 on Build GRCh38
6384263 c.70G>T p.V24L Chr.1: 43545145-43545145 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367985690 Chr.1: 43545147-43545147 on Build GRCh38
rs144716808 Chr.1: 43545305-43545305 on Build GRCh38
rs149085019 Chr.1: 43545016-43545016 on Build GRCh38
rs1021583607 Chr.1: 43545424-43545424 on Build GRCh38
rs766928002 Chr.1: 43545092-43545092 on Build GRCh38
rs368269221 Chr.1: 43545215-43545215 on Build GRCh38
rs762766721 Chr.1: 43545037-43545037 on Build GRCh38
rs1038578914 Chr.1: 43545227-43545227 on Build GRCh38
rs201337870 Chr.1: 43545153-43545153 on Build GRCh38
rs964315531 Chr.1: 43545257-43545257 on Build GRCh38
rs142286926 Chr.1: 43545154-43545154 on Build GRCh38
rs117228743 Chr.1: 43545130-43545130 on Build GRCh38
rs539339335 Chr.1: 43545330-43545330 on Build GRCh38
rs990056470 Chr.1: 43545267-43545267 on Build GRCh38
rs553173981 Chr.1: 43545367-43545367 on Build GRCh38
rs754662738 Chr.1: 43545069-43545069 on Build GRCh38
rs201191611 Chr.1: 43545014-43545014 on Build GRCh38
rs556913472 Chr.1: 43545395-43545395 on Build GRCh38
rs200019988 Chr.1: 43545179-43545179 on Build GRCh38
rs910852352 Chr.1: 43545251-43545251 on Build GRCh38
rs780606374 Chr.1: 43545121-43545121 on Build GRCh38
rs1055896338 Chr.1: 43545191-43545191 on Build GRCh38
rs373829166 Chr.1: 43545118-43545118 on Build GRCh38
rs546168990 Chr.1: 43545414-43545414 on Build GRCh38
rs566546362 Chr.1: 43545369-43545369 on Build GRCh38
rs189332496 Chr.1: 43545214-43545214 on Build GRCh38
rs751189277 Chr.1: 43545053-43545053 on Build GRCh38
rs1008301013 Chr.1: 43545349-43545349 on Build GRCh38
rs754414070 Chr.1: 43545025-43545025 on Build GRCh38
rs766128034 Chr.1: 43545180-43545180 on Build GRCh38
rs78406870 Chr.1: 43545390-43545390 on Build GRCh38
rs180687526 Chr.1: 43545307-43545307 on Build GRCh38
rs907234062 Chr.1: 43545041-43545041 on Build GRCh38
rs574507239 Chr.1: 43545389-43545389 on Build GRCh38
rs370234203 Chr.1: 43545105-43545105 on Build GRCh38
rs563605163 Chr.1: 43545122-43545122 on Build GRCh38
rs376974470 Chr.1: 43545104-43545104 on Build GRCh38
rs568062556 Chr.1: 43545249-43545249 on Build GRCh38
rs187216097 Chr.1: 43545385-43545385 on Build GRCh38
rs371739340 Chr.1: 43545177-43545177 on Build GRCh38
rs577499041 Chr.1: 43545412-43545412 on Build GRCh38
rs145508048 Chr.1: 43545127-43545127 on Build GRCh38
rs942589829 Chr.1: 43544974-43544974 on Build GRCh38
rs765964748 Chr.1: 43545052-43545052 on Build GRCh38
rs368002739 Chr.1: 43545250-43545250 on Build GRCh38
rs535122327 Chr.1: 43545376-43545376 on Build GRCh38
rs767978306 Chr.1: 43544987-43544987 on Build GRCh38
rs964335917 Chr.1: 43545099-43545099 on Build GRCh38
rs892938910 Chr.1: 43545012-43545012 on Build GRCh38
rs752168752 Chr.1: 43545268-43545268 on Build GRCh38
rs758369493 Chr.1: 43545361-43545361 on Build GRCh38
rs1041810840 Chr.1: 43545375-43545375 on Build GRCh38

View Full Product Details