Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 44783701-44784222 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BEST4
Entrez Gene ID
266675
Gene Name
bestrophin 4
Gene Aliases
VMD2L2
UniGene
Hs.302513
Gene Chromosome Location
Chr.1: 44781836-44792828 on Build GRCh38
Transcripts
NM_153274.2 XM_017001020.1 XM_017001021.1 XM_017001022.1 XM_017001023.1

SNP Detail

SNP ID Chromosome Location
rs930259916 Chr.1: 44783864-44783864 on Build GRCh38
rs944433624 Chr.1: 44784201-44784201 on Build GRCh38
rs897886304 Chr.1: 44783924-44783924 on Build GRCh38
rs1051393977 Chr.1: 44783954-44783954 on Build GRCh38
rs981992154 Chr.1: 44783782-44783782 on Build GRCh38
rs1031735573 Chr.1: 44784091-44784091 on Build GRCh38
rs566438500 Chr.1: 44784036-44784036 on Build GRCh38
rs1031683492 Chr.1: 44783724-44783724 on Build GRCh38
rs904541612 Chr.1: 44783988-44783988 on Build GRCh38
rs928798907 Chr.1: 44783785-44783785 on Build GRCh38
rs751691010 Chr.1: 44783788-44783788 on Build GRCh38
rs770783054 Chr.1: 44784198-44784198 on Build GRCh38
rs1043613279 Chr.1: 44783944-44783944 on Build GRCh38
rs970707793 Chr.1: 44783787-44783787 on Build GRCh38
rs1024890318 Chr.1: 44784115-44784115 on Build GRCh38
rs143236734 Chr.1: 44784133-44784133 on Build GRCh38
rs917536469 Chr.1: 44783766-44783766 on Build GRCh38
rs778827510 Chr.1: 44784176-44784176 on Build GRCh38
rs535453472 Chr.1: 44784065-44784065 on Build GRCh38
rs931890360 Chr.1: 44783936-44783936 on Build GRCh38
rs77982586 Chr.1: 44783820-44783820 on Build GRCh38
rs1014458310 Chr.1: 44784095-44784095 on Build GRCh38
rs543801847 Chr.1: 44784144-44784144 on Build GRCh38
rs539514969 Chr.1: 44783928-44783928 on Build GRCh38
rs992933088 Chr.1: 44783749-44783749 on Build GRCh38
rs951708930 Chr.1: 44784151-44784151 on Build GRCh38
rs1036458432 Chr.1: 44783879-44783879 on Build GRCh38
rs763031507 Chr.1: 44784181-44784181 on Build GRCh38
rs967585507 Chr.1: 44783759-44783759 on Build GRCh38
rs543999956 Chr.1: 44783747-44783747 on Build GRCh38
rs1019957912 Chr.1: 44783754-44783754 on Build GRCh38
rs561057731 Chr.1: 44783930-44783930 on Build GRCh38
rs944543586 Chr.1: 44783887-44783887 on Build GRCh38
rs781304590 Chr.1: 44783869-44783869 on Build GRCh38
rs891868615 Chr.1: 44784092-44784092 on Build GRCh38
rs575086618 Chr.1: 44784135-44784135 on Build GRCh38
rs983211371 Chr.1: 44783866-44783866 on Build GRCh38
rs983094970 Chr.1: 44784174-44784174 on Build GRCh38

View Full Product Details