Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 45339549-45340085 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MUTYH
Entrez Gene ID
4595
Gene Name
mutY DNA glycosylase
Gene Aliases
MYH
UniGene
Hs.271353
Gene Chromosome Location
Chr.1: 45329242-45340925 on Build GRCh38
Transcripts
NM_001048171.1 NM_001048172.1 NM_001048173.1 NM_001048174.1 NM_001128425.1 NM_001293190.1 NM_001293191.1 NM_001293192.1 NM_001293195.1 NM_001293196.1 NM_012222.2 XM_011541498.1 XM_011541499.1 XM_011541501.2 XM_011541502.2 XM_011541503.2 XM_011541504.2 XM_011541505.2 XM_011541506.1 XM_017001331.1 XM_017001332.1 XM_017001334.1 XM_017001335.1 XM_017001336.1 XM_017001337.1 XM_017001338.1 XM_017001339.1 NM_025077.3 XM_005270412.3 XM_005270413.4 XM_011540569.2

Gene Symbol
TOE1
Entrez Gene ID
114034
Gene Name
target of EGR1, member 1 (nuclear)
Gene Aliases
hCaf1z
UniGene
Hs.525091
Gene Chromosome Location
Chr.1: 45339457-45343978 on Build GRCh38
Transcripts
NM_001048171.1 NM_001048172.1 NM_001048173.1 NM_001048174.1 NM_001128425.1 NM_001293190.1 NM_001293191.1 NM_001293192.1 NM_001293195.1 NM_001293196.1 NM_012222.2 XM_011541498.1 XM_011541499.1 XM_011541501.2 XM_011541502.2 XM_011541503.2 XM_011541504.2 XM_011541505.2 XM_011541506.1 XM_017001331.1 XM_017001332.1 XM_017001334.1 XM_017001335.1 XM_017001336.1 XM_017001337.1 XM_017001338.1 XM_017001339.1 NM_025077.3 XM_005270412.3 XM_005270413.4 XM_011540569.2

SNP Detail

SNP ID Chromosome Location
rs963531958 Chr.1: 45339618-45339618 on Build GRCh38
rs772606790 Chr.1: 45339878-45339878 on Build GRCh38
rs758523512 Chr.1: 45339909-45339909 on Build GRCh38
rs537932462 Chr.1: 45340044-45340044 on Build GRCh38
rs756216661 Chr.1: 45340004-45340004 on Build GRCh38
rs780193031 Chr.1: 45339913-45339913 on Build GRCh38
rs1015460326 Chr.1: 45339682-45339682 on Build GRCh38
rs894805752 Chr.1: 45339802-45339802 on Build GRCh38
rs571304759 Chr.1: 45340062-45340062 on Build GRCh38
rs965093956 Chr.1: 45339960-45339960 on Build GRCh38
rs568514678 Chr.1: 45340005-45340005 on Build GRCh38
rs545047858 Chr.1: 45339756-45339756 on Build GRCh38
rs751532568 Chr.1: 45339916-45339916 on Build GRCh38
rs943435081 Chr.1: 45339685-45339685 on Build GRCh38
rs998064862 Chr.1: 45340042-45340042 on Build GRCh38
rs770414609 Chr.1: 45339940-45339940 on Build GRCh38
rs776043801 Chr.1: 45339951-45339951 on Build GRCh38
rs1026511507 Chr.1: 45339636-45339636 on Build GRCh38
rs769022410 Chr.1: 45339961-45339961 on Build GRCh38
rs551403495 Chr.1: 45340056-45340056 on Build GRCh38
rs146380711 Chr.1: 45339638-45339638 on Build GRCh38
rs542719516 Chr.1: 45339605-45339605 on Build GRCh38
rs748796325 Chr.1: 45339937-45339937 on Build GRCh38
rs1011974836 Chr.1: 45339820-45339820 on Build GRCh38
rs1035136860 Chr.1: 45339584-45339584 on Build GRCh38
rs115629902 Chr.1: 45339595-45339595 on Build GRCh38
rs185172591 Chr.1: 45339952-45339952 on Build GRCh38
rs3219469 Chr.1: 45339741-45339741 on Build GRCh38
rs1003985418 Chr.1: 45339998-45339998 on Build GRCh38
rs767958560 Chr.1: 45339794-45339794 on Build GRCh38
rs376824989 Chr.1: 45339967-45339967 on Build GRCh38
rs974766644 Chr.1: 45339962-45339962 on Build GRCh38
rs549470824 Chr.1: 45339933-45339933 on Build GRCh38
rs1004840661 Chr.1: 45339708-45339708 on Build GRCh38
rs780788634 Chr.1: 45339927-45339927 on Build GRCh38
rs936784358 Chr.1: 45339789-45339790 on Build GRCh38
rs954131709 Chr.1: 45340057-45340057 on Build GRCh38
rs974828783 Chr.1: 45339722-45339722 on Build GRCh38
rs1046022926 Chr.1: 45339604-45339604 on Build GRCh38
rs894886600 Chr.1: 45339593-45339593 on Build GRCh38
rs750433216 Chr.1: 45339899-45339899 on Build GRCh38
rs775952036 Chr.1: 45339885-45339885 on Build GRCh38
rs761085013 Chr.1: 45339886-45339886 on Build GRCh38
rs925381523 Chr.1: 45339787-45339787 on Build GRCh38
rs764439480 Chr.1: 45339891-45339891 on Build GRCh38
rs769355993 Chr.1: 45339856-45339856 on Build GRCh38
rs762127548 Chr.1: 45339982-45339982 on Build GRCh38
rs1032778925 Chr.1: 45340058-45340058 on Build GRCh38
rs370343800 Chr.1: 45339626-45339626 on Build GRCh38
rs986134085 Chr.1: 45340058-45340058 on Build GRCh38
rs752006773 Chr.1: 45339772-45339772 on Build GRCh38
rs3219468 Chr.1: 45339894-45339894 on Build GRCh38
rs911916414 Chr.1: 45339680-45339680 on Build GRCh38
rs765382093 Chr.1: 45339895-45339895 on Build GRCh38
rs1048565967 Chr.1: 45339988-45339988 on Build GRCh38
rs901016018 Chr.1: 45340007-45340007 on Build GRCh38
rs776814505 Chr.1: 45339980-45339980 on Build GRCh38
rs1026733287 Chr.1: 45339637-45339637 on Build GRCh38
rs946516871 Chr.1: 45339750-45339750 on Build GRCh38
rs760116695 Chr.1: 45339873-45339873 on Build GRCh38
rs987488636 Chr.1: 45339674-45339674 on Build GRCh38
rs777448905 Chr.1: 45339936-45339936 on Build GRCh38
rs937685923 Chr.1: 45339953-45339954 on Build GRCh38
rs560739226 Chr.1: 45339917-45339917 on Build GRCh38
rs988138269 Chr.1: 45339931-45339931 on Build GRCh38
rs772169586 Chr.1: 45339963-45339963 on Build GRCh38
rs372487707 Chr.1: 45339862-45339862 on Build GRCh38
rs77949009 Chr.1: 45339780-45339780 on Build GRCh38
rs769287218 Chr.1: 45339935-45339935 on Build GRCh38
rs888185369 Chr.1: 45339993-45339993 on Build GRCh38
rs562412204 Chr.1: 45339667-45339667 on Build GRCh38
rs762711011 Chr.1: 45339615-45339615 on Build GRCh38
rs529425621 Chr.1: 45339932-45339932 on Build GRCh38
rs939297908 Chr.1: 45339782-45339782 on Build GRCh38

View Full Product Details