Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 45342192-45342693 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TOE1
Entrez Gene ID
114034
Gene Name
target of EGR1, member 1 (nuclear)
Gene Aliases
hCaf1z
UniGene
Hs.525091
Gene Chromosome Location
Chr.1: 45339457-45343978 on Build GRCh38
Transcripts
NM_025077.3 XM_005270412.3 XM_005270413.4 XM_011540569.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6805986 c.695C>T p.A232V Chr.1: 45342586-45342586 on Build GRCh38
1602466 c.699T>G p.A233A Chr.1: 45342590-45342590 on Build GRCh38
5381105 c.744C>T p.F248F Chr.1: 45342635-45342635 on Build GRCh38
5443491 c.593T>G p.F198C Chr.1: 45342484-45342484 on Build GRCh38
6805982 c.646G>T p.A216S Chr.1: 45342537-45342537 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1040844847 Chr.1: 45342252-45342252 on Build GRCh38
rs775471846 Chr.1: 45342637-45342637 on Build GRCh38
rs753255466 Chr.1: 45342602-45342602 on Build GRCh38
rs746935900 Chr.1: 45342659-45342659 on Build GRCh38
rs766438052 Chr.1: 45342334-45342334 on Build GRCh38
rs769225253 Chr.1: 45342412-45342412 on Build GRCh38
rs142753932 Chr.1: 45342463-45342463 on Build GRCh38
rs145986883 Chr.1: 45342411-45342411 on Build GRCh38
rs778471696 Chr.1: 45342443-45342443 on Build GRCh38
rs767479473 Chr.1: 45342557-45342557 on Build GRCh38
rs747546453 Chr.1: 45342396-45342396 on Build GRCh38
rs893373736 Chr.1: 45342311-45342311 on Build GRCh38
rs374271681 Chr.1: 45342465-45342465 on Build GRCh38
rs35900406 Chr.1: 45342214-45342215 on Build GRCh38
rs371271084 Chr.1: 45342561-45342561 on Build GRCh38
rs377615673 Chr.1: 45342472-45342472 on Build GRCh38
rs780185153 Chr.1: 45342291-45342291 on Build GRCh38
rs891470892 Chr.1: 45342635-45342635 on Build GRCh38
rs1025115069 Chr.1: 45342416-45342416 on Build GRCh38
rs759497807 Chr.1: 45342428-45342428 on Build GRCh38
rs756103040 Chr.1: 45342267-45342267 on Build GRCh38
rs963268111 Chr.1: 45342556-45342556 on Build GRCh38
rs570113391 Chr.1: 45342292-45342292 on Build GRCh38
rs904431918 Chr.1: 45342570-45342570 on Build GRCh38
rs759563427 Chr.1: 45342350-45342351 on Build GRCh38
rs201403165 Chr.1: 45342608-45342608 on Build GRCh38
rs1049616324 Chr.1: 45342537-45342537 on Build GRCh38
rs781748718 Chr.1: 45342466-45342466 on Build GRCh38
rs1051192282 Chr.1: 45342241-45342241 on Build GRCh38
rs770222170 Chr.1: 45342468-45342468 on Build GRCh38
rs185406109 Chr.1: 45342527-45342527 on Build GRCh38
rs752569685 Chr.1: 45342435-45342435 on Build GRCh38
rs767258677 Chr.1: 45342434-45342434 on Build GRCh38
rs550198834 Chr.1: 45342290-45342290 on Build GRCh38
rs760414433 Chr.1: 45342436-45342436 on Build GRCh38
rs368907887 Chr.1: 45342354-45342354 on Build GRCh38
rs1011804373 Chr.1: 45342360-45342360 on Build GRCh38
rs538952271 Chr.1: 45342401-45342401 on Build GRCh38
rs761431701 Chr.1: 45342547-45342547 on Build GRCh38
rs1049691829 Chr.1: 45342297-45342297 on Build GRCh38
rs780430714 Chr.1: 45342393-45342393 on Build GRCh38
rs940085713 Chr.1: 45342471-45342471 on Build GRCh38
rs771133822 Chr.1: 45342420-45342420 on Build GRCh38
rs757795522 Chr.1: 45342585-45342585 on Build GRCh38
rs535191956 Chr.1: 45342445-45342445 on Build GRCh38
rs912679288 Chr.1: 45342272-45342272 on Build GRCh38
rs373393600 Chr.1: 45342377-45342377 on Build GRCh38
rs773514604 Chr.1: 45342414-45342414 on Build GRCh38
rs759496191 Chr.1: 45342503-45342503 on Build GRCh38
rs980931587 Chr.1: 45342438-45342438 on Build GRCh38
rs267598623 Chr.1: 45342442-45342442 on Build GRCh38
rs751451617 Chr.1: 45342341-45342341 on Build GRCh38
rs3748647 Chr.1: 45342500-45342500 on Build GRCh38
rs745974209 Chr.1: 45342612-45342612 on Build GRCh38
rs1002995018 Chr.1: 45342425-45342425 on Build GRCh38
rs774216136 Chr.1: 45342424-45342424 on Build GRCh38
rs763750237 Chr.1: 45342439-45342439 on Build GRCh38
rs999848167 Chr.1: 45342383-45342383 on Build GRCh38
rs1031785220 Chr.1: 45342387-45342387 on Build GRCh38
rs756609909 Chr.1: 45342604-45342604 on Build GRCh38
rs755114749 Chr.1: 45342270-45342270 on Build GRCh38
rs376295493 Chr.1: 45342603-45342603 on Build GRCh38
rs932453883 Chr.1: 45342536-45342536 on Build GRCh38
rs781727641 Chr.1: 45342594-45342594 on Build GRCh38
rs901008823 Chr.1: 45342280-45342280 on Build GRCh38
rs137896624 Chr.1: 45342609-45342609 on Build GRCh38
rs752344309 Chr.1: 45342357-45342357 on Build GRCh38
rs770967324 Chr.1: 45342478-45342478 on Build GRCh38
rs932525889 Chr.1: 45342287-45342287 on Build GRCh38
rs1013983292 Chr.1: 45342646-45342646 on Build GRCh38
rs749976929 Chr.1: 45342581-45342581 on Build GRCh38
rs756699552 Chr.1: 45342389-45342389 on Build GRCh38
rs533647558 Chr.1: 45342441-45342441 on Build GRCh38
rs772071374 Chr.1: 45342506-45342506 on Build GRCh38
rs777030573 Chr.1: 45342409-45342409 on Build GRCh38
rs778955773 Chr.1: 45342610-45342610 on Build GRCh38
rs35640482 Chr.1: 45342368-45342369 on Build GRCh38
rs755734398 Chr.1: 45342366-45342366 on Build GRCh38
rs145244176 Chr.1: 45342255-45342255 on Build GRCh38
rs778263701 Chr.1: 45342607-45342607 on Build GRCh38
rs536731618 Chr.1: 45342273-45342273 on Build GRCh38
rs771979228 Chr.1: 45342636-45342636 on Build GRCh38
rs754800220 Chr.1: 45342342-45342342 on Build GRCh38
rs575022056 Chr.1: 45342529-45342529 on Build GRCh38
rs908541095 Chr.1: 45342459-45342459 on Build GRCh38
rs760467874 Chr.1: 45342513-45342513 on Build GRCh38
rs775701267 Chr.1: 45342604-45342605 on Build GRCh38
rs777366352 Chr.1: 45342369-45342369 on Build GRCh38
rs902862301 Chr.1: 45342403-45342403 on Build GRCh38
rs763134006 Chr.1: 45342417-45342417 on Build GRCh38
rs747000027 Chr.1: 45342284-45342284 on Build GRCh38
rs767923015 Chr.1: 45342444-45342444 on Build GRCh38

View Full Product Details