Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 14039575-14040080 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL27RA
Entrez Gene ID
9466
Gene Name
interleukin 27 receptor subunit alpha
Gene Aliases
CRL1, IL-27RA, IL27R, TCCR, WSX1, zcytor1
UniGene
Hs.132781
Gene Chromosome Location
Chr.19: 14031740-14052919 on Build GRCh38
Transcripts
NM_004843.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3281531 c.421G>A p.E141K Chr.19: 14039797-14039797 on Build GRCh38
3529291 c.309C>T p.L103L Chr.19: 14039598-14039598 on Build GRCh38
7170521 c.395G>A p.R132Q Chr.19: 14039771-14039771 on Build GRCh38
1304082 c.427G>A p.D143N Chr.19: 14039803-14039803 on Build GRCh38
4414455 c.320G>A p.G107D Chr.19: 14039609-14039609 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs977488200 Chr.19: 14039818-14039818 on Build GRCh38
rs200728245 Chr.19: 14039693-14039693 on Build GRCh38
rs1046851728 Chr.19: 14039968-14039968 on Build GRCh38
rs199904982 Chr.19: 14039826-14039826 on Build GRCh38
rs757216991 Chr.19: 14039917-14039917 on Build GRCh38
rs754756373 Chr.19: 14039627-14039627 on Build GRCh38
rs149150973 Chr.19: 14039882-14039882 on Build GRCh38
rs373267847 Chr.19: 14039698-14039698 on Build GRCh38
rs764852881 Chr.19: 14039890-14039890 on Build GRCh38
rs770992284 Chr.19: 14039731-14039731 on Build GRCh38
rs763438386 Chr.19: 14039690-14039690 on Build GRCh38
rs552269402 Chr.19: 14040006-14040006 on Build GRCh38
rs369206630 Chr.19: 14039951-14039951 on Build GRCh38
rs529787928 Chr.19: 14039608-14039608 on Build GRCh38
rs377689625 Chr.19: 14039719-14039719 on Build GRCh38
rs372276918 Chr.19: 14039945-14039945 on Build GRCh38
rs776197914 Chr.19: 14039881-14039881 on Build GRCh38
rs555116302 Chr.19: 14039937-14039937 on Build GRCh38
rs375264330 Chr.19: 14039647-14039647 on Build GRCh38
rs763238631 Chr.19: 14039768-14039768 on Build GRCh38
rs747540021 Chr.19: 14039956-14039956 on Build GRCh38
rs749993806 Chr.19: 14039617-14039617 on Build GRCh38
rs781160329 Chr.19: 14039742-14039742 on Build GRCh38
rs186420427 Chr.19: 14039942-14039942 on Build GRCh38
rs373940066 Chr.19: 14039811-14039811 on Build GRCh38
rs376709516 Chr.19: 14039777-14039777 on Build GRCh38
rs772509590 Chr.19: 14039878-14039878 on Build GRCh38
rs765045284 Chr.19: 14039785-14039785 on Build GRCh38
rs540624044 Chr.19: 14040036-14040036 on Build GRCh38
rs145198960 Chr.19: 14039618-14039618 on Build GRCh38
rs1025371138 Chr.19: 14039748-14039748 on Build GRCh38
rs368706518 Chr.19: 14039626-14039626 on Build GRCh38
rs746082483 Chr.19: 14039752-14039752 on Build GRCh38
rs770102258 Chr.19: 14039671-14039671 on Build GRCh38
rs770919999 Chr.19: 14039651-14039651 on Build GRCh38
rs751472029 Chr.19: 14039823-14039823 on Build GRCh38
rs371086964 Chr.19: 14039765-14039765 on Build GRCh38
rs774876922 Chr.19: 14039770-14039770 on Build GRCh38
rs774621762 Chr.19: 14039863-14039863 on Build GRCh38
rs376795280 Chr.19: 14039946-14039946 on Build GRCh38
rs74824519 Chr.19: 14040047-14040047 on Build GRCh38
rs1019697719 Chr.19: 14039699-14039699 on Build GRCh38
rs143233265 Chr.19: 14039898-14039898 on Build GRCh38
rs758208255 Chr.19: 14039813-14039813 on Build GRCh38
rs772485804 Chr.19: 14039950-14039950 on Build GRCh38
rs748668943 Chr.19: 14039871-14039871 on Build GRCh38
rs772182491 Chr.19: 14039641-14039641 on Build GRCh38
rs775882702 Chr.19: 14039687-14039687 on Build GRCh38
rs746004197 Chr.19: 14039831-14039831 on Build GRCh38
rs752595176 Chr.19: 14039717-14039717 on Build GRCh38
rs758946711 Chr.19: 14039615-14039615 on Build GRCh38
rs768973217 Chr.19: 14039856-14039856 on Build GRCh38
rs776854880 Chr.19: 14039658-14039658 on Build GRCh38
rs757180872 Chr.19: 14039824-14039824 on Build GRCh38
rs374053127 Chr.19: 14039771-14039771 on Build GRCh38
rs763916613 Chr.19: 14039899-14039899 on Build GRCh38
rs754630325 Chr.19: 14039708-14039708 on Build GRCh38
rs750374493 Chr.19: 14039931-14039931 on Build GRCh38
rs373611906 Chr.19: 14039670-14039670 on Build GRCh38
rs778825813 Chr.19: 14039716-14039716 on Build GRCh38
rs539025031 Chr.19: 14039841-14039841 on Build GRCh38
rs765418656 Chr.19: 14039968-14039970 on Build GRCh38
rs749538342 Chr.19: 14039847-14039847 on Build GRCh38
rs577104639 Chr.19: 14040024-14040024 on Build GRCh38
rs756141524 Chr.19: 14039933-14039933 on Build GRCh38
rs190443624 Chr.19: 14039696-14039696 on Build GRCh38
rs58817586 Chr.19: 14039947-14039947 on Build GRCh38
rs764652272 Chr.19: 14039691-14039691 on Build GRCh38
rs1005350984 Chr.19: 14039764-14039764 on Build GRCh38
rs1005418235 Chr.19: 14040046-14040046 on Build GRCh38
rs778885144 Chr.19: 14039628-14039628 on Build GRCh38
rs368080572 Chr.19: 14039814-14039814 on Build GRCh38
rs181859110 Chr.19: 14039763-14039763 on Build GRCh38
rs779694594 Chr.19: 14039621-14039621 on Build GRCh38
rs748065051 Chr.19: 14039638-14039638 on Build GRCh38
rs371298670 Chr.19: 14039784-14039784 on Build GRCh38
rs767291532 Chr.19: 14039925-14039925 on Build GRCh38
rs765885275 Chr.19: 14039772-14039772 on Build GRCh38
rs756562006 Chr.19: 14039741-14039741 on Build GRCh38
rs538985417 Chr.19: 14039718-14039718 on Build GRCh38
rs370031536 Chr.19: 14039819-14039819 on Build GRCh38
rs536704566 Chr.19: 14039906-14039906 on Build GRCh38
rs766993532 Chr.19: 14039616-14039616 on Build GRCh38
rs567809883 Chr.19: 14039697-14039697 on Build GRCh38
rs1050715522 Chr.19: 14039688-14039688 on Build GRCh38
rs201794701 Chr.19: 14039897-14039897 on Build GRCh38
rs201656454 Chr.19: 14039646-14039646 on Build GRCh38

View Full Product Details