Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 14563052-14563553 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TECR
Entrez Gene ID
9524
Gene Name
trans-2,3-enoyl-CoA reductase
Gene Aliases
GPSN2, MRT14, SC2, TER
UniGene
Hs.515642
Gene Chromosome Location
Chr.19: 14527760-14565980 on Build GRCh38
Transcripts
NM_001321170.1 NM_138501.5 NR_038103.1 NR_038104.1 XM_006722945.1 XM_011528442.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6799247 c.67-7delC p.? Chr.19: 14563199-14563199 on Build GRCh38
4732970 c.95T>G p.I32S Chr.19: 14563234-14563234 on Build GRCh38
3282257 c.118C>A p.H40N Chr.19: 14563257-14563257 on Build GRCh38
1390963 c.79G>A p.A27T Chr.19: 14563218-14563218 on Build GRCh38
6149470 c.118+1G>C p.? Chr.19: 14563258-14563258 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776703090 Chr.19: 14563201-14563201 on Build GRCh38
rs746920276 Chr.19: 14563288-14563288 on Build GRCh38
rs748570128 Chr.19: 14563282-14563283 on Build GRCh38
rs111501133 Chr.19: 14563258-14563258 on Build GRCh38
rs192451814 Chr.19: 14563312-14563312 on Build GRCh38
rs891089 Chr.19: 14563423-14563423 on Build GRCh38
rs752066182 Chr.19: 14563225-14563225 on Build GRCh38
rs1041704620 Chr.19: 14563496-14563496 on Build GRCh38
rs372968700 Chr.19: 14563170-14563170 on Build GRCh38
rs994316374 Chr.19: 14563339-14563339 on Build GRCh38
rs950429148 Chr.19: 14563384-14563384 on Build GRCh38
rs747771192 Chr.19: 14563273-14563273 on Build GRCh38
rs758244290 Chr.19: 14563276-14563276 on Build GRCh38
rs879806199 Chr.19: 14563337-14563338 on Build GRCh38
rs370808218 Chr.19: 14563168-14563168 on Build GRCh38
rs556954502 Chr.19: 14563092-14563092 on Build GRCh38
rs1032569782 Chr.19: 14563109-14563109 on Build GRCh38
rs541489558 Chr.19: 14563272-14563272 on Build GRCh38
rs773366371 Chr.19: 14563114-14563114 on Build GRCh38
rs1042212095 Chr.19: 14563447-14563447 on Build GRCh38
rs879609794 Chr.19: 14563342-14563343 on Build GRCh38
rs373195944 Chr.19: 14563355-14563355 on Build GRCh38
rs188069168 Chr.19: 14563523-14563523 on Build GRCh38
rs994766168 Chr.19: 14563503-14563503 on Build GRCh38
rs748960213 Chr.19: 14563169-14563169 on Build GRCh38
rs766457201 Chr.19: 14563117-14563117 on Build GRCh38
rs145931907 Chr.19: 14563256-14563256 on Build GRCh38
rs745903458 Chr.19: 14563302-14563302 on Build GRCh38
rs932694817 Chr.19: 14563152-14563152 on Build GRCh38
rs879408244 Chr.19: 14563343-14563344 on Build GRCh38
rs924053244 Chr.19: 14563457-14563457 on Build GRCh38
rs373146902 Chr.19: 14563162-14563162 on Build GRCh38
rs746949549 Chr.19: 14563191-14563191 on Build GRCh38
rs765696958 Chr.19: 14563135-14563135 on Build GRCh38
rs568050308 Chr.19: 14563445-14563445 on Build GRCh38
rs772031577 Chr.19: 14563188-14563188 on Build GRCh38
rs1046253784 Chr.19: 14563395-14563395 on Build GRCh38
rs568873082 Chr.19: 14563524-14563524 on Build GRCh38
rs758995594 Chr.19: 14563155-14563155 on Build GRCh38
rs761747212 Chr.19: 14563107-14563107 on Build GRCh38
rs755629224 Chr.19: 14563167-14563167 on Build GRCh38
rs1014078807 Chr.19: 14563090-14563090 on Build GRCh38
rs563728766 Chr.19: 14563323-14563323 on Build GRCh38
rs965858131 Chr.19: 14563149-14563149 on Build GRCh38
rs137862909 Chr.19: 14563180-14563180 on Build GRCh38
rs879517542 Chr.19: 14563338-14563339 on Build GRCh38
rs765505868 Chr.19: 14563211-14563211 on Build GRCh38
rs902579454 Chr.19: 14563268-14563268 on Build GRCh38
rs554273185 Chr.19: 14563405-14563405 on Build GRCh38
rs187924859 Chr.19: 14563291-14563291 on Build GRCh38
rs754062540 Chr.19: 14563130-14563130 on Build GRCh38
rs945229805 Chr.19: 14563377-14563377 on Build GRCh38
rs1053750277 Chr.19: 14563475-14563475 on Build GRCh38
rs764495966 Chr.19: 14563222-14563222 on Build GRCh38
rs774386013 Chr.19: 14563180-14563181 on Build GRCh38
rs200550141 Chr.19: 14563270-14563270 on Build GRCh38
rs1033988365 Chr.19: 14563456-14563456 on Build GRCh38
rs958992871 Chr.19: 14563418-14563418 on Build GRCh38
rs76446392 Chr.19: 14563412-14563412 on Build GRCh38
rs540392987 Chr.19: 14563095-14563095 on Build GRCh38
rs1002309131 Chr.19: 14563184-14563184 on Build GRCh38
rs1019821687 Chr.19: 14563314-14563314 on Build GRCh38
rs760634686 Chr.19: 14563115-14563115 on Build GRCh38
rs925995105 Chr.19: 14563316-14563316 on Build GRCh38
rs759847557 Chr.19: 14563205-14563205 on Build GRCh38
rs768569087 Chr.19: 14563257-14563257 on Build GRCh38
rs765812213 Chr.19: 14563242-14563242 on Build GRCh38
rs770981335 Chr.19: 14563199-14563199 on Build GRCh38
rs889794007 Chr.19: 14563476-14563476 on Build GRCh38
rs199993628 Chr.19: 14563337-14563345 on Build GRCh38
rs961105760 Chr.19: 14563491-14563491 on Build GRCh38
rs185593335 Chr.19: 14563414-14563414 on Build GRCh38
rs777626678 Chr.19: 14563134-14563134 on Build GRCh38
rs1026510977 Chr.19: 14563372-14563372 on Build GRCh38
rs949207756 Chr.19: 14563241-14563241 on Build GRCh38
rs1053697842 Chr.19: 14563299-14563299 on Build GRCh38
rs535567825 Chr.19: 14563474-14563474 on Build GRCh38
rs750331140 Chr.19: 14563301-14563301 on Build GRCh38
rs201189282 Chr.19: 14563304-14563304 on Build GRCh38
rs546257217 Chr.19: 14563432-14563432 on Build GRCh38
rs777616566 Chr.19: 14563284-14563284 on Build GRCh38
rs1041291455 Chr.19: 14563121-14563121 on Build GRCh38
rs370460255 Chr.19: 14563181-14563181 on Build GRCh38
rs11539237 Chr.19: 14563203-14563203 on Build GRCh38
rs983901521 Chr.19: 14563193-14563193 on Build GRCh38
rs985618819 Chr.19: 14563118-14563118 on Build GRCh38
rs753052889 Chr.19: 14563140-14563140 on Build GRCh38
rs757780004 Chr.19: 14563235-14563235 on Build GRCh38
rs771943648 Chr.19: 14563110-14563110 on Build GRCh38
rs763449006 Chr.19: 14563218-14563218 on Build GRCh38
rs901410537 Chr.19: 14563502-14563502 on Build GRCh38
rs370284053 Chr.19: 14563426-14563426 on Build GRCh38
rs1006866665 Chr.19: 14563484-14563484 on Build GRCh38
rs386807078 Chr.19: 14563337-14563343 on Build GRCh38
rs112196248 Chr.19: 14563260-14563260 on Build GRCh38
rs1053216621 Chr.19: 14563326-14563326 on Build GRCh38
rs776029034 Chr.19: 14563217-14563217 on Build GRCh38
rs774128838 Chr.19: 14563096-14563096 on Build GRCh38
rs1033335249 Chr.19: 14563185-14563185 on Build GRCh38
rs917818570 Chr.19: 14563424-14563424 on Build GRCh38
rs776461646 Chr.19: 14563406-14563406 on Build GRCh38
rs1000267498 Chr.19: 14563165-14563166 on Build GRCh38
rs776595742 Chr.19: 14563292-14563292 on Build GRCh38
rs376415137 Chr.19: 14563164-14563164 on Build GRCh38
rs530705728 Chr.19: 14563337-14563337 on Build GRCh38
rs200634741 Chr.19: 14563189-14563189 on Build GRCh38

View Full Product Details