Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 16501061-16501548 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C19orf44
Entrez Gene ID
84167
Gene Name
chromosome 19 open reading frame 44
Gene Aliases
-
UniGene
Hs.631627
Gene Chromosome Location
Chr.19: 16496311-16521369 on Build GRCh38
Transcripts
NM_001288834.1 NM_032207.3 XM_006722920.3 XM_011528354.2 XM_017027350.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6614882 c.723T>C p.A241A Chr.19: 16501515-16501515 on Build GRCh38
6614883 c.718A>G p.S240G Chr.19: 16501510-16501510 on Build GRCh38
5501147 c.312C>A p.I104I Chr.19: 16501104-16501104 on Build GRCh38
5569429 c.620C>A p.T207K Chr.19: 16501412-16501412 on Build GRCh38
417720 c.663G>T p.L221F Chr.19: 16501455-16501455 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs728117 Chr.19: 16501200-16501200 on Build GRCh38
rs773814678 Chr.19: 16501099-16501099 on Build GRCh38
rs751613344 Chr.19: 16501292-16501292 on Build GRCh38
rs781437375 Chr.19: 16501212-16501212 on Build GRCh38
rs867651984 Chr.19: 16501265-16501265 on Build GRCh38
rs769718275 Chr.19: 16501373-16501373 on Build GRCh38
rs761287141 Chr.19: 16501100-16501100 on Build GRCh38
rs750835848 Chr.19: 16501213-16501213 on Build GRCh38
rs777296335 Chr.19: 16501220-16501220 on Build GRCh38
rs374292345 Chr.19: 16501153-16501153 on Build GRCh38
rs777530174 Chr.19: 16501415-16501415 on Build GRCh38
rs371363608 Chr.19: 16501218-16501218 on Build GRCh38
rs754009367 Chr.19: 16501111-16501111 on Build GRCh38
rs770949440 Chr.19: 16501224-16501224 on Build GRCh38
rs750592976 Chr.19: 16501267-16501267 on Build GRCh38
rs765429207 Chr.19: 16501124-16501124 on Build GRCh38
rs768975791 Chr.19: 16501391-16501391 on Build GRCh38
rs777482076 Chr.19: 16501470-16501472 on Build GRCh38
rs745819965 Chr.19: 16501477-16501477 on Build GRCh38
rs760592805 Chr.19: 16501169-16501169 on Build GRCh38
rs756481021 Chr.19: 16501302-16501302 on Build GRCh38
rs766802929 Chr.19: 16501340-16501341 on Build GRCh38
rs745744139 Chr.19: 16501372-16501372 on Build GRCh38
rs143681756 Chr.19: 16501425-16501425 on Build GRCh38
rs770662416 Chr.19: 16501172-16501172 on Build GRCh38
rs371756715 Chr.19: 16501188-16501188 on Build GRCh38
rs752375991 Chr.19: 16501307-16501307 on Build GRCh38
rs563097198 Chr.19: 16501159-16501159 on Build GRCh38
rs764114089 Chr.19: 16501286-16501286 on Build GRCh38
rs748915898 Chr.19: 16501087-16501087 on Build GRCh38
rs1056904636 Chr.19: 16501084-16501084 on Build GRCh38
rs754243003 Chr.19: 16501454-16501454 on Build GRCh38
rs758227350 Chr.19: 16501393-16501393 on Build GRCh38
rs756073383 Chr.19: 16501478-16501478 on Build GRCh38
rs774428481 Chr.19: 16501397-16501397 on Build GRCh38
rs776437885 Chr.19: 16501179-16501179 on Build GRCh38
rs201664638 Chr.19: 16501487-16501487 on Build GRCh38
rs759470948 Chr.19: 16501231-16501231 on Build GRCh38
rs751166122 Chr.19: 16501467-16501467 on Build GRCh38
rs150929772 Chr.19: 16501104-16501104 on Build GRCh38
rs543362543 Chr.19: 16501140-16501140 on Build GRCh38
rs754430509 Chr.19: 16501368-16501369 on Build GRCh38
rs776402055 Chr.19: 16501229-16501229 on Build GRCh38
rs762007255 Chr.19: 16501398-16501398 on Build GRCh38
rs778492628 Chr.19: 16501138-16501138 on Build GRCh38
rs753970847 Chr.19: 16501161-16501161 on Build GRCh38
rs544298324 Chr.19: 16501295-16501295 on Build GRCh38
rs755518095 Chr.19: 16501402-16501402 on Build GRCh38
rs148952679 Chr.19: 16501381-16501381 on Build GRCh38
rs368008120 Chr.19: 16501158-16501158 on Build GRCh38
rs773237666 Chr.19: 16501513-16501513 on Build GRCh38
rs775387544 Chr.19: 16501380-16501380 on Build GRCh38
rs728116 Chr.19: 16501167-16501167 on Build GRCh38
rs938368853 Chr.19: 16501439-16501439 on Build GRCh38
rs758018942 Chr.19: 16501311-16501311 on Build GRCh38
rs748263116 Chr.19: 16501498-16501498 on Build GRCh38
rs781556993 Chr.19: 16501142-16501142 on Build GRCh38
rs756715466 Chr.19: 16501483-16501484 on Build GRCh38
rs749546317 Chr.19: 16501384-16501384 on Build GRCh38
rs73513157 Chr.19: 16501275-16501275 on Build GRCh38
rs772218577 Chr.19: 16501512-16501512 on Build GRCh38
rs147828681 Chr.19: 16501216-16501216 on Build GRCh38
rs768163311 Chr.19: 16501090-16501090 on Build GRCh38
rs751027426 Chr.19: 16501484-16501485 on Build GRCh38
rs760961062 Chr.19: 16501324-16501325 on Build GRCh38
rs749152333 Chr.19: 16501480-16501480 on Build GRCh38
rs781249551 Chr.19: 16501349-16501349 on Build GRCh38
rs372943238 Chr.19: 16501458-16501458 on Build GRCh38
rs761780981 Chr.19: 16501293-16501293 on Build GRCh38
rs1038271332 Chr.19: 16501284-16501284 on Build GRCh38
rs769700747 Chr.19: 16501258-16501258 on Build GRCh38
rs113496336 Chr.19: 16501317-16501317 on Build GRCh38
rs141306136 Chr.19: 16501221-16501221 on Build GRCh38
rs775869881 Chr.19: 16501264-16501264 on Build GRCh38
rs756795441 Chr.19: 16501468-16501468 on Build GRCh38
rs373713416 Chr.19: 16501518-16501518 on Build GRCh38
rs770343725 Chr.19: 16501342-16501342 on Build GRCh38
rs759382859 Chr.19: 16501181-16501181 on Build GRCh38
rs780327720 Chr.19: 16501303-16501303 on Build GRCh38
rs768336435 Chr.19: 16501493-16501493 on Build GRCh38
rs765597437 Chr.19: 16501184-16501184 on Build GRCh38
rs367607617 Chr.19: 16501209-16501209 on Build GRCh38
rs774641690 Chr.19: 16501494-16501494 on Build GRCh38
rs756462546 Chr.19: 16501214-16501214 on Build GRCh38
rs79590176 Chr.19: 16501156-16501156 on Build GRCh38
rs891447595 Chr.19: 16501429-16501429 on Build GRCh38
rs200970238 Chr.19: 16501479-16501479 on Build GRCh38
rs373902605 Chr.19: 16501434-16501434 on Build GRCh38
rs746569479 Chr.19: 16501340-16501340 on Build GRCh38
rs368718509 Chr.19: 16501110-16501110 on Build GRCh38
rs188780108 Chr.19: 16501519-16501519 on Build GRCh38
rs192562625 Chr.19: 16501294-16501294 on Build GRCh38
rs113147081 Chr.19: 16501260-16501260 on Build GRCh38
rs764178955 Chr.19: 16501195-16501195 on Build GRCh38
rs760442677 Chr.19: 16501105-16501105 on Build GRCh38
rs763865304 Chr.19: 16501464-16501464 on Build GRCh38
rs978385990 Chr.19: 16501424-16501424 on Build GRCh38
rs755210580 Chr.19: 16501112-16501112 on Build GRCh38
rs771573597 Chr.19: 16501219-16501219 on Build GRCh38
rs747540279 Chr.19: 16501139-16501139 on Build GRCh38
rs370995893 Chr.19: 16501132-16501132 on Build GRCh38
rs767434785 Chr.19: 16501402-16501402 on Build GRCh38

View Full Product Details