Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 17365206-17365715 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLVAP
Entrez Gene ID
83483
Gene Name
plasmalemma vesicle associated protein
Gene Aliases
FELS, PV-1, PV1, gp68
UniGene
Hs.107125
Gene Chromosome Location
Chr.19: 17351448-17377349 on Build GRCh38
Transcripts
NM_031310.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
992779 c.1170C>G p.I390M Chr.19: 17365295-17365295 on Build GRCh38
5614616 c.863G>T p.R288L Chr.19: 17365602-17365602 on Build GRCh38
438903 c.1116G>A p.A372A Chr.19: 17365349-17365349 on Build GRCh38
5588683 c.864G>A p.R288R Chr.19: 17365601-17365601 on Build GRCh38
1391453 c.1115C>T p.A372V Chr.19: 17365350-17365350 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756960713 Chr.19: 17365582-17365582 on Build GRCh38
rs954262919 Chr.19: 17365302-17365302 on Build GRCh38
rs778293545 Chr.19: 17365434-17365434 on Build GRCh38
rs911676579 Chr.19: 17365328-17365328 on Build GRCh38
rs780468405 Chr.19: 17365660-17365660 on Build GRCh38
rs141514915 Chr.19: 17365594-17365594 on Build GRCh38
rs759726274 Chr.19: 17365513-17365513 on Build GRCh38
rs756084593 Chr.19: 17365423-17365423 on Build GRCh38
rs552294764 Chr.19: 17365676-17365676 on Build GRCh38
rs771680538 Chr.19: 17365441-17365441 on Build GRCh38
rs200491210 Chr.19: 17365576-17365576 on Build GRCh38
rs772475301 Chr.19: 17365603-17365603 on Build GRCh38
rs769469168 Chr.19: 17365685-17365685 on Build GRCh38
rs759964317 Chr.19: 17365465-17365465 on Build GRCh38
rs201354703 Chr.19: 17365440-17365440 on Build GRCh38
rs141106508 Chr.19: 17365401-17365401 on Build GRCh38
rs756372237 Chr.19: 17365659-17365659 on Build GRCh38
rs543086219 Chr.19: 17365641-17365641 on Build GRCh38
rs772667508 Chr.19: 17365550-17365550 on Build GRCh38
rs776456257 Chr.19: 17365609-17365609 on Build GRCh38
rs61739312 Chr.19: 17365669-17365669 on Build GRCh38
rs750288854 Chr.19: 17365410-17365410 on Build GRCh38
rs934707036 Chr.19: 17365545-17365545 on Build GRCh38
rs147035073 Chr.19: 17365568-17365568 on Build GRCh38
rs780097048 Chr.19: 17365424-17365424 on Build GRCh38
rs73016435 Chr.19: 17365336-17365336 on Build GRCh38
rs150753312 Chr.19: 17365626-17365626 on Build GRCh38
rs374123136 Chr.19: 17365590-17365590 on Build GRCh38
rs892262273 Chr.19: 17365561-17365561 on Build GRCh38
rs771318318 Chr.19: 17365460-17365460 on Build GRCh38
rs377281215 Chr.19: 17365370-17365370 on Build GRCh38
rs138789000 Chr.19: 17365519-17365519 on Build GRCh38
rs138584008 Chr.19: 17365569-17365569 on Build GRCh38
rs775604876 Chr.19: 17365524-17365524 on Build GRCh38
rs376519104 Chr.19: 17365503-17365503 on Build GRCh38
rs760495218 Chr.19: 17365474-17365474 on Build GRCh38
rs539524093 Chr.19: 17365463-17365463 on Build GRCh38
rs537070619 Chr.19: 17365429-17365429 on Build GRCh38
rs149357796 Chr.19: 17365553-17365553 on Build GRCh38
rs766293089 Chr.19: 17365476-17365476 on Build GRCh38
rs745795317 Chr.19: 17365358-17365358 on Build GRCh38
rs1028750051 Chr.19: 17365653-17365653 on Build GRCh38
rs779118041 Chr.19: 17365667-17365667 on Build GRCh38
rs1028607662 Chr.19: 17365634-17365634 on Build GRCh38
rs374998380 Chr.19: 17365536-17365536 on Build GRCh38
rs35339649 Chr.19: 17365267-17365267 on Build GRCh38
rs111681752 Chr.19: 17365679-17365679 on Build GRCh38
rs144053602 Chr.19: 17365578-17365578 on Build GRCh38
rs771478951 Chr.19: 17365402-17365402 on Build GRCh38
rs200728670 Chr.19: 17365648-17365648 on Build GRCh38
rs753976923 Chr.19: 17365293-17365293 on Build GRCh38
rs561048879 Chr.19: 17365588-17365588 on Build GRCh38
rs111831522 Chr.19: 17365285-17365285 on Build GRCh38
rs375725191 Chr.19: 17365241-17365241 on Build GRCh38
rs763597063 Chr.19: 17365579-17365579 on Build GRCh38
rs750965023 Chr.19: 17365265-17365265 on Build GRCh38
rs756174826 Chr.19: 17365368-17365368 on Build GRCh38
rs369525652 Chr.19: 17365409-17365409 on Build GRCh38
rs781372688 Chr.19: 17365456-17365457 on Build GRCh38
rs758015518 Chr.19: 17365495-17365495 on Build GRCh38
rs141002499 Chr.19: 17365459-17365459 on Build GRCh38
rs150210307 Chr.19: 17365502-17365502 on Build GRCh38
rs375676127 Chr.19: 17365472-17365472 on Build GRCh38
rs201285895 Chr.19: 17365635-17365635 on Build GRCh38
rs1028613392 Chr.19: 17365485-17365485 on Build GRCh38
rs770251971 Chr.19: 17365523-17365523 on Build GRCh38
rs146532352 Chr.19: 17365392-17365392 on Build GRCh38
rs202200013 Chr.19: 17365518-17365518 on Build GRCh38
rs767798441 Chr.19: 17365254-17365254 on Build GRCh38
rs144011339 Chr.19: 17365649-17365649 on Build GRCh38
rs376756648 Chr.19: 17365565-17365565 on Build GRCh38
rs148762093 Chr.19: 17365593-17365593 on Build GRCh38
rs142500391 Chr.19: 17365344-17365344 on Build GRCh38
rs112072429 Chr.19: 17365425-17365425 on Build GRCh38
rs748929936 Chr.19: 17365319-17365319 on Build GRCh38
rs761158492 Chr.19: 17365393-17365393 on Build GRCh38
rs896334725 Chr.19: 17365598-17365598 on Build GRCh38
rs147299275 Chr.19: 17365589-17365589 on Build GRCh38
rs370148435 Chr.19: 17365631-17365631 on Build GRCh38
rs1052700069 Chr.19: 17365548-17365548 on Build GRCh38
rs765104141 Chr.19: 17365618-17365618 on Build GRCh38
rs747771773 Chr.19: 17365380-17365380 on Build GRCh38
rs768876641 Chr.19: 17365541-17365541 on Build GRCh38
rs768079144 Chr.19: 17365375-17365375 on Build GRCh38
rs3810208 Chr.19: 17365442-17365442 on Build GRCh38
rs774009238 Chr.19: 17365378-17365378 on Build GRCh38
rs560705260 Chr.19: 17365356-17365356 on Build GRCh38
rs761178855 Chr.19: 17365260-17365260 on Build GRCh38
rs535383500 Chr.19: 17365349-17365349 on Build GRCh38
rs773739320 Chr.19: 17365350-17365350 on Build GRCh38
rs781598089 Chr.19: 17365583-17365583 on Build GRCh38
rs750109599 Chr.19: 17365656-17365656 on Build GRCh38
rs759212354 Chr.19: 17365611-17365611 on Build GRCh38
rs150260311 Chr.19: 17365602-17365602 on Build GRCh38
rs1042522184 Chr.19: 17365612-17365612 on Build GRCh38
rs749810877 Chr.19: 17365662-17365662 on Build GRCh38
rs113668445 Chr.19: 17365284-17365284 on Build GRCh38
rs114328379 Chr.19: 17365240-17365240 on Build GRCh38
rs773260384 Chr.19: 17365671-17365671 on Build GRCh38
rs763208613 Chr.19: 17365471-17365471 on Build GRCh38
rs773654858 Chr.19: 17365257-17365257 on Build GRCh38
rs752712824 Chr.19: 17365577-17365577 on Build GRCh38
rs971815592 Chr.19: 17365528-17365528 on Build GRCh38
rs367820138 Chr.19: 17365247-17365247 on Build GRCh38
rs755134547 Chr.19: 17365320-17365320 on Build GRCh38
rs779684076 Chr.19: 17365369-17365369 on Build GRCh38
rs532294385 Chr.19: 17365654-17365654 on Build GRCh38
rs370704284 Chr.19: 17365581-17365581 on Build GRCh38
rs199712527 Chr.19: 17365477-17365477 on Build GRCh38
rs776662915 Chr.19: 17365397-17365397 on Build GRCh38
rs201791359 Chr.19: 17365412-17365412 on Build GRCh38
rs139092467 Chr.19: 17365642-17365642 on Build GRCh38
rs769912880 Chr.19: 17365466-17365466 on Build GRCh38
rs772418879 Chr.19: 17365388-17365388 on Build GRCh38
rs748283687 Chr.19: 17365668-17365668 on Build GRCh38
rs372736269 Chr.19: 17365279-17365279 on Build GRCh38
rs200292631 Chr.19: 17365493-17365493 on Build GRCh38
rs754942023 Chr.19: 17365481-17365481 on Build GRCh38
rs778477668 Chr.19: 17365487-17365487 on Build GRCh38

View Full Product Details