Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 19227196-19227696 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCAN
Entrez Gene ID
1463
Gene Name
neurocan
Gene Aliases
CSPG3
UniGene
Hs.169047
Gene Chromosome Location
Chr.19: 19211973-19252252 on Build GRCh38
Transcripts
NM_004386.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4705801 c.1950C>A p.I650I Chr.19: 19227570-19227570 on Build GRCh38
1712079 c.1841C>T p.P614L Chr.19: 19227461-19227461 on Build GRCh38
6084147 c.1907T>A p.L636Q Chr.19: 19227527-19227527 on Build GRCh38
3668213 c.1842C>A p.P614P Chr.19: 19227462-19227462 on Build GRCh38
5389157 c.1696C>T p.L566L Chr.19: 19227316-19227316 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752868038 Chr.19: 19227550-19227550 on Build GRCh38
rs763424005 Chr.19: 19227307-19227307 on Build GRCh38
rs781629289 Chr.19: 19227348-19227348 on Build GRCh38
rs987179408 Chr.19: 19227224-19227224 on Build GRCh38
rs201043438 Chr.19: 19227602-19227602 on Build GRCh38
rs934997771 Chr.19: 19227362-19227362 on Build GRCh38
rs754950280 Chr.19: 19227455-19227455 on Build GRCh38
rs954489241 Chr.19: 19227217-19227222 on Build GRCh38
rs140529255 Chr.19: 19227306-19227306 on Build GRCh38
rs766766149 Chr.19: 19227445-19227445 on Build GRCh38
rs375952303 Chr.19: 19227473-19227473 on Build GRCh38
rs575262028 Chr.19: 19227534-19227534 on Build GRCh38
rs115916487 Chr.19: 19227653-19227653 on Build GRCh38
rs949253302 Chr.19: 19227236-19227236 on Build GRCh38
rs201315418 Chr.19: 19227506-19227506 on Build GRCh38
rs138659174 Chr.19: 19227502-19227502 on Build GRCh38
rs757496531 Chr.19: 19227346-19227346 on Build GRCh38
rs924769150 Chr.19: 19227437-19227437 on Build GRCh38
rs768397223 Chr.19: 19227611-19227611 on Build GRCh38
rs751470313 Chr.19: 19227302-19227302 on Build GRCh38
rs370556685 Chr.19: 19227508-19227508 on Build GRCh38
rs1026532318 Chr.19: 19227216-19227216 on Build GRCh38
rs373677142 Chr.19: 19227354-19227354 on Build GRCh38
rs757125847 Chr.19: 19227565-19227565 on Build GRCh38
rs746874998 Chr.19: 19227472-19227472 on Build GRCh38
rs111534277 Chr.19: 19227617-19227617 on Build GRCh38
rs373628957 Chr.19: 19227517-19227517 on Build GRCh38
rs764563622 Chr.19: 19227330-19227330 on Build GRCh38
rs983682959 Chr.19: 19227461-19227461 on Build GRCh38
rs145802020 Chr.19: 19227433-19227433 on Build GRCh38
rs745717935 Chr.19: 19227479-19227479 on Build GRCh38
rs369506098 Chr.19: 19227610-19227611 on Build GRCh38
rs750859324 Chr.19: 19227418-19227418 on Build GRCh38
rs774662675 Chr.19: 19227406-19227406 on Build GRCh38
rs751889617 Chr.19: 19227468-19227468 on Build GRCh38
rs187728357 Chr.19: 19227610-19227610 on Build GRCh38
rs777171049 Chr.19: 19227623-19227623 on Build GRCh38
rs371049970 Chr.19: 19227321-19227321 on Build GRCh38
rs368925637 Chr.19: 19227532-19227532 on Build GRCh38
rs761188774 Chr.19: 19227530-19227530 on Build GRCh38
rs759202432 Chr.19: 19227246-19227246 on Build GRCh38
rs752710402 Chr.19: 19227648-19227648 on Build GRCh38
rs762512331 Chr.19: 19227400-19227400 on Build GRCh38
rs951063842 Chr.19: 19227458-19227458 on Build GRCh38
rs759651783 Chr.19: 19227627-19227627 on Build GRCh38
rs531602650 Chr.19: 19227398-19227398 on Build GRCh38
rs747752512 Chr.19: 19227613-19227613 on Build GRCh38
rs771376825 Chr.19: 19227618-19227618 on Build GRCh38
rs144580072 Chr.19: 19227615-19227615 on Build GRCh38
rs202175922 Chr.19: 19227467-19227467 on Build GRCh38
rs779791812 Chr.19: 19227675-19227675 on Build GRCh38
rs776083940 Chr.19: 19227241-19227241 on Build GRCh38
rs977618896 Chr.19: 19227434-19227434 on Build GRCh38
rs768119647 Chr.19: 19227519-19227519 on Build GRCh38
rs765551496 Chr.19: 19227642-19227642 on Build GRCh38
rs370466971 Chr.19: 19227416-19227416 on Build GRCh38
rs576135458 Chr.19: 19227654-19227654 on Build GRCh38
rs201511416 Chr.19: 19227372-19227372 on Build GRCh38
rs762403187 Chr.19: 19227409-19227409 on Build GRCh38
rs755845467 Chr.19: 19227592-19227592 on Build GRCh38
rs756438427 Chr.19: 19227366-19227366 on Build GRCh38
rs780252481 Chr.19: 19227367-19227367 on Build GRCh38
rs775056326 Chr.19: 19227488-19227488 on Build GRCh38
rs990833100 Chr.19: 19227494-19227494 on Build GRCh38
rs149325641 Chr.19: 19227576-19227576 on Build GRCh38
rs373906684 Chr.19: 19227449-19227449 on Build GRCh38
rs137981080 Chr.19: 19227403-19227403 on Build GRCh38
rs756074366 Chr.19: 19227666-19227666 on Build GRCh38
rs868079643 Chr.19: 19227477-19227477 on Build GRCh38
rs369232036 Chr.19: 19227630-19227630 on Build GRCh38
rs769482303 Chr.19: 19227397-19227397 on Build GRCh38
rs772123011 Chr.19: 19227368-19227368 on Build GRCh38
rs199935130 Chr.19: 19227596-19227596 on Build GRCh38
rs754060519 Chr.19: 19227533-19227533 on Build GRCh38
rs200427306 Chr.19: 19227558-19227558 on Build GRCh38
rs750311153 Chr.19: 19227661-19227661 on Build GRCh38
rs770616255 Chr.19: 19227230-19227230 on Build GRCh38
rs372189026 Chr.19: 19227614-19227614 on Build GRCh38
rs561035532 Chr.19: 19227331-19227331 on Build GRCh38
rs201087476 Chr.19: 19227390-19227390 on Build GRCh38
rs764028917 Chr.19: 19227267-19227269 on Build GRCh38
rs748897933 Chr.19: 19227608-19227608 on Build GRCh38
rs764684464 Chr.19: 19227268-19227268 on Build GRCh38
rs942378006 Chr.19: 19227620-19227620 on Build GRCh38
rs765193656 Chr.19: 19227548-19227548 on Build GRCh38
rs150327139 Chr.19: 19227382-19227382 on Build GRCh38
rs773700316 Chr.19: 19227529-19227529 on Build GRCh38
rs769428851 Chr.19: 19227487-19227487 on Build GRCh38
rs140117865 Chr.19: 19227612-19227612 on Build GRCh38
rs745487557 Chr.19: 19227391-19227391 on Build GRCh38
rs745806145 Chr.19: 19227581-19227581 on Build GRCh38
rs765896733 Chr.19: 19227358-19227358 on Build GRCh38
rs764171896 Chr.19: 19227656-19227656 on Build GRCh38
rs916377393 Chr.19: 19227218-19227218 on Build GRCh38

View Full Product Details