Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 21056507-21057055 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF430
Entrez Gene ID
80264
Gene Name
zinc finger protein 430
Gene Aliases
-
UniGene
Hs.729202
Gene Chromosome Location
Chr.19: 21020620-21060046 on Build GRCh38
Transcripts
NM_001172671.1 NM_025189.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4600642 c.532C>A p.Q178K Chr.19: 21056840-21056840 on Build GRCh38
3531320 c.725T>A p.F242Y Chr.19: 21057033-21057033 on Build GRCh38
6433606 c.508A>T p.T170S Chr.19: 21056816-21056816 on Build GRCh38
7176592 c.593G>A p.R198K Chr.19: 21056901-21056901 on Build GRCh38
6398558 c.440C>A p.T147K Chr.19: 21056748-21056748 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369825441 Chr.19: 21056670-21056670 on Build GRCh38
rs773279046 Chr.19: 21056796-21056796 on Build GRCh38
rs771102557 Chr.19: 21056822-21056822 on Build GRCh38
rs370609116 Chr.19: 21056842-21056842 on Build GRCh38
rs144891389 Chr.19: 21056712-21056712 on Build GRCh38
rs765657821 Chr.19: 21056837-21056837 on Build GRCh38
rs150504986 Chr.19: 21056776-21056776 on Build GRCh38
rs759005737 Chr.19: 21056713-21056713 on Build GRCh38
rs749091077 Chr.19: 21056921-21056921 on Build GRCh38
rs766517895 Chr.19: 21056685-21056685 on Build GRCh38
rs773702221 Chr.19: 21056929-21056930 on Build GRCh38
rs751035178 Chr.19: 21056727-21056727 on Build GRCh38
rs989302686 Chr.19: 21056569-21056569 on Build GRCh38
rs376456283 Chr.19: 21056756-21056756 on Build GRCh38
rs572758723 Chr.19: 21056934-21056934 on Build GRCh38
rs755781546 Chr.19: 21056903-21056903 on Build GRCh38
rs142242499 Chr.19: 21056898-21056898 on Build GRCh38
rs749179163 Chr.19: 21056781-21056781 on Build GRCh38
rs755478286 Chr.19: 21056598-21056598 on Build GRCh38
rs187080608 Chr.19: 21056554-21056554 on Build GRCh38
rs761227954 Chr.19: 21057026-21057027 on Build GRCh38
rs759968025 Chr.19: 21056701-21056701 on Build GRCh38
rs764567373 Chr.19: 21056969-21056969 on Build GRCh38
rs772095140 Chr.19: 21056680-21056680 on Build GRCh38
rs746622822 Chr.19: 21056635-21056635 on Build GRCh38
rs550182423 Chr.19: 21056543-21056543 on Build GRCh38
rs561516548 Chr.19: 21056974-21056974 on Build GRCh38
rs542350615 Chr.19: 21056943-21056943 on Build GRCh38
rs951104335 Chr.19: 21056584-21056584 on Build GRCh38
rs779852785 Chr.19: 21056780-21056780 on Build GRCh38
rs779560668 Chr.19: 21056601-21056601 on Build GRCh38
rs199744484 Chr.19: 21056675-21056675 on Build GRCh38
rs759878122 Chr.19: 21056832-21056832 on Build GRCh38
rs746484913 Chr.19: 21056794-21056794 on Build GRCh38
rs185470665 Chr.19: 21056928-21056928 on Build GRCh38
rs756326834 Chr.19: 21056583-21056583 on Build GRCh38
rs766720832 Chr.19: 21056588-21056588 on Build GRCh38
rs776882876 Chr.19: 21056946-21056946 on Build GRCh38
rs750046481 Chr.19: 21056682-21056682 on Build GRCh38
rs886549332 Chr.19: 21056693-21056693 on Build GRCh38
rs776972801 Chr.19: 21056823-21056823 on Build GRCh38
rs78534959 Chr.19: 21056612-21056612 on Build GRCh38
rs756518284 Chr.19: 21056556-21056556 on Build GRCh38
rs754170570 Chr.19: 21056592-21056592 on Build GRCh38
rs760669365 Chr.19: 21056814-21056814 on Build GRCh38
rs765747796 Chr.19: 21056703-21056703 on Build GRCh38
rs936403278 Chr.19: 21056557-21056557 on Build GRCh38
rs772178982 Chr.19: 21056793-21056795 on Build GRCh38
rs904866284 Chr.19: 21056550-21056550 on Build GRCh38
rs139084188 Chr.19: 21056681-21056681 on Build GRCh38
rs934744150 Chr.19: 21056724-21056724 on Build GRCh38
rs778706138 Chr.19: 21056933-21056933 on Build GRCh38
rs754580454 Chr.19: 21056868-21056868 on Build GRCh38
rs149888883 Chr.19: 21056688-21056688 on Build GRCh38
rs753124776 Chr.19: 21056839-21056839 on Build GRCh38
rs775920873 Chr.19: 21056952-21056952 on Build GRCh38
rs201566618 Chr.19: 21057002-21057002 on Build GRCh38
rs748723570 Chr.19: 21056604-21056604 on Build GRCh38
rs754158104 Chr.19: 21056696-21056696 on Build GRCh38
rs763212213 Chr.19: 21056960-21056960 on Build GRCh38
rs762411446 Chr.19: 21057024-21057024 on Build GRCh38
rs774687079 Chr.19: 21057004-21057004 on Build GRCh38
rs777279999 Chr.19: 21056723-21056723 on Build GRCh38
rs916408671 Chr.19: 21056578-21056578 on Build GRCh38
rs750623735 Chr.19: 21056581-21056581 on Build GRCh38
rs200822899 Chr.19: 21056852-21056852 on Build GRCh38
rs7256437 Chr.19: 21056918-21056918 on Build GRCh38
rs370882800 Chr.19: 21056936-21056936 on Build GRCh38
rs144326071 Chr.19: 21056672-21056672 on Build GRCh38
rs148335981 Chr.19: 21056947-21056947 on Build GRCh38
rs757960660 Chr.19: 21056609-21056609 on Build GRCh38
rs373575225 Chr.19: 21056738-21056738 on Build GRCh38
rs776419488 Chr.19: 21056666-21056666 on Build GRCh38
rs116573353 Chr.19: 21056854-21056854 on Build GRCh38
rs535240998 Chr.19: 21056689-21056689 on Build GRCh38
rs777188984 Chr.19: 21056611-21056611 on Build GRCh38
rs748203333 Chr.19: 21056662-21056663 on Build GRCh38
rs1006299493 Chr.19: 21056699-21056699 on Build GRCh38
rs141116955 Chr.19: 21056762-21056762 on Build GRCh38
rs965906544 Chr.19: 21056615-21056615 on Build GRCh38
rs375938971 Chr.19: 21056647-21056647 on Build GRCh38

View Full Product Details