Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 23373655-23374162 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF91
Entrez Gene ID
7644
Gene Name
zinc finger protein 91
Gene Aliases
HPF7, HTF10
UniGene
Hs.531632
Gene Chromosome Location
Chr.19: 23305004-23395560 on Build GRCh38
Transcripts
NM_001300951.1 NM_003430.3 XM_006722880.3 XM_011528265.2 XM_011528266.2 XM_017027235.1 XM_017027236.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1392231 c.176C>A p.P59Q Chr.19: 23373819-23373819 on Build GRCh38
288088 c.192T>C p.Y64Y Chr.19: 23373803-23373803 on Build GRCh38
6263649 c.166C>G p.L56V Chr.19: 23373829-23373829 on Build GRCh38
4858548 c.203G>A p.G68E Chr.19: 23373792-23373792 on Build GRCh38
1589976 c.198G>A p.E66E Chr.19: 23373797-23373797 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1034469947 Chr.19: 23374135-23374135 on Build GRCh38
rs377228317 Chr.19: 23373802-23373802 on Build GRCh38
rs537512418 Chr.19: 23374028-23374028 on Build GRCh38
rs377459278 Chr.19: 23373877-23373877 on Build GRCh38
rs866813184 Chr.19: 23374092-23374092 on Build GRCh38
rs772071180 Chr.19: 23373843-23373843 on Build GRCh38
rs17554633 Chr.19: 23374050-23374050 on Build GRCh38
rs373466656 Chr.19: 23373820-23373820 on Build GRCh38
rs570539879 Chr.19: 23373933-23373933 on Build GRCh38
rs374732824 Chr.19: 23373693-23373693 on Build GRCh38
rs1049572487 Chr.19: 23373821-23373821 on Build GRCh38
rs761803358 Chr.19: 23373731-23373731 on Build GRCh38
rs200627969 Chr.19: 23373759-23373759 on Build GRCh38
rs369674549 Chr.19: 23373757-23373757 on Build GRCh38
rs770882565 Chr.19: 23373716-23373716 on Build GRCh38
rs904753379 Chr.19: 23374079-23374079 on Build GRCh38
rs757869359 Chr.19: 23373777-23373777 on Build GRCh38
rs372907267 Chr.19: 23373769-23373769 on Build GRCh38
rs377387685 Chr.19: 23373829-23373829 on Build GRCh38
rs34740519 Chr.19: 23373803-23373803 on Build GRCh38
rs748206589 Chr.19: 23373838-23373838 on Build GRCh38
rs1023556639 Chr.19: 23373915-23373915 on Build GRCh38
rs973448340 Chr.19: 23373758-23373758 on Build GRCh38
rs750335705 Chr.19: 23374124-23374124 on Build GRCh38
rs1001778607 Chr.19: 23374103-23374103 on Build GRCh38
rs763959145 Chr.19: 23373717-23373717 on Build GRCh38
rs776916774 Chr.19: 23373887-23373887 on Build GRCh38
rs375618205 Chr.19: 23374090-23374090 on Build GRCh38
rs777307696 Chr.19: 23373698-23373698 on Build GRCh38
rs557571273 Chr.19: 23373684-23373684 on Build GRCh38
rs773389392 Chr.19: 23373874-23373874 on Build GRCh38
rs891560512 Chr.19: 23373994-23373994 on Build GRCh38
rs148956643 Chr.19: 23373748-23373748 on Build GRCh38
rs771283571 Chr.19: 23373882-23373882 on Build GRCh38
rs752197684 Chr.19: 23373774-23373774 on Build GRCh38
rs960607381 Chr.19: 23373871-23373871 on Build GRCh38
rs773440636 Chr.19: 23373744-23373744 on Build GRCh38
rs1040777372 Chr.19: 23373925-23373925 on Build GRCh38
rs369964129 Chr.19: 23373705-23373705 on Build GRCh38
rs760984987 Chr.19: 23373747-23373747 on Build GRCh38
rs200994728 Chr.19: 23373737-23373737 on Build GRCh38
rs368985302 Chr.19: 23374093-23374093 on Build GRCh38
rs751299264 Chr.19: 23373796-23373796 on Build GRCh38
rs371406488 Chr.19: 23373691-23373691 on Build GRCh38
rs373305708 Chr.19: 23373720-23373720 on Build GRCh38
rs980662965 Chr.19: 23373681-23373681 on Build GRCh38
rs376532436 Chr.19: 23373855-23373855 on Build GRCh38
rs201748917 Chr.19: 23373780-23373780 on Build GRCh38
rs745793518 Chr.19: 23373825-23373825 on Build GRCh38
rs747276961 Chr.19: 23373876-23373876 on Build GRCh38
rs777442535 Chr.19: 23373778-23373778 on Build GRCh38
rs1013836893 Chr.19: 23373897-23373897 on Build GRCh38
rs774557781 Chr.19: 23373728-23373728 on Build GRCh38
rs745848571 Chr.19: 23373718-23373718 on Build GRCh38
rs969262490 Chr.19: 23373920-23373920 on Build GRCh38

View Full Product Details