Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 31278627-31279127 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSHZ3
Entrez Gene ID
57616
Gene Name
teashirt zinc finger homeobox 3
Gene Aliases
TSH3, ZNF537
UniGene
Hs.278436
Gene Chromosome Location
Chr.19: 31274945-31349284 on Build GRCh38
Transcripts

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
994523 c.142G>A p.V48M Chr.19: 31279102-31279102 on Build GRCh38
565580 c.1107G>A p.Q369Q Chr.19: 31278686-31278686 on Build GRCh38
5056500 c.718C>T p.R240C Chr.19: 31279075-31279075 on Build GRCh38
5582872 c.203C>T p.P68L Chr.19: 31279041-31279041 on Build GRCh38
2750645 c.166T>A p.Y56N Chr.19: 31279078-31279078 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749597184 Chr.19: 31279053-31279053 on Build GRCh38
rs267605409 Chr.19: 31278997-31278997 on Build GRCh38
rs918714863 Chr.19: 31278994-31278994 on Build GRCh38
rs917796527 Chr.19: 31278948-31278948 on Build GRCh38
rs138541165 Chr.19: 31279091-31279091 on Build GRCh38
rs772707105 Chr.19: 31279070-31279070 on Build GRCh38
rs777524623 Chr.19: 31278694-31278694 on Build GRCh38
rs770806071 Chr.19: 31278701-31278701 on Build GRCh38
rs773916270 Chr.19: 31278977-31278977 on Build GRCh38
rs776665273 Chr.19: 31278947-31278947 on Build GRCh38
rs200286730 Chr.19: 31278711-31278711 on Build GRCh38
rs923574447 Chr.19: 31279072-31279072 on Build GRCh38
rs746061219 Chr.19: 31278951-31278951 on Build GRCh38
rs754151995 Chr.19: 31278751-31278751 on Build GRCh38
rs752282606 Chr.19: 31278780-31278780 on Build GRCh38
rs976295486 Chr.19: 31279075-31279075 on Build GRCh38
rs750249835 Chr.19: 31278865-31278865 on Build GRCh38
rs778652998 Chr.19: 31279034-31279034 on Build GRCh38
rs766061784 Chr.19: 31278715-31278716 on Build GRCh38
rs776184849 Chr.19: 31278853-31278853 on Build GRCh38
rs771485583 Chr.19: 31278797-31278797 on Build GRCh38
rs143883315 Chr.19: 31279054-31279054 on Build GRCh38
rs777071664 Chr.19: 31278849-31278849 on Build GRCh38
rs552223166 Chr.19: 31278653-31278653 on Build GRCh38
rs774427176 Chr.19: 31278719-31278719 on Build GRCh38
rs755412223 Chr.19: 31278656-31278656 on Build GRCh38
rs761814857 Chr.19: 31278721-31278721 on Build GRCh38
rs146913156 Chr.19: 31278828-31278828 on Build GRCh38
rs137987422 Chr.19: 31278959-31278959 on Build GRCh38
rs200791129 Chr.19: 31278770-31278770 on Build GRCh38
rs761188265 Chr.19: 31278981-31278981 on Build GRCh38
rs535604105 Chr.19: 31278982-31278982 on Build GRCh38
rs765055013 Chr.19: 31279090-31279090 on Build GRCh38
rs747007733 Chr.19: 31278908-31278908 on Build GRCh38
rs753720547 Chr.19: 31278717-31278718 on Build GRCh38
rs763611766 Chr.19: 31278971-31278971 on Build GRCh38
rs201015382 Chr.19: 31279035-31279035 on Build GRCh38
rs748250164 Chr.19: 31278832-31278832 on Build GRCh38
rs557330136 Chr.19: 31279086-31279086 on Build GRCh38
rs759916924 Chr.19: 31278851-31278851 on Build GRCh38
rs769727817 Chr.19: 31278831-31278831 on Build GRCh38
rs770126054 Chr.19: 31279047-31279047 on Build GRCh38
rs752245787 Chr.19: 31278668-31278668 on Build GRCh38
rs772311820 Chr.19: 31278732-31278732 on Build GRCh38
rs780817273 Chr.19: 31278705-31278705 on Build GRCh38
rs75499863 Chr.19: 31278843-31278843 on Build GRCh38
rs377716978 Chr.19: 31278837-31278837 on Build GRCh38
rs887864001 Chr.19: 31278861-31278861 on Build GRCh38
rs766810539 Chr.19: 31278747-31278747 on Build GRCh38
rs61744715 Chr.19: 31278963-31278963 on Build GRCh38
rs760381593 Chr.19: 31279073-31279073 on Build GRCh38
rs200738522 Chr.19: 31278856-31278856 on Build GRCh38
rs773136339 Chr.19: 31278842-31278842 on Build GRCh38
rs754908096 Chr.19: 31278885-31278885 on Build GRCh38
rs994020303 Chr.19: 31278936-31278936 on Build GRCh38
rs528782093 Chr.19: 31278852-31278852 on Build GRCh38
rs781097660 Chr.19: 31278817-31278817 on Build GRCh38
rs565926372 Chr.19: 31279038-31279038 on Build GRCh38
rs531021804 Chr.19: 31278864-31278864 on Build GRCh38
rs754419344 Chr.19: 31278651-31278651 on Build GRCh38
rs753638329 Chr.19: 31279085-31279085 on Build GRCh38
rs374355143 Chr.19: 31278809-31278809 on Build GRCh38
rs139336851 Chr.19: 31278854-31278854 on Build GRCh38
rs766230775 Chr.19: 31279004-31279004 on Build GRCh38
rs771048734 Chr.19: 31278848-31278848 on Build GRCh38
rs868368486 Chr.19: 31278672-31278672 on Build GRCh38
rs755093450 Chr.19: 31278721-31278722 on Build GRCh38
rs188441334 Chr.19: 31278868-31278868 on Build GRCh38
rs373776019 Chr.19: 31278869-31278869 on Build GRCh38
rs142329923 Chr.19: 31278800-31278800 on Build GRCh38
rs755734150 Chr.19: 31279002-31279002 on Build GRCh38
rs760787125 Chr.19: 31278745-31278745 on Build GRCh38
rs142807470 Chr.19: 31278710-31278710 on Build GRCh38
rs149474575 Chr.19: 31278974-31278974 on Build GRCh38
rs756854055 Chr.19: 31278811-31278811 on Build GRCh38
rs746155760 Chr.19: 31279041-31279041 on Build GRCh38
rs28609894 Chr.19: 31279103-31279103 on Build GRCh38
rs979100128 Chr.19: 31278859-31278859 on Build GRCh38
rs762360444 Chr.19: 31279062-31279062 on Build GRCh38
rs199546310 Chr.19: 31278819-31278819 on Build GRCh38
rs754959706 Chr.19: 31279020-31279020 on Build GRCh38
rs12461253 Chr.19: 31278857-31278857 on Build GRCh38
rs771523387 Chr.19: 31279069-31279069 on Build GRCh38
rs775613015 Chr.19: 31278983-31278983 on Build GRCh38
rs760719932 Chr.19: 31279050-31279050 on Build GRCh38
rs200052169 Chr.19: 31279015-31279015 on Build GRCh38
rs756947039 Chr.19: 31278860-31278860 on Build GRCh38
rs768854873 Chr.19: 31279055-31279055 on Build GRCh38
rs772491128 Chr.19: 31278893-31278893 on Build GRCh38
rs555294304 Chr.19: 31279036-31279036 on Build GRCh38
rs375634411 Chr.19: 31279040-31279040 on Build GRCh38
rs757928110 Chr.19: 31278790-31278790 on Build GRCh38
rs1037129129 Chr.19: 31278716-31278716 on Build GRCh38
rs147659074 Chr.19: 31278774-31278774 on Build GRCh38
rs61747216 Chr.19: 31278890-31278890 on Build GRCh38
rs917738425 Chr.19: 31278704-31278704 on Build GRCh38
rs760182546 Chr.19: 31278760-31278760 on Build GRCh38
rs773436087 Chr.19: 31278739-31278739 on Build GRCh38
rs746799556 Chr.19: 31278695-31278695 on Build GRCh38
rs376393548 Chr.19: 31278662-31278662 on Build GRCh38
rs771192598 Chr.19: 31278919-31278919 on Build GRCh38
rs770320228 Chr.19: 31278956-31278956 on Build GRCh38
rs186519057 Chr.19: 31279074-31279074 on Build GRCh38
rs749265079 Chr.19: 31278872-31278872 on Build GRCh38
rs1040664426 Chr.19: 31278847-31278847 on Build GRCh38
rs369970992 Chr.19: 31278801-31278801 on Build GRCh38

View Full Product Details