Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 46303739-46304227 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIF3A
Entrez Gene ID
64344
Gene Name
hypoxia inducible factor 3 alpha subunit
Gene Aliases
HIF-3A, HIF3-alpha-1, IPAS, MOP7, PASD7, bHLHe17
UniGene
Hs.420830
Gene Chromosome Location
Chr.19: 46297046-46343433 on Build GRCh38
Transcripts
NM_022462.4 NM_152794.3 NM_152795.3 NM_152796.2 XM_005259152.4 XM_005259153.3 XM_005259155.4 XM_005259156.4 XM_017027132.1 XM_017027133.1 XM_017027134.1 XM_017027135.1 XM_017027136.1 XM_017027137.1 XM_017027138.1 XM_017027139.1 XM_017027140.1 XM_017027141.1 XM_017027142.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7200690 c.165C>T p.C55C Chr.19: 46304043-46304043 on Build GRCh38
6423569 c.144C>A p.H48Q Chr.19: 46304021-46304021 on Build GRCh38
6423570 c.143C>A p.T48N Chr.19: 46304021-46304021 on Build GRCh38
4816352 c.207G>A p.P69P Chr.19: 46304085-46304085 on Build GRCh38
7200688 c.172C>T p.R58C Chr.19: 46304043-46304043 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756165739 Chr.19: 46304040-46304040 on Build GRCh38
rs768347653 Chr.19: 46303949-46303949 on Build GRCh38
rs375846168 Chr.19: 46304018-46304018 on Build GRCh38
rs778626472 Chr.19: 46303898-46303898 on Build GRCh38
rs763398201 Chr.19: 46303966-46303966 on Build GRCh38
rs199779703 Chr.19: 46304137-46304138 on Build GRCh38
rs569086569 Chr.19: 46304004-46304004 on Build GRCh38
rs769137791 Chr.19: 46304043-46304043 on Build GRCh38
rs756194874 Chr.19: 46304103-46304103 on Build GRCh38
rs771902082 Chr.19: 46304007-46304007 on Build GRCh38
rs781310174 Chr.19: 46303983-46303983 on Build GRCh38
rs764011338 Chr.19: 46304113-46304113 on Build GRCh38
rs561573536 Chr.19: 46303913-46303913 on Build GRCh38
rs540143517 Chr.19: 46303832-46303832 on Build GRCh38
rs1038125441 Chr.19: 46303826-46303826 on Build GRCh38
rs760175137 Chr.19: 46303945-46303945 on Build GRCh38
rs747745830 Chr.19: 46304140-46304140 on Build GRCh38
rs1057174237 Chr.19: 46304170-46304170 on Build GRCh38
rs761440699 Chr.19: 46304041-46304041 on Build GRCh38
rs976535588 Chr.19: 46304014-46304014 on Build GRCh38
rs113691430 Chr.19: 46303777-46303777 on Build GRCh38
rs776320049 Chr.19: 46304036-46304036 on Build GRCh38
rs779946029 Chr.19: 46304130-46304130 on Build GRCh38
rs1035602964 Chr.19: 46304179-46304179 on Build GRCh38
rs927337057 Chr.19: 46304146-46304146 on Build GRCh38
rs757833861 Chr.19: 46303843-46303843 on Build GRCh38
rs929451703 Chr.19: 46303842-46303842 on Build GRCh38
rs766595847 Chr.19: 46303971-46303971 on Build GRCh38
rs751859823 Chr.19: 46304062-46304062 on Build GRCh38
rs750776790 Chr.19: 46303847-46303847 on Build GRCh38
rs532948502 Chr.19: 46303808-46303808 on Build GRCh38
rs760251315 Chr.19: 46303831-46303831 on Build GRCh38
rs751812041 Chr.19: 46303979-46303979 on Build GRCh38
rs919326700 Chr.19: 46304016-46304016 on Build GRCh38
rs758253537 Chr.19: 46304120-46304120 on Build GRCh38
rs551056583 Chr.19: 46304208-46304208 on Build GRCh38
rs759807487 Chr.19: 46304082-46304082 on Build GRCh38
rs1010299132 Chr.19: 46304185-46304185 on Build GRCh38
rs746769253 Chr.19: 46304131-46304131 on Build GRCh38
rs992153229 Chr.19: 46304188-46304188 on Build GRCh38
rs758588946 Chr.19: 46303860-46303860 on Build GRCh38
rs367570195 Chr.19: 46304019-46304019 on Build GRCh38
rs752756082 Chr.19: 46303984-46303984 on Build GRCh38
rs563901616 Chr.19: 46303764-46303764 on Build GRCh38
rs745730884 Chr.19: 46304005-46304005 on Build GRCh38
rs1042283799 Chr.19: 46304198-46304198 on Build GRCh38
rs1039286291 Chr.19: 46304073-46304073 on Build GRCh38
rs751685617 Chr.19: 46303862-46303862 on Build GRCh38
rs781184848 Chr.19: 46303870-46303870 on Build GRCh38
rs770846110 Chr.19: 46303889-46303889 on Build GRCh38
rs926576266 Chr.19: 46303987-46303987 on Build GRCh38
rs765754233 Chr.19: 46303846-46303846 on Build GRCh38
rs116420807 Chr.19: 46304045-46304045 on Build GRCh38
rs941277565 Chr.19: 46304117-46304117 on Build GRCh38
rs751361030 Chr.19: 46303893-46303894 on Build GRCh38
rs909693229 Chr.19: 46304087-46304087 on Build GRCh38
rs201468772 Chr.19: 46303901-46303901 on Build GRCh38
rs904658064 Chr.19: 46304204-46304204 on Build GRCh38
rs897942541 Chr.19: 46303836-46303836 on Build GRCh38
rs960735275 Chr.19: 46304001-46304001 on Build GRCh38
rs749293528 Chr.19: 46303881-46303881 on Build GRCh38
rs142517237 Chr.19: 46303928-46303928 on Build GRCh38
rs765807165 Chr.19: 46303957-46303957 on Build GRCh38
rs746716033 Chr.19: 46304022-46304022 on Build GRCh38
rs867611831 Chr.19: 46304065-46304065 on Build GRCh38
rs530997581 Chr.19: 46304177-46304177 on Build GRCh38
rs1011297371 Chr.19: 46304137-46304137 on Build GRCh38
rs937325389 Chr.19: 46304149-46304149 on Build GRCh38
rs895919114 Chr.19: 46304178-46304178 on Build GRCh38
rs754397971 Chr.19: 46303837-46303837 on Build GRCh38
rs755066168 Chr.19: 46303863-46303863 on Build GRCh38
rs1056553130 Chr.19: 46303809-46303809 on Build GRCh38
rs771711197 Chr.19: 46303910-46303910 on Build GRCh38
rs371806876 Chr.19: 46304048-46304048 on Build GRCh38
rs768399021 Chr.19: 46304033-46304033 on Build GRCh38
rs754780099 Chr.19: 46304133-46304133 on Build GRCh38
rs1031413091 Chr.19: 46303914-46303914 on Build GRCh38
rs757228711 Chr.19: 46303991-46303991 on Build GRCh38
rs767699026 Chr.19: 46304092-46304092 on Build GRCh38
rs896012437 Chr.19: 46303927-46303927 on Build GRCh38
rs943563294 Chr.19: 46304064-46304064 on Build GRCh38
rs926654753 Chr.19: 46303765-46303765 on Build GRCh38
rs115548009 Chr.19: 46303982-46303982 on Build GRCh38
rs752864358 Chr.19: 46304094-46304094 on Build GRCh38
rs766837195 Chr.19: 46304052-46304052 on Build GRCh38
rs1035734206 Chr.19: 46303922-46303922 on Build GRCh38
rs750856621 Chr.19: 46303958-46303958 on Build GRCh38
rs1039076184 Chr.19: 46303867-46303867 on Build GRCh38
rs762458259 Chr.19: 46303953-46303953 on Build GRCh38
rs781015034 Chr.19: 46304135-46304135 on Build GRCh38

View Full Product Details