Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 46608109-46608603 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALM3
Entrez Gene ID
808
Gene Name
calmodulin 3
Gene Aliases
HEL-S-72, PHKD, PHKD3
UniGene
Hs.515487
Gene Chromosome Location
Chr.19: 46601255-46610782 on Build GRCh38
Transcripts
NM_005184.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7348508 c.259C>T p.R87* Chr.19: 46608562-46608562 on Build GRCh38
3769869 c.69T>G p.D23E Chr.19: 46608231-46608231 on Build GRCh38
998445 c.108G>A p.V36V Chr.19: 46608270-46608270 on Build GRCh38
4079587 c.240A>C p.T80T Chr.19: 46608543-46608543 on Build GRCh38
6573596 c.260G>A p.R87Q Chr.19: 46608563-46608563 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs966713319 Chr.19: 46608402-46608402 on Build GRCh38
rs369539598 Chr.19: 46608184-46608184 on Build GRCh38
rs753670904 Chr.19: 46608352-46608352 on Build GRCh38
rs536630307 Chr.19: 46608411-46608411 on Build GRCh38
rs761456118 Chr.19: 46608356-46608356 on Build GRCh38
rs1002766364 Chr.19: 46608182-46608182 on Build GRCh38
rs774567046 Chr.19: 46608439-46608439 on Build GRCh38
rs377206004 Chr.19: 46608173-46608173 on Build GRCh38
rs760657005 Chr.19: 46608309-46608309 on Build GRCh38
rs180722019 Chr.19: 46608358-46608358 on Build GRCh38
rs764944151 Chr.19: 46608357-46608357 on Build GRCh38
rs749423113 Chr.19: 46608435-46608435 on Build GRCh38
rs760752804 Chr.19: 46608443-46608443 on Build GRCh38
rs376024469 Chr.19: 46608445-46608445 on Build GRCh38
rs771082049 Chr.19: 46608437-46608437 on Build GRCh38
rs748438358 Chr.19: 46608187-46608187 on Build GRCh38
rs577585466 Chr.19: 46608466-46608466 on Build GRCh38
rs755348000 Chr.19: 46608552-46608552 on Build GRCh38
rs762484332 Chr.19: 46608487-46608487 on Build GRCh38
rs764047686 Chr.19: 46608327-46608327 on Build GRCh38
rs747463054 Chr.19: 46608181-46608181 on Build GRCh38
rs757777400 Chr.19: 46608178-46608178 on Build GRCh38
rs756526606 Chr.19: 46608569-46608569 on Build GRCh38
rs770046799 Chr.19: 46608191-46608191 on Build GRCh38
rs375106354 Chr.19: 46608321-46608321 on Build GRCh38
rs375684773 Chr.19: 46608259-46608259 on Build GRCh38
rs887638849 Chr.19: 46608144-46608144 on Build GRCh38
rs781598565 Chr.19: 46608386-46608386 on Build GRCh38
rs773251048 Chr.19: 46608243-46608243 on Build GRCh38
rs944414245 Chr.19: 46608427-46608427 on Build GRCh38
rs34089805 Chr.19: 46608504-46608504 on Build GRCh38
rs763100441 Chr.19: 46608244-46608244 on Build GRCh38
rs752104056 Chr.19: 46608374-46608374 on Build GRCh38
rs761694804 Chr.19: 46608464-46608464 on Build GRCh38
rs370430097 Chr.19: 46608183-46608183 on Build GRCh38
rs3729761 Chr.19: 46608486-46608486 on Build GRCh38
rs749960299 Chr.19: 46608170-46608170 on Build GRCh38
rs373292581 Chr.19: 46608146-46608146 on Build GRCh38
rs753584176 Chr.19: 46608154-46608154 on Build GRCh38
rs559783307 Chr.19: 46608576-46608576 on Build GRCh38
rs1003514568 Chr.19: 46608190-46608190 on Build GRCh38
rs3729760 Chr.19: 46608406-46608406 on Build GRCh38
rs753593596 Chr.19: 46608465-46608466 on Build GRCh38
rs775591771 Chr.19: 46608286-46608286 on Build GRCh38
rs756928364 Chr.19: 46608155-46608155 on Build GRCh38
rs368593188 Chr.19: 46608428-46608428 on Build GRCh38
rs186405375 Chr.19: 46608359-46608359 on Build GRCh38
rs755475396 Chr.19: 46608382-46608382 on Build GRCh38
rs115265989 Chr.19: 46608570-46608570 on Build GRCh38
rs749917251 Chr.19: 46608310-46608311 on Build GRCh38
rs764890178 Chr.19: 46608469-46608469 on Build GRCh38
rs3729759 Chr.19: 46608388-46608388 on Build GRCh38
rs550294796 Chr.19: 46608158-46608158 on Build GRCh38
rs147734536 Chr.19: 46608540-46608540 on Build GRCh38
rs1012699471 Chr.19: 46608168-46608168 on Build GRCh38
rs779521135 Chr.19: 46608364-46608364 on Build GRCh38
rs559310165 Chr.19: 46608372-46608374 on Build GRCh38
rs1044180500 Chr.19: 46608562-46608562 on Build GRCh38
rs780566686 Chr.19: 46608179-46608179 on Build GRCh38
rs145596510 Chr.19: 46608513-46608513 on Build GRCh38
rs753104718 Chr.19: 46608555-46608555 on Build GRCh38
rs544400553 Chr.19: 46608447-46608447 on Build GRCh38
rs887681678 Chr.19: 46608537-46608537 on Build GRCh38

View Full Product Details