Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49464311-49464814 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDH16A1
Entrez Gene ID
126133
Gene Name
aldehyde dehydrogenase 16 family member A1
Gene Aliases
-
UniGene
Hs.719998
Gene Chromosome Location
Chr.19: 49453216-49471050 on Build GRCh38
Transcripts
NM_001145396.1 NM_153329.3 XM_011526441.1 XM_011526442.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
272327 c.1365C>T p.H455H Chr.19: 49464450-49464450 on Build GRCh38
3893042 c.1531C>T p.P511S Chr.19: 49464725-49464725 on Build GRCh38
7201477 c.1567A>C p.S523R Chr.19: 49464761-49464761 on Build GRCh38
1244807 c.1335C>G p.L445L Chr.19: 49464420-49464420 on Build GRCh38
2752599 c.1437C>T p.D479D Chr.19: 49464522-49464522 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201084975 Chr.19: 49464527-49464527 on Build GRCh38
rs375627359 Chr.19: 49464377-49464377 on Build GRCh38
rs761695664 Chr.19: 49464469-49464469 on Build GRCh38
rs780070423 Chr.19: 49464458-49464458 on Build GRCh38
rs775229384 Chr.19: 49464727-49464727 on Build GRCh38
rs201777977 Chr.19: 49464594-49464594 on Build GRCh38
rs756948267 Chr.19: 49464444-49464444 on Build GRCh38
rs917255833 Chr.19: 49464338-49464338 on Build GRCh38
rs750231212 Chr.19: 49464753-49464753 on Build GRCh38
rs1044342828 Chr.19: 49464538-49464538 on Build GRCh38
rs767457200 Chr.19: 49464484-49464484 on Build GRCh38
rs745343121 Chr.19: 49464390-49464390 on Build GRCh38
rs371054096 Chr.19: 49464481-49464481 on Build GRCh38
rs995101582 Chr.19: 49464704-49464704 on Build GRCh38
rs990125979 Chr.19: 49464771-49464771 on Build GRCh38
rs200335556 Chr.19: 49464560-49464560 on Build GRCh38
rs10421522 Chr.19: 49464739-49464739 on Build GRCh38
rs371313328 Chr.19: 49464365-49464365 on Build GRCh38
rs764646307 Chr.19: 49464541-49464541 on Build GRCh38
rs549966663 Chr.19: 49464554-49464554 on Build GRCh38
rs34601909 Chr.19: 49464551-49464551 on Build GRCh38
rs1000386645 Chr.19: 49464345-49464345 on Build GRCh38
rs1027691405 Chr.19: 49464662-49464662 on Build GRCh38
rs774379640 Chr.19: 49464480-49464480 on Build GRCh38
rs769845538 Chr.19: 49464701-49464701 on Build GRCh38
rs368976678 Chr.19: 49464599-49464599 on Build GRCh38
rs750214804 Chr.19: 49464341-49464341 on Build GRCh38
rs771330332 Chr.19: 49464660-49464660 on Build GRCh38
rs777763681 Chr.19: 49464614-49464614 on Build GRCh38
rs150696383 Chr.19: 49464671-49464671 on Build GRCh38
rs776261322 Chr.19: 49464744-49464744 on Build GRCh38
rs370026616 Chr.19: 49464579-49464579 on Build GRCh38
rs199547632 Chr.19: 49464585-49464585 on Build GRCh38
rs768780600 Chr.19: 49464519-49464519 on Build GRCh38
rs779475910 Chr.19: 49464515-49464515 on Build GRCh38
rs146453544 Chr.19: 49464667-49464667 on Build GRCh38
rs566399437 Chr.19: 49464342-49464342 on Build GRCh38
rs371432624 Chr.19: 49464559-49464559 on Build GRCh38
rs766049631 Chr.19: 49464556-49464556 on Build GRCh38
rs371592208 Chr.19: 49464593-49464593 on Build GRCh38
rs777479119 Chr.19: 49464566-49464566 on Build GRCh38
rs138995716 Chr.19: 49464650-49464650 on Build GRCh38
rs778174915 Chr.19: 49464706-49464706 on Build GRCh38
rs760420970 Chr.19: 49464793-49464793 on Build GRCh38
rs370566117 Chr.19: 49464539-49464539 on Build GRCh38
rs750274439 Chr.19: 49464743-49464743 on Build GRCh38
rs4802609 Chr.19: 49464470-49464470 on Build GRCh38
rs545337650 Chr.19: 49464334-49464334 on Build GRCh38
rs779552359 Chr.19: 49464769-49464769 on Build GRCh38
rs540018165 Chr.19: 49464344-49464344 on Build GRCh38
rs560383515 Chr.19: 49464549-49464549 on Build GRCh38
rs778725984 Chr.19: 49464674-49464674 on Build GRCh38
rs763313609 Chr.19: 49464417-49464417 on Build GRCh38
rs774666530 Chr.19: 49464523-49464523 on Build GRCh38
rs771412846 Chr.19: 49464451-49464451 on Build GRCh38
rs749099899 Chr.19: 49464615-49464615 on Build GRCh38
rs755256724 Chr.19: 49464679-49464679 on Build GRCh38
rs527613185 Chr.19: 49464510-49464510 on Build GRCh38
rs768690823 Chr.19: 49464467-49464467 on Build GRCh38
rs747393130 Chr.19: 49464596-49464597 on Build GRCh38
rs772501763 Chr.19: 49464772-49464772 on Build GRCh38
rs781260919 Chr.19: 49464520-49464520 on Build GRCh38
rs200088188 Chr.19: 49464651-49464651 on Build GRCh38
rs11558188 Chr.19: 49464724-49464724 on Build GRCh38
rs372663614 Chr.19: 49464619-49464619 on Build GRCh38
rs151226186 Chr.19: 49464641-49464641 on Build GRCh38
rs144710024 Chr.19: 49464437-49464437 on Build GRCh38
rs781377623 Chr.19: 49464699-49464699 on Build GRCh38
rs369626137 Chr.19: 49464592-49464592 on Build GRCh38
rs746495747 Chr.19: 49464518-49464518 on Build GRCh38
rs375713785 Chr.19: 49464511-49464511 on Build GRCh38
rs754273617 Chr.19: 49464542-49464542 on Build GRCh38
rs747180806 Chr.19: 49464621-49464621 on Build GRCh38
rs763939268 Chr.19: 49464405-49464405 on Build GRCh38
rs761222144 Chr.19: 49464490-49464490 on Build GRCh38
rs139981295 Chr.19: 49464445-49464445 on Build GRCh38
rs145782809 Chr.19: 49464672-49464672 on Build GRCh38
rs868744182 Chr.19: 49464776-49464776 on Build GRCh38
rs368739759 Chr.19: 49464570-49464570 on Build GRCh38
rs900116709 Chr.19: 49464353-49464353 on Build GRCh38
rs767088349 Chr.19: 49464603-49464603 on Build GRCh38
rs749754755 Chr.19: 49464718-49464718 on Build GRCh38
rs559991892 Chr.19: 49464488-49464488 on Build GRCh38
rs367667391 Chr.19: 49464557-49464557 on Build GRCh38
rs776728408 Chr.19: 49464627-49464627 on Build GRCh38
rs1042090201 Chr.19: 49464462-49464462 on Build GRCh38
rs372218834 Chr.19: 49464738-49464738 on Build GRCh38
rs766007240 Chr.19: 49464550-49464558 on Build GRCh38
rs373759432 Chr.19: 49464376-49464376 on Build GRCh38
rs200873076 Chr.19: 49464655-49464655 on Build GRCh38
rs914343728 Chr.19: 49464792-49464792 on Build GRCh38
rs149196725 Chr.19: 49464719-49464719 on Build GRCh38
rs773590065 Chr.19: 49464412-49464412 on Build GRCh38
rs757608621 Chr.19: 49464504-49464504 on Build GRCh38
rs111803922 Chr.19: 49464450-49464450 on Build GRCh38
rs752233539 Chr.19: 49464607-49464607 on Build GRCh38
rs201895597 Chr.19: 49464522-49464522 on Build GRCh38
rs375093482 Chr.19: 49464433-49464433 on Build GRCh38
rs558103745 Chr.19: 49464346-49464346 on Build GRCh38
rs898170516 Chr.19: 49464702-49464702 on Build GRCh38
rs775474907 Chr.19: 49464536-49464536 on Build GRCh38
rs374982664 Chr.19: 49464399-49464399 on Build GRCh38
rs771634532 Chr.19: 49464767-49464767 on Build GRCh38
rs924602848 Chr.19: 49464597-49464597 on Build GRCh38
rs756048720 Chr.19: 49464613-49464613 on Build GRCh38
rs776172585 Chr.19: 49464630-49464631 on Build GRCh38
rs893083778 Chr.19: 49464369-49464369 on Build GRCh38
rs959208467 Chr.19: 49464575-49464575 on Build GRCh38
rs756171469 Chr.19: 49464761-49464761 on Build GRCh38
rs141388688 Chr.19: 49464745-49464745 on Build GRCh38
rs147203240 Chr.19: 49464461-49464461 on Build GRCh38
rs564414489 Chr.19: 49464770-49464770 on Build GRCh38
rs760111667 Chr.19: 49464732-49464732 on Build GRCh38
rs751727995 Chr.19: 49464429-49464429 on Build GRCh38
rs199956596 Chr.19: 49464757-49464757 on Build GRCh38
rs142205566 Chr.19: 49464521-49464521 on Build GRCh38
rs1041815131 Chr.19: 49464343-49464343 on Build GRCh38
rs772118363 Chr.19: 49464396-49464396 on Build GRCh38
rs368289777 Chr.19: 49464441-49464441 on Build GRCh38
rs752325190 Chr.19: 49464669-49464669 on Build GRCh38
rs776606895 Chr.19: 49464468-49464468 on Build GRCh38
rs114984918 Chr.19: 49464591-49464591 on Build GRCh38
rs10422648 Chr.19: 49464656-49464656 on Build GRCh38
rs771336085 Chr.19: 49464476-49464476 on Build GRCh38
rs775040837 Chr.19: 49464774-49464774 on Build GRCh38
rs369105748 Chr.19: 49464403-49464403 on Build GRCh38
rs541495383 Chr.19: 49464586-49464586 on Build GRCh38
rs200796027 Chr.19: 49464498-49464498 on Build GRCh38
rs760383838 Chr.19: 49464549-49464550 on Build GRCh38
rs140593990 Chr.19: 49464494-49464494 on Build GRCh38
rs774898389 Chr.19: 49464723-49464723 on Build GRCh38
rs777958673 Chr.19: 49464765-49464765 on Build GRCh38
rs2293009 Chr.19: 49464423-49464423 on Build GRCh38
rs748605370 Chr.19: 49464366-49464366 on Build GRCh38
rs759629351 Chr.19: 49464525-49464525 on Build GRCh38
rs998719705 Chr.19: 49464359-49464359 on Build GRCh38
rs146615636 Chr.19: 49464678-49464678 on Build GRCh38
rs754271873 Chr.19: 49464496-49464496 on Build GRCh38
rs756534916 Chr.19: 49464436-49464436 on Build GRCh38
rs753080420 Chr.19: 49464435-49464435 on Build GRCh38
rs749297466 Chr.19: 49464766-49464766 on Build GRCh38
rs76240224 Chr.19: 49464680-49464680 on Build GRCh38
rs888777884 Chr.19: 49464335-49464335 on Build GRCh38
rs755001290 Chr.19: 49464430-49464430 on Build GRCh38
rs370793991 Chr.19: 49464386-49464386 on Build GRCh38
rs761847775 Chr.19: 49464740-49464740 on Build GRCh38

View Full Product Details