Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49799753-49800282 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP2A1
Entrez Gene ID
160
Gene Name
adaptor related protein complex 2 alpha 1 subunit
Gene Aliases
ADTAA, AP2-ALPHA, CLAPA1
UniGene
Hs.467125
Gene Chromosome Location
Chr.19: 49766923-49807114 on Build GRCh38
Transcripts
NM_014203.2 NM_130787.2 XM_011526556.2 XM_011526557.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5056836 c.1352G>A p.R451H Chr.19: 49800047-49800047 on Build GRCh38
2753316 c.1272C>T p.I424I Chr.19: 49799766-49799766 on Build GRCh38
713377 c.1436C>G p.A479G Chr.19: 49800131-49800131 on Build GRCh38
1304926 c.1391G>A p.R464H Chr.19: 49800086-49800086 on Build GRCh38
3536965 c.1445C>T p.T482I Chr.19: 49800140-49800140 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745506973 Chr.19: 49799962-49799962 on Build GRCh38
rs371530889 Chr.19: 49800133-49800133 on Build GRCh38
rs897618513 Chr.19: 49800221-49800221 on Build GRCh38
rs1017229429 Chr.19: 49799858-49799858 on Build GRCh38
rs770288996 Chr.19: 49799982-49799982 on Build GRCh38
rs763288834 Chr.19: 49799796-49799797 on Build GRCh38
rs781014128 Chr.19: 49799943-49799943 on Build GRCh38
rs201639831 Chr.19: 49800175-49800175 on Build GRCh38
rs374354429 Chr.19: 49800141-49800141 on Build GRCh38
rs73590490 Chr.19: 49799941-49799941 on Build GRCh38
rs763787974 Chr.19: 49800158-49800158 on Build GRCh38
rs748476589 Chr.19: 49800110-49800110 on Build GRCh38
rs896697817 Chr.19: 49799957-49799957 on Build GRCh38
rs768733259 Chr.19: 49799942-49799943 on Build GRCh38
rs914383544 Chr.19: 49800230-49800230 on Build GRCh38
rs140072315 Chr.19: 49800202-49800202 on Build GRCh38
rs771936918 Chr.19: 49800194-49800195 on Build GRCh38
rs770655556 Chr.19: 49799791-49799791 on Build GRCh38
rs752485707 Chr.19: 49800033-49800033 on Build GRCh38
rs201906388 Chr.19: 49800000-49800000 on Build GRCh38
rs1026068157 Chr.19: 49799776-49799776 on Build GRCh38
rs986806883 Chr.19: 49800180-49800180 on Build GRCh38
rs749773354 Chr.19: 49800118-49800118 on Build GRCh38
rs368404769 Chr.19: 49800161-49800161 on Build GRCh38
rs185296139 Chr.19: 49799782-49799782 on Build GRCh38
rs774463341 Chr.19: 49800004-49800004 on Build GRCh38
rs755250608 Chr.19: 49799942-49799942 on Build GRCh38
rs376823198 Chr.19: 49800184-49800184 on Build GRCh38
rs559611052 Chr.19: 49800215-49800215 on Build GRCh38
rs778547986 Chr.19: 49800069-49800069 on Build GRCh38
rs146010013 Chr.19: 49799991-49799991 on Build GRCh38
rs767661333 Chr.19: 49800021-49800021 on Build GRCh38
rs756706797 Chr.19: 49800099-49800099 on Build GRCh38
rs200754162 Chr.19: 49799964-49799964 on Build GRCh38
rs189554440 Chr.19: 49799923-49799923 on Build GRCh38
rs770048090 Chr.19: 49800114-49800114 on Build GRCh38
rs751157666 Chr.19: 49800190-49800190 on Build GRCh38
rs946815740 Chr.19: 49800251-49800253 on Build GRCh38
rs965003478 Chr.19: 49799899-49799899 on Build GRCh38
rs915533421 Chr.19: 49799826-49799826 on Build GRCh38
rs563879994 Chr.19: 49800256-49800256 on Build GRCh38
rs749904873 Chr.19: 49800181-49800181 on Build GRCh38
rs189561929 Chr.19: 49800227-49800227 on Build GRCh38
rs773683243 Chr.19: 49800115-49800115 on Build GRCh38
rs533348822 Chr.19: 49799811-49799811 on Build GRCh38
rs761892112 Chr.19: 49799956-49799956 on Build GRCh38
rs915233026 Chr.19: 49800166-49800166 on Build GRCh38
rs1054184861 Chr.19: 49799822-49799822 on Build GRCh38
rs376939716 Chr.19: 49799952-49799952 on Build GRCh38
rs779596720 Chr.19: 49800100-49800100 on Build GRCh38
rs757324763 Chr.19: 49800063-49800063 on Build GRCh38
rs767515085 Chr.19: 49800197-49800197 on Build GRCh38
rs977198596 Chr.19: 49800027-49800027 on Build GRCh38
rs770393212 Chr.19: 49799785-49799785 on Build GRCh38
rs759719552 Chr.19: 49800012-49800012 on Build GRCh38
rs777302536 Chr.19: 49799784-49799784 on Build GRCh38
rs778636010 Chr.19: 49799960-49799960 on Build GRCh38
rs763624613 Chr.19: 49800022-49800022 on Build GRCh38
rs759631724 Chr.19: 49800138-49800138 on Build GRCh38
rs184250533 Chr.19: 49800057-49800057 on Build GRCh38
rs753456849 Chr.19: 49800047-49800047 on Build GRCh38
rs149504055 Chr.19: 49799959-49799959 on Build GRCh38
rs766432944 Chr.19: 49800001-49800001 on Build GRCh38
rs894136196 Chr.19: 49800228-49800228 on Build GRCh38
rs774504131 Chr.19: 49799942-49799945 on Build GRCh38
rs570465699 Chr.19: 49799910-49799910 on Build GRCh38
rs774649528 Chr.19: 49799949-49799949 on Build GRCh38
rs779680239 Chr.19: 49800185-49800185 on Build GRCh38
rs750478513 Chr.19: 49799928-49799928 on Build GRCh38
rs764028672 Chr.19: 49800054-49800054 on Build GRCh38
rs576480748 Chr.19: 49800140-49800140 on Build GRCh38
rs551896115 Chr.19: 49799895-49799895 on Build GRCh38
rs551688871 Chr.19: 49800243-49800243 on Build GRCh38
rs199728232 Chr.19: 49799947-49799947 on Build GRCh38
rs930381743 Chr.19: 49800226-49800226 on Build GRCh38
rs1012564038 Chr.19: 49800239-49800239 on Build GRCh38
rs765298475 Chr.19: 49800179-49800179 on Build GRCh38
rs561974666 Chr.19: 49800176-49800176 on Build GRCh38
rs368042642 Chr.19: 49800132-49800132 on Build GRCh38
rs377250932 Chr.19: 49799796-49799796 on Build GRCh38
rs748493956 Chr.19: 49799972-49799972 on Build GRCh38
rs751774802 Chr.19: 49799936-49799936 on Build GRCh38
rs374167558 Chr.19: 49799924-49799924 on Build GRCh38
rs763032812 Chr.19: 49799933-49799933 on Build GRCh38
rs200552286 Chr.19: 49799935-49799935 on Build GRCh38
rs181564717 Chr.19: 49799954-49799954 on Build GRCh38

View Full Product Details