Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 57377045-57377555 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF547
Entrez Gene ID
284306
Gene Name
zinc finger protein 547
Gene Aliases
-
UniGene
Hs.446620
Gene Chromosome Location
Chr.19: 57363435-57379559 on Build GRCh38
Transcripts
NM_173631.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6525509 c.250C>A p.Q84K Chr.19: 57377226-57377226 on Build GRCh38
1397007 c.290A>G p.K97R Chr.19: 57377266-57377266 on Build GRCh38
6442975 c.292G>C p.D98H Chr.19: 57377268-57377268 on Build GRCh38
6152634 c.367C>T p.Q123* Chr.19: 57377343-57377343 on Build GRCh38
224010 c.407G>A p.G136E Chr.19: 57377383-57377383 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs538353346 Chr.19: 57377159-57377159 on Build GRCh38
rs927430482 Chr.19: 57377185-57377185 on Build GRCh38
rs373698675 Chr.19: 57377283-57377283 on Build GRCh38
rs377518505 Chr.19: 57377205-57377205 on Build GRCh38
rs766226161 Chr.19: 57377364-57377364 on Build GRCh38
rs867849999 Chr.19: 57377281-57377281 on Build GRCh38
rs544829574 Chr.19: 57377323-57377323 on Build GRCh38
rs995368661 Chr.19: 57377464-57377464 on Build GRCh38
rs10413160 Chr.19: 57377234-57377234 on Build GRCh38
rs141205853 Chr.19: 57377084-57377084 on Build GRCh38
rs768585756 Chr.19: 57377334-57377334 on Build GRCh38
rs138029123 Chr.19: 57377282-57377282 on Build GRCh38
rs1028988461 Chr.19: 57377465-57377465 on Build GRCh38
rs372728345 Chr.19: 57377098-57377098 on Build GRCh38
rs753423928 Chr.19: 57377257-57377258 on Build GRCh38
rs889601366 Chr.19: 57377286-57377286 on Build GRCh38
rs112341705 Chr.19: 57377376-57377376 on Build GRCh38
rs1019297076 Chr.19: 57377090-57377090 on Build GRCh38
rs778345904 Chr.19: 57377158-57377158 on Build GRCh38
rs755846192 Chr.19: 57377144-57377144 on Build GRCh38
rs958784321 Chr.19: 57377193-57377193 on Build GRCh38
rs748404990 Chr.19: 57377493-57377493 on Build GRCh38
rs151324019 Chr.19: 57377346-57377346 on Build GRCh38
rs761092222 Chr.19: 57377081-57377081 on Build GRCh38
rs201630393 Chr.19: 57377121-57377121 on Build GRCh38
rs767250581 Chr.19: 57377125-57377125 on Build GRCh38
rs201774240 Chr.19: 57377294-57377294 on Build GRCh38
rs934546849 Chr.19: 57377257-57377257 on Build GRCh38
rs965389774 Chr.19: 57377106-57377106 on Build GRCh38
rs370936486 Chr.19: 57377381-57377381 on Build GRCh38
rs546234801 Chr.19: 57377118-57377118 on Build GRCh38
rs747828436 Chr.19: 57377163-57377163 on Build GRCh38
rs773411586 Chr.19: 57377516-57377516 on Build GRCh38
rs531953082 Chr.19: 57377482-57377482 on Build GRCh38
rs561319969 Chr.19: 57377332-57377332 on Build GRCh38
rs201727290 Chr.19: 57377311-57377311 on Build GRCh38
rs114959714 Chr.19: 57377307-57377307 on Build GRCh38
rs761891228 Chr.19: 57377354-57377354 on Build GRCh38
rs141431125 Chr.19: 57377306-57377306 on Build GRCh38
rs766061812 Chr.19: 57377277-57377277 on Build GRCh38
rs753794377 Chr.19: 57377278-57377278 on Build GRCh38
rs1015177022 Chr.19: 57377429-57377429 on Build GRCh38
rs202125151 Chr.19: 57377425-57377425 on Build GRCh38
rs1012121867 Chr.19: 57377109-57377109 on Build GRCh38
rs752627277 Chr.19: 57377395-57377395 on Build GRCh38
rs779369087 Chr.19: 57377224-57377224 on Build GRCh38
rs749189364 Chr.19: 57377151-57377151 on Build GRCh38
rs139850139 Chr.19: 57377156-57377156 on Build GRCh38
rs778606519 Chr.19: 57377513-57377513 on Build GRCh38
rs750171240 Chr.19: 57377138-57377138 on Build GRCh38
rs201182688 Chr.19: 57377221-57377221 on Build GRCh38
rs751966845 Chr.19: 57377452-57377452 on Build GRCh38
rs200222024 Chr.19: 57377223-57377223 on Build GRCh38
rs369017713 Chr.19: 57377137-57377137 on Build GRCh38
rs867588796 Chr.19: 57377442-57377442 on Build GRCh38
rs150397433 Chr.19: 57377504-57377504 on Build GRCh38
rs775457284 Chr.19: 57377527-57377527 on Build GRCh38
rs756822993 Chr.19: 57377414-57377414 on Build GRCh38
rs575635555 Chr.19: 57377317-57377317 on Build GRCh38
rs373576598 Chr.19: 57377523-57377523 on Build GRCh38
rs760102586 Chr.19: 57377208-57377208 on Build GRCh38
rs764432403 Chr.19: 57377225-57377225 on Build GRCh38
rs751249464 Chr.19: 57377400-57377400 on Build GRCh38
rs746413433 Chr.19: 57377322-57377322 on Build GRCh38
rs770020777 Chr.19: 57377213-57377213 on Build GRCh38
rs759482432 Chr.19: 57377379-57377379 on Build GRCh38
rs763155611 Chr.19: 57377333-57377333 on Build GRCh38
rs771687206 Chr.19: 57377181-57377181 on Build GRCh38
rs777126015 Chr.19: 57377399-57377399 on Build GRCh38
rs201194074 Chr.19: 57377398-57377398 on Build GRCh38
rs146966640 Chr.19: 57377218-57377218 on Build GRCh38
rs894855046 Chr.19: 57377365-57377365 on Build GRCh38
rs370434517 Chr.19: 57377520-57377520 on Build GRCh38
rs773463996 Chr.19: 57377189-57377189 on Build GRCh38
rs202089799 Chr.19: 57377197-57377197 on Build GRCh38
rs372467148 Chr.19: 57377080-57377080 on Build GRCh38
rs749733667 Chr.19: 57377454-57377454 on Build GRCh38
rs77834888 Chr.19: 57377435-57377435 on Build GRCh38
rs548325402 Chr.19: 57377295-57377295 on Build GRCh38
rs752146844 Chr.19: 57377285-57377285 on Build GRCh38
rs546847485 Chr.19: 57377427-57377427 on Build GRCh38
rs994233631 Chr.19: 57377343-57377343 on Build GRCh38
rs1053346603 Chr.19: 57377351-57377351 on Build GRCh38
rs754538745 Chr.19: 57377316-57377317 on Build GRCh38
rs372503408 Chr.19: 57377147-57377147 on Build GRCh38
rs772446130 Chr.19: 57377078-57377078 on Build GRCh38
rs369291877 Chr.19: 57377087-57377087 on Build GRCh38
rs769369323 Chr.19: 57377324-57377324 on Build GRCh38

View Full Product Details