Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 57621624-57622143 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF134
Entrez Gene ID
7693
Gene Name
zinc finger protein 134
Gene Aliases
pHZ-15
UniGene
Hs.469694
Gene Chromosome Location
Chr.19: 57614219-57624717 on Build GRCh38
Transcripts
NM_003435.4

SNP Detail

SNP ID Chromosome Location
rs962009480 Chr.19: 57621843-57621843 on Build GRCh38
rs763238578 Chr.19: 57621752-57621752 on Build GRCh38
rs774551502 Chr.19: 57621839-57621839 on Build GRCh38
rs1028572903 Chr.19: 57621792-57621792 on Build GRCh38
rs1053373634 Chr.19: 57621874-57621874 on Build GRCh38
rs182974945 Chr.19: 57621884-57621884 on Build GRCh38
rs572275072 Chr.19: 57621941-57621941 on Build GRCh38
rs899231492 Chr.19: 57621978-57621978 on Build GRCh38
rs576600180 Chr.19: 57621712-57621712 on Build GRCh38
rs936378630 Chr.19: 57621871-57621871 on Build GRCh38
rs570446803 Chr.19: 57621845-57621845 on Build GRCh38
rs568674568 Chr.19: 57621746-57621746 on Build GRCh38
rs945010938 Chr.19: 57621981-57621981 on Build GRCh38
rs1049610707 Chr.19: 57622057-57622057 on Build GRCh38
rs553831917 Chr.19: 57621827-57621827 on Build GRCh38
rs868121055 Chr.19: 57621790-57621790 on Build GRCh38
rs1056083806 Chr.19: 57621682-57621682 on Build GRCh38
rs939595892 Chr.19: 57621679-57621679 on Build GRCh38
rs892111277 Chr.19: 57621711-57621711 on Build GRCh38
rs879487563 Chr.19: 57622095-57622095 on Build GRCh38
rs1015050590 Chr.19: 57621955-57621955 on Build GRCh38
rs890945922 Chr.19: 57622119-57622119 on Build GRCh38
rs770227314 Chr.19: 57621730-57621730 on Build GRCh38
rs963446027 Chr.19: 57621747-57621747 on Build GRCh38
rs906376778 Chr.19: 57621942-57621942 on Build GRCh38
rs926778227 Chr.19: 57621921-57621921 on Build GRCh38
rs10403536 Chr.19: 57621875-57621875 on Build GRCh38
rs905482094 Chr.19: 57621665-57621665 on Build GRCh38
rs774903924 Chr.19: 57621674-57621674 on Build GRCh38
rs953333609 Chr.19: 57622006-57622006 on Build GRCh38
rs973400082 Chr.19: 57621751-57621751 on Build GRCh38
rs541671079 Chr.19: 57621935-57621935 on Build GRCh38
rs1005117666 Chr.19: 57621948-57621948 on Build GRCh38
rs555654186 Chr.19: 57621906-57621906 on Build GRCh38
rs749631532 Chr.19: 57621652-57621652 on Build GRCh38
rs559496247 Chr.19: 57622004-57622004 on Build GRCh38
rs753615009 Chr.19: 57622050-57622050 on Build GRCh38
rs970946643 Chr.19: 57621722-57621722 on Build GRCh38
rs980996168 Chr.19: 57621914-57621914 on Build GRCh38
rs953596932 Chr.19: 57621812-57621812 on Build GRCh38
rs555505751 Chr.19: 57621936-57621936 on Build GRCh38
rs1012065921 Chr.19: 57621648-57621648 on Build GRCh38
rs567212752 Chr.19: 57622003-57622003 on Build GRCh38
rs903694164 Chr.19: 57621738-57621738 on Build GRCh38
rs560152579 Chr.19: 57622017-57622017 on Build GRCh38
rs773556583 Chr.19: 57621735-57621735 on Build GRCh38
rs115775338 Chr.19: 57621879-57621879 on Build GRCh38
rs766787164 Chr.19: 57621838-57621838 on Build GRCh38
rs190036746 Chr.19: 57621825-57621825 on Build GRCh38
rs1056616303 Chr.19: 57621951-57621951 on Build GRCh38
rs1042102 Chr.19: 57621728-57621728 on Build GRCh38

View Full Product Details