Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 57633077-57633580 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF211
Entrez Gene ID
10520
Gene Name
zinc finger protein 211
Gene Aliases
C2H2-25, CH2H2-25, ZNF-25
UniGene
Hs.590977
Gene Chromosome Location
Chr.19: 57633167-57644046 on Build GRCh38
Transcripts
NM_001265597.2 NM_001265598.2 NM_001265599.2 NM_001265600.2 NM_001322306.1 NM_006385.4 NM_198855.3 NR_049752.2 NR_049753.2 NR_049754.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6828506 c.89C>T p.S30L Chr.19: 57633435-57633435 on Build GRCh38
1397144 c.1A>G p.M1V Chr.19: 57633347-57633347 on Build GRCh38
6403703 c.20G>A p.G7D Chr.19: 57633366-57633366 on Build GRCh38
4621081 c.59G>T p.R20L Chr.19: 57633405-57633405 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs573805036 Chr.19: 57633265-57633265 on Build GRCh38
rs943869094 Chr.19: 57633273-57633273 on Build GRCh38
rs201176921 Chr.19: 57633387-57633387 on Build GRCh38
rs775291795 Chr.19: 57633357-57633358 on Build GRCh38
rs77671222 Chr.19: 57633506-57633506 on Build GRCh38
rs773420264 Chr.19: 57633249-57633249 on Build GRCh38
rs762825836 Chr.19: 57633358-57633358 on Build GRCh38
rs1053790113 Chr.19: 57633336-57633336 on Build GRCh38
rs370362787 Chr.19: 57633382-57633382 on Build GRCh38
rs764701761 Chr.19: 57633448-57633448 on Build GRCh38
rs936747104 Chr.19: 57633537-57633537 on Build GRCh38
rs1025156823 Chr.19: 57633215-57633215 on Build GRCh38
rs773194514 Chr.19: 57633469-57633469 on Build GRCh38
rs766394224 Chr.19: 57633260-57633260 on Build GRCh38
rs758794412 Chr.19: 57633442-57633442 on Build GRCh38
rs771800398 Chr.19: 57633466-57633466 on Build GRCh38
rs773995396 Chr.19: 57633381-57633381 on Build GRCh38
rs769151805 Chr.19: 57633554-57633554 on Build GRCh38
rs746152307 Chr.19: 57633459-57633459 on Build GRCh38
rs759076666 Chr.19: 57633413-57633413 on Build GRCh38
rs781487802 Chr.19: 57633369-57633369 on Build GRCh38
rs577601973 Chr.19: 57633160-57633161 on Build GRCh38
rs779137984 Chr.19: 57633354-57633354 on Build GRCh38
rs746913909 Chr.19: 57633472-57633472 on Build GRCh38
rs370924466 Chr.19: 57633345-57633345 on Build GRCh38
rs767365367 Chr.19: 57633292-57633292 on Build GRCh38
rs10420097 Chr.19: 57633193-57633193 on Build GRCh38
rs919378861 Chr.19: 57633515-57633515 on Build GRCh38
rs776409217 Chr.19: 57633405-57633405 on Build GRCh38
rs760193898 Chr.19: 57633389-57633389 on Build GRCh38
rs919855551 Chr.19: 57633131-57633131 on Build GRCh38
rs979338785 Chr.19: 57633156-57633156 on Build GRCh38
rs765678912 Chr.19: 57633423-57633423 on Build GRCh38
rs753292447 Chr.19: 57633359-57633359 on Build GRCh38
rs1000732532 Chr.19: 57633408-57633408 on Build GRCh38
rs991868378 Chr.19: 57633209-57633209 on Build GRCh38
rs929514774 Chr.19: 57633330-57633330 on Build GRCh38
rs537111962 Chr.19: 57633187-57633187 on Build GRCh38
rs906722411 Chr.19: 57633352-57633352 on Build GRCh38
rs943925557 Chr.19: 57633507-57633507 on Build GRCh38
rs9749442 Chr.19: 57633151-57633151 on Build GRCh38
rs377630462 Chr.19: 57633529-57633529 on Build GRCh38
rs553271755 Chr.19: 57633324-57633324 on Build GRCh38
rs966603236 Chr.19: 57633214-57633214 on Build GRCh38
rs759033392 Chr.19: 57633479-57633479 on Build GRCh38
rs944762319 Chr.19: 57633162-57633162 on Build GRCh38
rs768326141 Chr.19: 57633321-57633321 on Build GRCh38
rs757427591 Chr.19: 57633452-57633452 on Build GRCh38
rs1026640581 Chr.19: 57633461-57633461 on Build GRCh38
rs768536569 Chr.19: 57633377-57633377 on Build GRCh38
rs28721436 Chr.19: 57633492-57633492 on Build GRCh38
rs113714661 Chr.19: 57633195-57633195 on Build GRCh38
rs369869811 Chr.19: 57633410-57633410 on Build GRCh38
rs376127475 Chr.19: 57633320-57633320 on Build GRCh38
rs780213750 Chr.19: 57633465-57633465 on Build GRCh38
rs879074307 Chr.19: 57633164-57633164 on Build GRCh38
rs970913345 Chr.19: 57633222-57633222 on Build GRCh38
rs925245885 Chr.19: 57633225-57633225 on Build GRCh38
rs201650717 Chr.19: 57633363-57633363 on Build GRCh38
rs778374400 Chr.19: 57633362-57633362 on Build GRCh38
rs747893937 Chr.19: 57633385-57633385 on Build GRCh38
rs781103212 Chr.19: 57633454-57633454 on Build GRCh38
rs542451879 Chr.19: 57633267-57633267 on Build GRCh38
rs1050837474 Chr.19: 57633160-57633160 on Build GRCh38
rs567367930 Chr.19: 57633517-57633517 on Build GRCh38
rs938251406 Chr.19: 57633538-57633538 on Build GRCh38
rs571529807 Chr.19: 57633129-57633129 on Build GRCh38
rs765695321 Chr.19: 57633246-57633246 on Build GRCh38
rs565395390 Chr.19: 57633390-57633390 on Build GRCh38
rs575101411 Chr.19: 57633241-57633241 on Build GRCh38
rs932546595 Chr.19: 57633150-57633150 on Build GRCh38
rs777749679 Chr.19: 57633477-57633477 on Build GRCh38
rs10420575 Chr.19: 57633180-57633180 on Build GRCh38
rs1053865737 Chr.19: 57633557-57633557 on Build GRCh38
rs979672673 Chr.19: 57633216-57633216 on Build GRCh38
rs919551207 Chr.19: 57633253-57633253 on Build GRCh38
rs187290810 Chr.19: 57633212-57633212 on Build GRCh38
rs533039689 Chr.19: 57633349-57633349 on Build GRCh38
rs114740943 Chr.19: 57633256-57633256 on Build GRCh38
rs749962838 Chr.19: 57633344-57633344 on Build GRCh38
rs550828894 Chr.19: 57633165-57633165 on Build GRCh38
rs936717809 Chr.19: 57633312-57633312 on Build GRCh38
rs576184512 Chr.19: 57633160-57633160 on Build GRCh38
rs767063223 Chr.19: 57633332-57633332 on Build GRCh38
rs367833699 Chr.19: 57633161-57633161 on Build GRCh38
rs752053466 Chr.19: 57633451-57633451 on Build GRCh38
rs761708411 Chr.19: 57633331-57633331 on Build GRCh38
rs756312785 Chr.19: 57633464-57633464 on Build GRCh38
rs959629128 Chr.19: 57633206-57633206 on Build GRCh38
rs143187549 Chr.19: 57633521-57633521 on Build GRCh38
rs769074682 Chr.19: 57633192-57633192 on Build GRCh38
rs973757029 Chr.19: 57633455-57633455 on Build GRCh38
rs544726019 Chr.19: 57633522-57633522 on Build GRCh38
rs112876388 Chr.19: 57633159-57633160 on Build GRCh38
rs199670185 Chr.19: 57633372-57633372 on Build GRCh38
rs558643772 Chr.19: 57633224-57633224 on Build GRCh38
rs896737244 Chr.19: 57633555-57633555 on Build GRCh38
rs534612596 Chr.19: 57633110-57633110 on Build GRCh38
rs144205771 Chr.19: 57633386-57633386 on Build GRCh38
rs9749449 Chr.19: 57633347-57633347 on Build GRCh38
rs1049248757 Chr.19: 57633525-57633525 on Build GRCh38
rs540576553 Chr.19: 57633311-57633311 on Build GRCh38
rs1033318360 Chr.19: 57633194-57633194 on Build GRCh38
rs369088499 Chr.19: 57633251-57633251 on Build GRCh38
rs758946683 Chr.19: 57633360-57633360 on Build GRCh38
rs368204022 Chr.19: 57633350-57633350 on Build GRCh38
rs770511818 Chr.19: 57633476-57633476 on Build GRCh38
rs1012538471 Chr.19: 57633346-57633346 on Build GRCh38
rs902612642 Chr.19: 57633290-57633290 on Build GRCh38
rs374691145 Chr.19: 57633435-57633435 on Build GRCh38
rs1042896764 Chr.19: 57633278-57633278 on Build GRCh38
rs961387861 Chr.19: 57633250-57633250 on Build GRCh38
rs1055417852 Chr.19: 57633541-57633541 on Build GRCh38

View Full Product Details