Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 58068781-58069284 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF135
Entrez Gene ID
7694
Gene Name
zinc finger protein 135
Gene Aliases
ZNF61, ZNF78L1, pHZ-17, pT3
UniGene
Hs.85863
Gene Chromosome Location
Chr.19: 58059227-58069755 on Build GRCh38
Transcripts
NM_001164529.1 NM_001164530.1 NM_001289401.1 NM_001289402.1 NM_003436.3 NM_007134.1 XM_005259211.1 XM_006723362.3 XM_006723363.3 XM_017027240.1 XM_017027241.1 XM_017027242.1

SNP Detail

SNP ID Chromosome Location
rs1004270074 Chr.19: 58068919-58068919 on Build GRCh38
rs56951330 Chr.19: 58069015-58069015 on Build GRCh38
rs530257247 Chr.19: 58069098-58069098 on Build GRCh38
rs569540723 Chr.19: 58069246-58069246 on Build GRCh38
rs914249119 Chr.19: 58069072-58069072 on Build GRCh38
rs566884504 Chr.19: 58069129-58069129 on Build GRCh38
rs559270011 Chr.19: 58068840-58068840 on Build GRCh38
rs757642129 Chr.19: 58068889-58068889 on Build GRCh38
rs189111903 Chr.19: 58069116-58069116 on Build GRCh38
rs962663380 Chr.19: 58068922-58068922 on Build GRCh38
rs60681775 Chr.19: 58068917-58068918 on Build GRCh38
rs554727160 Chr.19: 58068871-58068871 on Build GRCh38
rs754874159 Chr.19: 58068841-58068841 on Build GRCh38
rs117509647 Chr.19: 58068987-58068987 on Build GRCh38
rs191884696 Chr.19: 58068869-58068869 on Build GRCh38
rs375382274 Chr.19: 58068928-58068928 on Build GRCh38
rs538638520 Chr.19: 58069165-58069165 on Build GRCh38
rs752329647 Chr.19: 58068907-58068907 on Build GRCh38
rs766264045 Chr.19: 58068808-58068808 on Build GRCh38
rs562409149 Chr.19: 58069055-58069055 on Build GRCh38
rs955810374 Chr.19: 58069057-58069057 on Build GRCh38
rs567666729 Chr.19: 58069075-58069075 on Build GRCh38
rs544778781 Chr.19: 58068870-58068870 on Build GRCh38
rs138422284 Chr.19: 58068924-58068924 on Build GRCh38
rs1057402 Chr.19: 58068909-58068909 on Build GRCh38
rs531424671 Chr.19: 58069060-58069060 on Build GRCh38
rs773163690 Chr.19: 58068925-58068925 on Build GRCh38
rs1016964159 Chr.19: 58068920-58068920 on Build GRCh38
rs12973718 Chr.19: 58069247-58069247 on Build GRCh38
rs758941244 Chr.19: 58069035-58069035 on Build GRCh38
rs1013175725 Chr.19: 58068930-58068930 on Build GRCh38
rs767429661 Chr.19: 58068903-58068903 on Build GRCh38
rs886248220 Chr.19: 58069196-58069196 on Build GRCh38
rs751361899 Chr.19: 58068809-58068809 on Build GRCh38
rs997756866 Chr.19: 58068826-58068826 on Build GRCh38
rs1051213397 Chr.19: 58068832-58068832 on Build GRCh38
rs555159106 Chr.19: 58069257-58069257 on Build GRCh38
rs60372547 Chr.19: 58069179-58069179 on Build GRCh38
rs11669919 Chr.19: 58069026-58069026 on Build GRCh38
rs933199226 Chr.19: 58069130-58069130 on Build GRCh38
rs543629912 Chr.19: 58069253-58069254 on Build GRCh38
rs183319182 Chr.19: 58069070-58069070 on Build GRCh38

View Full Product Details