Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 10577557-10578067 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NOL10
Entrez Gene ID
79954
Gene Name
nucleolar protein 10
Gene Aliases
PQBP5
UniGene
Hs.222494
Gene Chromosome Location
Chr.2: 10570766-10689991 on Build GRCh38
Transcripts
NM_001261392.1 NM_001261394.1 NM_024894.3 NR_048552.1 XM_011510398.2 XM_011510399.2 XM_017004990.1 XM_017004991.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2981731 c.1938G>A p.T646T Chr.2: 10577645-10577645 on Build GRCh38
3564795 c.1743G>A p.L581L Chr.2: 10577690-10577690 on Build GRCh38
4001119 c.1753G>A p.D585N Chr.2: 10577680-10577680 on Build GRCh38
7094776 c.1711C>T p.R571C Chr.2: 10577722-10577722 on Build GRCh38
3693721 c.1917C>T p.G639G Chr.2: 10577666-10577666 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779403728 Chr.2: 10577768-10577768 on Build GRCh38
rs199612731 Chr.2: 10577619-10577619 on Build GRCh38
rs772171165 Chr.2: 10577627-10577628 on Build GRCh38
rs1014955318 Chr.2: 10578031-10578031 on Build GRCh38
rs139579430 Chr.2: 10578034-10578034 on Build GRCh38
rs907967632 Chr.2: 10577705-10577705 on Build GRCh38
rs184990484 Chr.2: 10577928-10577928 on Build GRCh38
rs750035042 Chr.2: 10577761-10577761 on Build GRCh38
rs954714145 Chr.2: 10577671-10577671 on Build GRCh38
rs760665036 Chr.2: 10577710-10577710 on Build GRCh38
rs7560546 Chr.2: 10577657-10577657 on Build GRCh38
rs752159056 Chr.2: 10577772-10577772 on Build GRCh38
rs180951117 Chr.2: 10577822-10577822 on Build GRCh38
rs1003313853 Chr.2: 10577646-10577646 on Build GRCh38
rs138862112 Chr.2: 10577649-10577649 on Build GRCh38
rs760662007 Chr.2: 10577873-10577873 on Build GRCh38
rs983727812 Chr.2: 10578012-10578012 on Build GRCh38
rs757789453 Chr.2: 10577643-10577643 on Build GRCh38
rs200543723 Chr.2: 10577681-10577681 on Build GRCh38
rs372970827 Chr.2: 10577602-10577602 on Build GRCh38
rs891584061 Chr.2: 10577824-10577824 on Build GRCh38
rs769756550 Chr.2: 10577777-10577778 on Build GRCh38
rs935508126 Chr.2: 10577819-10577819 on Build GRCh38
rs376868734 Chr.2: 10577677-10577677 on Build GRCh38
rs761634365 Chr.2: 10577742-10577742 on Build GRCh38
rs781516883 Chr.2: 10577672-10577672 on Build GRCh38
rs746172794 Chr.2: 10577622-10577622 on Build GRCh38
rs115718510 Chr.2: 10578003-10578003 on Build GRCh38
rs142465541 Chr.2: 10577764-10577764 on Build GRCh38
rs369260781 Chr.2: 10577697-10577697 on Build GRCh38
rs747470027 Chr.2: 10577655-10577655 on Build GRCh38
rs1016786924 Chr.2: 10578015-10578015 on Build GRCh38
rs34667465 Chr.2: 10577650-10577651 on Build GRCh38
rs763949569 Chr.2: 10577620-10577620 on Build GRCh38
rs773073994 Chr.2: 10577784-10577785 on Build GRCh38
rs9973456 Chr.2: 10578004-10578004 on Build GRCh38
rs3214369 Chr.2: 10577775-10577778 on Build GRCh38
rs916818553 Chr.2: 10577766-10577766 on Build GRCh38
rs752563225 Chr.2: 10577615-10577615 on Build GRCh38
rs747908343 Chr.2: 10577774-10577777 on Build GRCh38
rs12473957 Chr.2: 10577588-10577588 on Build GRCh38
rs778670486 Chr.2: 10577639-10577639 on Build GRCh38
rs534246420 Chr.2: 10577937-10577937 on Build GRCh38
rs199849845 Chr.2: 10577778-10577778 on Build GRCh38
rs781014617 Chr.2: 10577767-10577769 on Build GRCh38
rs753592229 Chr.2: 10577622-10577622 on Build GRCh38
rs779010981 Chr.2: 10577616-10577618 on Build GRCh38
rs763151272 Chr.2: 10577682-10577682 on Build GRCh38
rs2287059 Chr.2: 10577680-10577680 on Build GRCh38
rs775535748 Chr.2: 10577700-10577700 on Build GRCh38
rs1034815316 Chr.2: 10577943-10577943 on Build GRCh38
rs554862289 Chr.2: 10577894-10577894 on Build GRCh38
rs764956288 Chr.2: 10577743-10577743 on Build GRCh38
rs904870471 Chr.2: 10577910-10577910 on Build GRCh38
rs375907878 Chr.2: 10577768-10577770 on Build GRCh38
rs556363160 Chr.2: 10578028-10578028 on Build GRCh38
rs558670752 Chr.2: 10577985-10577985 on Build GRCh38
rs921972397 Chr.2: 10577790-10577790 on Build GRCh38
rs7570118 Chr.2: 10577881-10577881 on Build GRCh38
rs1011231871 Chr.2: 10577844-10577844 on Build GRCh38
rs11684446 Chr.2: 10577800-10577800 on Build GRCh38
rs753718070 Chr.2: 10577721-10577721 on Build GRCh38
rs2287058 Chr.2: 10577637-10577637 on Build GRCh38
rs550821395 Chr.2: 10577794-10577794 on Build GRCh38
rs150549863 Chr.2: 10577784-10577784 on Build GRCh38
rs900185355 Chr.2: 10577904-10577904 on Build GRCh38
rs753989200 Chr.2: 10577883-10577883 on Build GRCh38
rs764076325 Chr.2: 10577711-10577711 on Build GRCh38

View Full Product Details