Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 16561846-16562390 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM49A
Entrez Gene ID
81553
Gene Name
family with sequence similarity 49 member A
Gene Aliases
-
UniGene
Hs.467769
Gene Chromosome Location
Chr.2: 16549459-16665866 on Build GRCh38
Transcripts
NM_030797.3 XM_005262630.3 XM_006712108.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6509131 c.388G>T p.E130* Chr.2: 16562052-16562052 on Build GRCh38
1206370 c.367A>G p.R123G Chr.2: 16562073-16562073 on Build GRCh38
5440068 c.397G>T p.E133* Chr.2: 16562043-16562043 on Build GRCh38
1206371 c.418C>T p.R140* Chr.2: 16562022-16562022 on Build GRCh38
1008919 c.301A>C p.K101Q Chr.2: 16562139-16562139 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs544084290 Chr.2: 16562190-16562190 on Build GRCh38
rs765647821 Chr.2: 16562140-16562140 on Build GRCh38
rs959416262 Chr.2: 16562265-16562265 on Build GRCh38
rs776202730 Chr.2: 16562022-16562022 on Build GRCh38
rs901055767 Chr.2: 16562355-16562355 on Build GRCh38
rs947124661 Chr.2: 16561886-16561886 on Build GRCh38
rs962212475 Chr.2: 16562201-16562202 on Build GRCh38
rs552792588 Chr.2: 16561900-16561900 on Build GRCh38
rs762840433 Chr.2: 16562087-16562087 on Build GRCh38
rs868184603 Chr.2: 16562043-16562043 on Build GRCh38
rs563078046 Chr.2: 16562341-16562341 on Build GRCh38
rs150652954 Chr.2: 16562116-16562116 on Build GRCh38
rs754509996 Chr.2: 16562168-16562168 on Build GRCh38
rs780436971 Chr.2: 16562157-16562157 on Build GRCh38
rs934012114 Chr.2: 16562004-16562004 on Build GRCh38
rs918527006 Chr.2: 16562339-16562339 on Build GRCh38
rs1036912031 Chr.2: 16561930-16561930 on Build GRCh38
rs757674617 Chr.2: 16562139-16562139 on Build GRCh38
rs764375091 Chr.2: 16562134-16562134 on Build GRCh38
rs747929826 Chr.2: 16562044-16562044 on Build GRCh38
rs972705381 Chr.2: 16562220-16562220 on Build GRCh38
rs754154704 Chr.2: 16562135-16562135 on Build GRCh38
rs554484294 Chr.2: 16562191-16562191 on Build GRCh38
rs779532114 Chr.2: 16561963-16561963 on Build GRCh38
rs555744737 Chr.2: 16561890-16561890 on Build GRCh38
rs1041289219 Chr.2: 16562345-16562345 on Build GRCh38
rs746512351 Chr.2: 16562166-16562166 on Build GRCh38
rs773016980 Chr.2: 16562061-16562061 on Build GRCh38
rs6751062 Chr.2: 16562185-16562185 on Build GRCh38
rs938941856 Chr.2: 16562273-16562273 on Build GRCh38
rs746426096 Chr.2: 16561987-16561987 on Build GRCh38
rs769612268 Chr.2: 16562059-16562059 on Build GRCh38
rs878997822 Chr.2: 16562256-16562256 on Build GRCh38
rs780787743 Chr.2: 16562175-16562175 on Build GRCh38
rs779575597 Chr.2: 16561997-16561997 on Build GRCh38
rs1051568488 Chr.2: 16562351-16562351 on Build GRCh38
rs889898221 Chr.2: 16562083-16562083 on Build GRCh38
rs747843052 Chr.2: 16562177-16562177 on Build GRCh38
rs941320727 Chr.2: 16561927-16561927 on Build GRCh38
rs138782523 Chr.2: 16562282-16562282 on Build GRCh38
rs868158172 Chr.2: 16562054-16562054 on Build GRCh38
rs1020563542 Chr.2: 16562173-16562173 on Build GRCh38
rs982206954 Chr.2: 16562231-16562231 on Build GRCh38
rs897043348 Chr.2: 16561999-16561999 on Build GRCh38
rs1034676151 Chr.2: 16562234-16562234 on Build GRCh38
rs766235281 Chr.2: 16562089-16562089 on Build GRCh38
rs566566030 Chr.2: 16561912-16561912 on Build GRCh38
rs746504941 Chr.2: 16562017-16562017 on Build GRCh38
rs770890257 Chr.2: 16562189-16562189 on Build GRCh38
rs71447258 Chr.2: 16562069-16562069 on Build GRCh38
rs750847325 Chr.2: 16562152-16562152 on Build GRCh38
rs1042910254 Chr.2: 16561895-16561895 on Build GRCh38
rs186994744 Chr.2: 16561918-16561918 on Build GRCh38
rs775584664 Chr.2: 16562096-16562096 on Build GRCh38
rs758872214 Chr.2: 16562155-16562155 on Build GRCh38
rs540232879 Chr.2: 16562230-16562230 on Build GRCh38
rs574488655 Chr.2: 16562201-16562201 on Build GRCh38
rs13017146 Chr.2: 16561969-16561969 on Build GRCh38
rs779504378 Chr.2: 16562287-16562287 on Build GRCh38
rs991087908 Chr.2: 16561893-16561893 on Build GRCh38
rs146120289 Chr.2: 16562021-16562021 on Build GRCh38
rs749097610 Chr.2: 16562188-16562188 on Build GRCh38
rs192501458 Chr.2: 16562288-16562288 on Build GRCh38
rs924422905 Chr.2: 16562328-16562328 on Build GRCh38
rs372656752 Chr.2: 16562169-16562169 on Build GRCh38
rs546049966 Chr.2: 16562321-16562321 on Build GRCh38
rs1006968395 Chr.2: 16562107-16562107 on Build GRCh38
rs990581587 Chr.2: 16562281-16562281 on Build GRCh38
rs972511263 Chr.2: 16562293-16562293 on Build GRCh38

View Full Product Details