Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 25164502-25164997 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POMC
Entrez Gene ID
5443
Gene Name
proopiomelanocortin
Gene Aliases
ACTH, CLIP, LPH, MSH, NPP, POC
UniGene
Hs.1897
Gene Chromosome Location
Chr.2: 25160853-25168851 on Build GRCh38
Transcripts
NM_000939.3 NM_001035256.2 NM_001319204.1 NM_001319205.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4093259 c.22C>T p.R8C Chr.2: 25164751-25164751 on Build GRCh38
7449299 c.96C>G p.S32R Chr.2: 25164677-25164677 on Build GRCh38
4093258 c.111C>T p.L37L Chr.2: 25164662-25164662 on Build GRCh38
329554 c.23G>A p.R8H Chr.2: 25164750-25164750 on Build GRCh38
720765 c.5C>A p.P2Q Chr.2: 25164768-25164768 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs564736300 Chr.2: 25164559-25164559 on Build GRCh38
rs142715775 Chr.2: 25164656-25164656 on Build GRCh38
rs373955826 Chr.2: 25164604-25164604 on Build GRCh38
rs8192605 Chr.2: 25164755-25164755 on Build GRCh38
rs531531453 Chr.2: 25164567-25164567 on Build GRCh38
rs772771717 Chr.2: 25164530-25164530 on Build GRCh38
rs143923583 Chr.2: 25164699-25164699 on Build GRCh38
rs1018907588 Chr.2: 25164702-25164702 on Build GRCh38
rs780567223 Chr.2: 25164802-25164802 on Build GRCh38
rs768177368 Chr.2: 25164814-25164814 on Build GRCh38
rs756296347 Chr.2: 25164776-25164776 on Build GRCh38
rs750208860 Chr.2: 25164682-25164682 on Build GRCh38
rs777746663 Chr.2: 25164708-25164708 on Build GRCh38
rs546413064 Chr.2: 25164846-25164846 on Build GRCh38
rs768369820 Chr.2: 25164756-25164756 on Build GRCh38
rs764179008 Chr.2: 25164781-25164781 on Build GRCh38
rs937738497 Chr.2: 25164681-25164681 on Build GRCh38
rs1021304379 Chr.2: 25164955-25164955 on Build GRCh38
rs753856820 Chr.2: 25164783-25164783 on Build GRCh38
rs765158244 Chr.2: 25164759-25164759 on Build GRCh38
rs150178677 Chr.2: 25164646-25164646 on Build GRCh38
rs779629993 Chr.2: 25164729-25164729 on Build GRCh38
rs758791952 Chr.2: 25164797-25164797 on Build GRCh38
rs552737194 Chr.2: 25164964-25164964 on Build GRCh38
rs1020642742 Chr.2: 25164622-25164622 on Build GRCh38
rs929309552 Chr.2: 25164832-25164832 on Build GRCh38
rs201981936 Chr.2: 25164653-25164653 on Build GRCh38
rs184096782 Chr.2: 25164909-25164909 on Build GRCh38
rs779540150 Chr.2: 25164787-25164787 on Build GRCh38
rs749782556 Chr.2: 25164638-25164638 on Build GRCh38
rs778322412 Chr.2: 25164637-25164637 on Build GRCh38
rs146551109 Chr.2: 25164750-25164750 on Build GRCh38
rs766183863 Chr.2: 25164762-25164762 on Build GRCh38
rs1011064833 Chr.2: 25164974-25164976 on Build GRCh38
rs757720012 Chr.2: 25164718-25164718 on Build GRCh38
rs139750421 Chr.2: 25164747-25164747 on Build GRCh38
rs892627147 Chr.2: 25164556-25164556 on Build GRCh38
rs746815510 Chr.2: 25164752-25164753 on Build GRCh38
rs546529957 Chr.2: 25164558-25164558 on Build GRCh38
rs866305615 Chr.2: 25164700-25164700 on Build GRCh38
rs771288680 Chr.2: 25164639-25164639 on Build GRCh38
rs757397032 Chr.2: 25164784-25164784 on Build GRCh38
rs368474505 Chr.2: 25164621-25164621 on Build GRCh38
rs775229698 Chr.2: 25164744-25164744 on Build GRCh38
rs760210424 Chr.2: 25164660-25164660 on Build GRCh38
rs746202967 Chr.2: 25164736-25164736 on Build GRCh38
rs371271950 Chr.2: 25164531-25164531 on Build GRCh38
rs887861277 Chr.2: 25164850-25164850 on Build GRCh38
rs768938294 Chr.2: 25164890-25164890 on Build GRCh38
rs927614600 Chr.2: 25164887-25164887 on Build GRCh38
rs528518879 Chr.2: 25164607-25164607 on Build GRCh38
rs575862836 Chr.2: 25164538-25164538 on Build GRCh38
rs200057379 Chr.2: 25164636-25164636 on Build GRCh38
rs965989005 Chr.2: 25164563-25164563 on Build GRCh38
rs983949746 Chr.2: 25164773-25164773 on Build GRCh38
rs757563301 Chr.2: 25164692-25164695 on Build GRCh38
rs565125686 Chr.2: 25164572-25164572 on Build GRCh38
rs201107465 Chr.2: 25164665-25164665 on Build GRCh38
rs763850526 Chr.2: 25164815-25164815 on Build GRCh38
rs772614148 Chr.2: 25164742-25164742 on Build GRCh38
rs761386423 Chr.2: 25164751-25164751 on Build GRCh38
rs773178150 Chr.2: 25164761-25164761 on Build GRCh38
rs137879524 Chr.2: 25164659-25164659 on Build GRCh38
rs761447218 Chr.2: 25164757-25164757 on Build GRCh38
rs372746207 Chr.2: 25164810-25164810 on Build GRCh38
rs142402460 Chr.2: 25164703-25164703 on Build GRCh38
rs892233509 Chr.2: 25164962-25164962 on Build GRCh38
rs570382495 Chr.2: 25164931-25164931 on Build GRCh38
rs764950859 Chr.2: 25164669-25164669 on Build GRCh38
rs202076407 Chr.2: 25164767-25164767 on Build GRCh38
rs767700712 Chr.2: 25164690-25164690 on Build GRCh38
rs758258712 Chr.2: 25164683-25164683 on Build GRCh38
rs901663756 Chr.2: 25164914-25164914 on Build GRCh38
rs576568490 Chr.2: 25164932-25164932 on Build GRCh38
rs752925315 Chr.2: 25164769-25164769 on Build GRCh38
rs763531551 Chr.2: 25164661-25164661 on Build GRCh38
rs574485920 Chr.2: 25164526-25164526 on Build GRCh38
rs749738402 Chr.2: 25164717-25164717 on Build GRCh38
rs1007644863 Chr.2: 25164884-25164884 on Build GRCh38
rs746399347 Chr.2: 25164790-25164790 on Build GRCh38
rs200370644 Chr.2: 25164657-25164657 on Build GRCh38
rs919491316 Chr.2: 25164831-25164831 on Build GRCh38
rs776588032 Chr.2: 25164664-25164664 on Build GRCh38
rs557321408 Chr.2: 25164533-25164533 on Build GRCh38
rs779090267 Chr.2: 25164729-25164730 on Build GRCh38
rs776016303 Chr.2: 25164756-25164756 on Build GRCh38
rs774861632 Chr.2: 25164648-25164648 on Build GRCh38

View Full Product Details