Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 26728169-26728691 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNK3
Entrez Gene ID
3777
Gene Name
potassium two pore domain channel subfamily K member 3
Gene Aliases
K2p3.1, OAT1, PPH4, TASK, TASK-1, TBAK1
UniGene
Hs.24040
Gene Chromosome Location
Chr.2: 26692713-26733420 on Build GRCh38
Transcripts
NM_002246.2 XM_005264293.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6157682 c.1008C>T p.S336S Chr.2: 26728391-26728391 on Build GRCh38
4644439 c.1061G>T p.S354I Chr.2: 26728444-26728444 on Build GRCh38
6157681 c.1014G>T p.T338T Chr.2: 26728397-26728397 on Build GRCh38
4638296 c.932C>T p.S311L Chr.2: 26728315-26728315 on Build GRCh38
4634824 c.841A>G p.T281A Chr.2: 26728224-26728224 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748278164 Chr.2: 26728477-26728477 on Build GRCh38
rs751965823 Chr.2: 26728426-26728426 on Build GRCh38
rs916938508 Chr.2: 26728433-26728433 on Build GRCh38
rs778740492 Chr.2: 26728466-26728466 on Build GRCh38
rs375930383 Chr.2: 26728397-26728397 on Build GRCh38
rs777780035 Chr.2: 26728450-26728450 on Build GRCh38
rs746830880 Chr.2: 26728356-26728356 on Build GRCh38
rs745644356 Chr.2: 26728472-26728472 on Build GRCh38
rs202132140 Chr.2: 26728421-26728421 on Build GRCh38
rs753913235 Chr.2: 26728260-26728260 on Build GRCh38
rs543371252 Chr.2: 26728436-26728436 on Build GRCh38
rs775696759 Chr.2: 26728359-26728359 on Build GRCh38
rs774951646 Chr.2: 26728591-26728591 on Build GRCh38
rs774904281 Chr.2: 26728489-26728489 on Build GRCh38
rs930974747 Chr.2: 26728669-26728669 on Build GRCh38
rs867242780 Chr.2: 26728543-26728543 on Build GRCh38
rs753377260 Chr.2: 26728523-26728523 on Build GRCh38
rs767249538 Chr.2: 26728430-26728430 on Build GRCh38
rs368524386 Chr.2: 26728215-26728215 on Build GRCh38
rs898376327 Chr.2: 26728658-26728658 on Build GRCh38
rs544168447 Chr.2: 26728198-26728198 on Build GRCh38
rs747301305 Chr.2: 26728361-26728361 on Build GRCh38
rs781402431 Chr.2: 26728571-26728571 on Build GRCh38
rs773539407 Chr.2: 26728406-26728406 on Build GRCh38
rs758136070 Chr.2: 26728569-26728569 on Build GRCh38
rs866503101 Chr.2: 26728297-26728297 on Build GRCh38
rs759941263 Chr.2: 26728498-26728498 on Build GRCh38
rs969157808 Chr.2: 26728519-26728519 on Build GRCh38
rs201539884 Chr.2: 26728544-26728544 on Build GRCh38
rs760241147 Chr.2: 26728511-26728511 on Build GRCh38
rs367712936 Chr.2: 26728485-26728485 on Build GRCh38
rs777532152 Chr.2: 26728353-26728353 on Build GRCh38
rs561536552 Chr.2: 26728479-26728479 on Build GRCh38
rs746544523 Chr.2: 26728228-26728228 on Build GRCh38
rs776738377 Chr.2: 26728374-26728374 on Build GRCh38
rs35356422 Chr.2: 26728615-26728615 on Build GRCh38
rs781291012 Chr.2: 26728237-26728237 on Build GRCh38
rs752341544 Chr.2: 26728510-26728510 on Build GRCh38
rs760761985 Chr.2: 26728250-26728250 on Build GRCh38
rs749364968 Chr.2: 26728589-26728589 on Build GRCh38
rs760706655 Chr.2: 26728200-26728200 on Build GRCh38
rs927373603 Chr.2: 26728561-26728561 on Build GRCh38
rs764611480 Chr.2: 26728207-26728207 on Build GRCh38
rs879253615 Chr.2: 26728423-26728423 on Build GRCh38
rs1053971363 Chr.2: 26728224-26728226 on Build GRCh38
rs770050192 Chr.2: 26728576-26728576 on Build GRCh38
rs771597975 Chr.2: 26728590-26728590 on Build GRCh38
rs578012367 Chr.2: 26728266-26728266 on Build GRCh38
rs571255777 Chr.2: 26728598-26728598 on Build GRCh38
rs779537390 Chr.2: 26728227-26728227 on Build GRCh38
rs924076295 Chr.2: 26728545-26728545 on Build GRCh38
rs762626613 Chr.2: 26728247-26728247 on Build GRCh38
rs989493030 Chr.2: 26728507-26728507 on Build GRCh38
rs768903706 Chr.2: 26728476-26728476 on Build GRCh38
rs896419240 Chr.2: 26728405-26728405 on Build GRCh38
rs868657359 Chr.2: 26728273-26728273 on Build GRCh38
rs769847307 Chr.2: 26728243-26728243 on Build GRCh38
rs748250891 Chr.2: 26728574-26728574 on Build GRCh38
rs750738078 Chr.2: 26728217-26728217 on Build GRCh38
rs758115202 Chr.2: 26728218-26728218 on Build GRCh38
rs772499560 Chr.2: 26728612-26728612 on Build GRCh38
rs142003111 Chr.2: 26728329-26728329 on Build GRCh38
rs780914900 Chr.2: 26728335-26728335 on Build GRCh38
rs527873552 Chr.2: 26728429-26728429 on Build GRCh38
rs750247526 Chr.2: 26728562-26728562 on Build GRCh38
rs556393203 Chr.2: 26728210-26728210 on Build GRCh38
rs749116440 Chr.2: 26728465-26728465 on Build GRCh38
rs754539196 Chr.2: 26728331-26728331 on Build GRCh38
rs773110341 Chr.2: 26728245-26728245 on Build GRCh38
rs763435797 Chr.2: 26728481-26728481 on Build GRCh38
rs879194273 Chr.2: 26728355-26728355 on Build GRCh38
rs750837699 Chr.2: 26728283-26728283 on Build GRCh38
rs752862203 Chr.2: 26728195-26728195 on Build GRCh38
rs765801365 Chr.2: 26728403-26728403 on Build GRCh38
rs766585282 Chr.2: 26728424-26728424 on Build GRCh38
rs112204193 Chr.2: 26728451-26728451 on Build GRCh38
rs890491608 Chr.2: 26728659-26728659 on Build GRCh38
rs774439143 Chr.2: 26728185-26728185 on Build GRCh38
rs763388281 Chr.2: 26728417-26728417 on Build GRCh38
rs767302701 Chr.2: 26728506-26728506 on Build GRCh38
rs757193393 Chr.2: 26728529-26728529 on Build GRCh38
rs868278570 Chr.2: 26728552-26728552 on Build GRCh38
rs771260628 Chr.2: 26728602-26728602 on Build GRCh38
rs764097891 Chr.2: 26728255-26728255 on Build GRCh38
rs756119916 Chr.2: 26728337-26728337 on Build GRCh38
rs878979169 Chr.2: 26728490-26728490 on Build GRCh38
rs150626749 Chr.2: 26728336-26728336 on Build GRCh38
rs763581939 Chr.2: 26728514-26728514 on Build GRCh38
rs1036932096 Chr.2: 26728431-26728431 on Build GRCh38
rs908000319 Chr.2: 26728211-26728211 on Build GRCh38
rs1029431375 Chr.2: 26728263-26728263 on Build GRCh38
rs768854800 Chr.2: 26728372-26728372 on Build GRCh38
rs879221554 Chr.2: 26728559-26728559 on Build GRCh38
rs765638250 Chr.2: 26728277-26728277 on Build GRCh38
rs775526030 Chr.2: 26728248-26728248 on Build GRCh38
rs868035571 Chr.2: 26728540-26728540 on Build GRCh38
rs372175509 Chr.2: 26728316-26728316 on Build GRCh38
rs768231464 Chr.2: 26728236-26728236 on Build GRCh38
rs773773071 Chr.2: 26728480-26728480 on Build GRCh38
rs939853530 Chr.2: 26728605-26728605 on Build GRCh38
rs767612748 Chr.2: 26728191-26728191 on Build GRCh38
rs759867333 Chr.2: 26728187-26728187 on Build GRCh38
rs986931065 Chr.2: 26728209-26728209 on Build GRCh38
rs757625253 Chr.2: 26728213-26728213 on Build GRCh38
rs988826109 Chr.2: 26728235-26728235 on Build GRCh38
rs758646201 Chr.2: 26728303-26728303 on Build GRCh38
rs995173175 Chr.2: 26728463-26728463 on Build GRCh38
rs1037389757 Chr.2: 26728629-26728629 on Build GRCh38
rs1663002 Chr.2: 26728581-26728581 on Build GRCh38

View Full Product Details