Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27201267-27201761 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC5A6
Entrez Gene ID
8884
Gene Name
solute carrier family 5 member 6
Gene Aliases
SMVT
UniGene
Hs.435735
Gene Chromosome Location
Chr.2: 27199587-27212307 on Build GRCh38
Transcripts
NM_021095.2 NR_028323.1 XM_006712128.1 XM_006712129.1 XM_006712130.1 XM_011533146.2 XM_017005216.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4093480 c.1501G>A p.V501I Chr.2: 27201709-27201709 on Build GRCh38
4093479 c.1590C>A p.Y530* Chr.2: 27201408-27201408 on Build GRCh38
6038288 c.1600A>G p.N534D Chr.2: 27201398-27201398 on Build GRCh38
5396791 c.1645A>T p.T549S Chr.2: 27201353-27201353 on Build GRCh38
1407384 c.1538_1540delTCT p.F513delF Chr.2: 27201670-27201672 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs563212359 Chr.2: 27201576-27201576 on Build GRCh38
rs752758073 Chr.2: 27201472-27201472 on Build GRCh38
rs759809649 Chr.2: 27201657-27201657 on Build GRCh38
rs777004667 Chr.2: 27201689-27201699 on Build GRCh38
rs745584989 Chr.2: 27201497-27201497 on Build GRCh38
rs779884335 Chr.2: 27201716-27201716 on Build GRCh38
rs370762620 Chr.2: 27201488-27201488 on Build GRCh38
rs761698728 Chr.2: 27201439-27201439 on Build GRCh38
rs886435922 Chr.2: 27201582-27201582 on Build GRCh38
rs767090307 Chr.2: 27201304-27201304 on Build GRCh38
rs564386986 Chr.2: 27201333-27201333 on Build GRCh38
rs138079521 Chr.2: 27201359-27201359 on Build GRCh38
rs1028087677 Chr.2: 27201722-27201722 on Build GRCh38
rs751792770 Chr.2: 27201448-27201448 on Build GRCh38
rs760626016 Chr.2: 27201315-27201315 on Build GRCh38
rs142757011 Chr.2: 27201700-27201700 on Build GRCh38
rs771714496 Chr.2: 27201387-27201387 on Build GRCh38
rs186754671 Chr.2: 27201500-27201500 on Build GRCh38
rs564922108 Chr.2: 27201367-27201367 on Build GRCh38
rs758598811 Chr.2: 27201323-27201323 on Build GRCh38
rs140682068 Chr.2: 27201640-27201640 on Build GRCh38
rs778073407 Chr.2: 27201298-27201298 on Build GRCh38
rs1006898 Chr.2: 27201628-27201628 on Build GRCh38
rs772985969 Chr.2: 27201650-27201650 on Build GRCh38
rs986447912 Chr.2: 27201462-27201462 on Build GRCh38
rs746592998 Chr.2: 27201719-27201719 on Build GRCh38
rs985650827 Chr.2: 27201510-27201510 on Build GRCh38
rs142905141 Chr.2: 27201709-27201709 on Build GRCh38
rs1036930576 Chr.2: 27201612-27201612 on Build GRCh38
rs376257096 Chr.2: 27201337-27201337 on Build GRCh38
rs760754638 Chr.2: 27201421-27201421 on Build GRCh38
rs767700862 Chr.2: 27201667-27201667 on Build GRCh38
rs750797839 Chr.2: 27201710-27201710 on Build GRCh38
rs192806302 Chr.2: 27201630-27201630 on Build GRCh38
rs115224636 Chr.2: 27201586-27201586 on Build GRCh38
rs768225616 Chr.2: 27201729-27201729 on Build GRCh38
rs778308149 Chr.2: 27201480-27201480 on Build GRCh38
rs760835085 Chr.2: 27201676-27201676 on Build GRCh38
rs1064845 Chr.2: 27201735-27201735 on Build GRCh38
rs757703719 Chr.2: 27201704-27201704 on Build GRCh38
rs368529182 Chr.2: 27201489-27201489 on Build GRCh38
rs764025829 Chr.2: 27201685-27201685 on Build GRCh38
rs550834773 Chr.2: 27201571-27201571 on Build GRCh38
rs771163664 Chr.2: 27201354-27201354 on Build GRCh38
rs768609582 Chr.2: 27201494-27201494 on Build GRCh38
rs370089425 Chr.2: 27201341-27201341 on Build GRCh38
rs761597123 Chr.2: 27201643-27201643 on Build GRCh38
rs116756872 Chr.2: 27201507-27201507 on Build GRCh38
rs547451922 Chr.2: 27201556-27201556 on Build GRCh38
rs767647856 Chr.2: 27201469-27201469 on Build GRCh38
rs748656917 Chr.2: 27201336-27201336 on Build GRCh38
rs751739995 Chr.2: 27201331-27201331 on Build GRCh38
rs200415509 Chr.2: 27201332-27201332 on Build GRCh38
rs775137520 Chr.2: 27201388-27201388 on Build GRCh38
rs779357206 Chr.2: 27201496-27201496 on Build GRCh38
rs755123868 Chr.2: 27201452-27201452 on Build GRCh38
rs368015240 Chr.2: 27201476-27201476 on Build GRCh38
rs112006254 Chr.2: 27201329-27201329 on Build GRCh38
rs549131587 Chr.2: 27201651-27201651 on Build GRCh38
rs1023765546 Chr.2: 27201380-27201380 on Build GRCh38
rs201074076 Chr.2: 27201313-27201313 on Build GRCh38
rs578214644 Chr.2: 27201343-27201343 on Build GRCh38
rs980238378 Chr.2: 27201532-27201532 on Build GRCh38
rs556451974 Chr.2: 27201321-27201321 on Build GRCh38
rs776331506 Chr.2: 27201437-27201437 on Build GRCh38
rs780500266 Chr.2: 27201734-27201734 on Build GRCh38
rs374107283 Chr.2: 27201447-27201447 on Build GRCh38
rs747051003 Chr.2: 27201316-27201316 on Build GRCh38
rs751748605 Chr.2: 27201665-27201665 on Build GRCh38
rs139846984 Chr.2: 27201496-27201496 on Build GRCh38
rs771096745 Chr.2: 27201393-27201393 on Build GRCh38
rs561737304 Chr.2: 27201505-27201505 on Build GRCh38
rs140443329 Chr.2: 27201436-27201436 on Build GRCh38
rs752809492 Chr.2: 27201675-27201675 on Build GRCh38
rs760322392 Chr.2: 27201473-27201474 on Build GRCh38

View Full Product Details