Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27276497-27276924 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC5G
Entrez Gene ID
285126
Gene Name
DnaJ heat shock protein family (Hsp40) member C5 gamma
Gene Aliases
CSP-gamma
UniGene
Hs.116303
Gene Chromosome Location
Chr.2: 27275421-27281430 on Build GRCh38
Transcripts
NM_001303127.1 NM_001303128.1 NM_173650.2 NR_130126.1 XM_006711995.3 XM_017003872.1 XM_017003873.1 XM_017003874.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4093523 c.61G>A p.V21M Chr.2: 27276789-27276789 on Build GRCh38
1306670 c.107C>A p.S36Y Chr.2: 27276835-27276835 on Build GRCh38
2939719 c.84C>A p.A28A Chr.2: 27276812-27276812 on Build GRCh38
4515141 c.99C>T p.F33F Chr.2: 27276827-27276827 on Build GRCh38
2939718 c.81C>T p.G27G Chr.2: 27276809-27276809 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755196274 Chr.2: 27276828-27276828 on Build GRCh38
rs142361760 Chr.2: 27276756-27276756 on Build GRCh38
rs773244111 Chr.2: 27276696-27276696 on Build GRCh38
rs1037450992 Chr.2: 27276665-27276665 on Build GRCh38
rs940444504 Chr.2: 27276662-27276662 on Build GRCh38
rs17005979 Chr.2: 27276739-27276739 on Build GRCh38
rs768119856 Chr.2: 27276682-27276682 on Build GRCh38
rs61754191 Chr.2: 27276807-27276807 on Build GRCh38
rs907595535 Chr.2: 27276641-27276641 on Build GRCh38
rs938339350 Chr.2: 27276573-27276573 on Build GRCh38
rs992465462 Chr.2: 27276578-27276578 on Build GRCh38
rs771412830 Chr.2: 27276765-27276765 on Build GRCh38
rs770823287 Chr.2: 27276714-27276714 on Build GRCh38
rs374325862 Chr.2: 27276816-27276816 on Build GRCh38
rs773775819 Chr.2: 27276856-27276856 on Build GRCh38
rs747865057 Chr.2: 27276703-27276703 on Build GRCh38
rs780524321 Chr.2: 27276647-27276647 on Build GRCh38
rs570689817 Chr.2: 27276540-27276540 on Build GRCh38
rs980369768 Chr.2: 27276562-27276562 on Build GRCh38
rs764134862 Chr.2: 27276731-27276731 on Build GRCh38
rs780742016 Chr.2: 27276684-27276684 on Build GRCh38
rs200578672 Chr.2: 27276776-27276776 on Build GRCh38
rs765759486 Chr.2: 27276755-27276755 on Build GRCh38
rs989209033 Chr.2: 27276677-27276677 on Build GRCh38
rs751229428 Chr.2: 27276784-27276784 on Build GRCh38
rs760943600 Chr.2: 27276727-27276727 on Build GRCh38
rs866459754 Chr.2: 27276779-27276779 on Build GRCh38
rs749116882 Chr.2: 27276701-27276701 on Build GRCh38
rs970616572 Chr.2: 27276544-27276544 on Build GRCh38
rs765270534 Chr.2: 27276854-27276854 on Build GRCh38
rs762007084 Chr.2: 27276842-27276842 on Build GRCh38
rs898989501 Chr.2: 27276671-27276671 on Build GRCh38
rs146746840 Chr.2: 27276673-27276673 on Build GRCh38
rs540426372 Chr.2: 27276694-27276694 on Build GRCh38
rs775890343 Chr.2: 27276723-27276723 on Build GRCh38
rs370777845 Chr.2: 27276817-27276817 on Build GRCh38
rs200523042 Chr.2: 27276815-27276815 on Build GRCh38
rs777906918 Chr.2: 27276810-27276810 on Build GRCh38
rs369997758 Chr.2: 27276808-27276808 on Build GRCh38
rs776671681 Chr.2: 27276838-27276838 on Build GRCh38
rs151264953 Chr.2: 27276757-27276757 on Build GRCh38
rs761762407 Chr.2: 27276745-27276745 on Build GRCh38
rs576484435 Chr.2: 27276879-27276879 on Build GRCh38
rs377405405 Chr.2: 27276865-27276865 on Build GRCh38
rs746633419 Chr.2: 27276679-27276679 on Build GRCh38
rs565024612 Chr.2: 27276620-27276620 on Build GRCh38
rs373141439 Chr.2: 27276561-27276561 on Build GRCh38
rs755635652 Chr.2: 27276809-27276809 on Build GRCh38
rs537419895 Chr.2: 27276897-27276897 on Build GRCh38
rs927561377 Chr.2: 27276569-27276569 on Build GRCh38
rs200966551 Chr.2: 27276717-27276717 on Build GRCh38
rs751956791 Chr.2: 27276888-27276888 on Build GRCh38
rs906462528 Chr.2: 27276883-27276883 on Build GRCh38
rs766815834 Chr.2: 27276869-27276869 on Build GRCh38
rs747551863 Chr.2: 27276686-27276686 on Build GRCh38

View Full Product Details