Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 33186765-33187267 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LTBP1
Entrez Gene ID
4052
Gene Name
latent transforming growth factor beta binding protein 1
Gene Aliases
-
UniGene
Hs.619315
Gene Chromosome Location
Chr.2: 32947129-33399509 on Build GRCh38
Transcripts
NM_000627.3 NM_001166264.1 NM_001166265.1 NM_001166266.1 NM_206943.2 XM_005264317.3 XM_005264318.3 XM_011532853.2 XM_011532855.2 XM_011532856.2 XM_011532857.2 XM_011532858.2 XM_011532859.2 XM_011532860.2 XM_011532861.2 XM_011532862.2 XM_017004108.1 XM_017004109.1 XM_017004110.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1592138 c.423C>T p.T141T Chr.2: 33187055-33187055 on Build GRCh38
4994127 c.1336T>G p.S446A Chr.2: 33186990-33186990 on Build GRCh38
7082373 c.411C>T p.T137T Chr.2: 33187043-33187043 on Build GRCh38
1592140 c.260G>T p.C87F Chr.2: 33186892-33186892 on Build GRCh38
4001652 c.378G>A p.A126A Chr.2: 33187010-33187010 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760602343 Chr.2: 33187132-33187132 on Build GRCh38
rs748645847 Chr.2: 33187086-33187086 on Build GRCh38
rs770308014 Chr.2: 33187019-33187019 on Build GRCh38
rs774443073 Chr.2: 33187108-33187108 on Build GRCh38
rs561643397 Chr.2: 33187131-33187131 on Build GRCh38
rs143447673 Chr.2: 33187101-33187101 on Build GRCh38
rs748948546 Chr.2: 33187008-33187008 on Build GRCh38
rs758106962 Chr.2: 33187100-33187101 on Build GRCh38
rs377312659 Chr.2: 33186827-33186827 on Build GRCh38
rs1045450019 Chr.2: 33186835-33186835 on Build GRCh38
rs1001247690 Chr.2: 33186862-33186862 on Build GRCh38
rs1034423581 Chr.2: 33186873-33186873 on Build GRCh38
rs775249584 Chr.2: 33186802-33186802 on Build GRCh38
rs1065324 Chr.2: 33187010-33187010 on Build GRCh38
rs542598800 Chr.2: 33187006-33187006 on Build GRCh38
rs201669598 Chr.2: 33187056-33187056 on Build GRCh38
rs1008428229 Chr.2: 33187225-33187225 on Build GRCh38
rs906991066 Chr.2: 33187147-33187147 on Build GRCh38
rs752433122 Chr.2: 33186999-33186999 on Build GRCh38
rs917570135 Chr.2: 33186804-33186804 on Build GRCh38
rs372621206 Chr.2: 33187004-33187004 on Build GRCh38
rs923766139 Chr.2: 33187137-33187137 on Build GRCh38
rs986891885 Chr.2: 33186992-33186992 on Build GRCh38
rs767502677 Chr.2: 33186982-33186982 on Build GRCh38
rs773782781 Chr.2: 33186902-33186902 on Build GRCh38
rs144716939 Chr.2: 33186924-33186924 on Build GRCh38
rs775586232 Chr.2: 33187126-33187126 on Build GRCh38
rs530376661 Chr.2: 33187138-33187138 on Build GRCh38
rs777279636 Chr.2: 33187013-33187013 on Build GRCh38
rs763208458 Chr.2: 33187024-33187024 on Build GRCh38
rs957674605 Chr.2: 33186883-33186883 on Build GRCh38
rs541238538 Chr.2: 33187107-33187107 on Build GRCh38
rs770495458 Chr.2: 33186870-33186870 on Build GRCh38
rs117488785 Chr.2: 33186809-33186809 on Build GRCh38
rs545440987 Chr.2: 33187037-33187037 on Build GRCh38
rs769350777 Chr.2: 33186919-33186919 on Build GRCh38
rs1006861565 Chr.2: 33187045-33187045 on Build GRCh38
rs752651855 Chr.2: 33187039-33187039 on Build GRCh38
rs759627500 Chr.2: 33187035-33187035 on Build GRCh38
rs565829105 Chr.2: 33186912-33186912 on Build GRCh38
rs781746951 Chr.2: 33187085-33187085 on Build GRCh38
rs748706302 Chr.2: 33186865-33186865 on Build GRCh38
rs749875763 Chr.2: 33187097-33187099 on Build GRCh38
rs1020507708 Chr.2: 33187189-33187189 on Build GRCh38
rs76241135 Chr.2: 33187156-33187156 on Build GRCh38
rs751630713 Chr.2: 33186975-33186975 on Build GRCh38
rs371416508 Chr.2: 33187113-33187113 on Build GRCh38
rs565249174 Chr.2: 33187098-33187098 on Build GRCh38
rs772115525 Chr.2: 33187121-33187121 on Build GRCh38
rs906866184 Chr.2: 33186852-33186852 on Build GRCh38
rs141247547 Chr.2: 33187235-33187235 on Build GRCh38
rs866249737 Chr.2: 33186848-33186848 on Build GRCh38
rs113801394 Chr.2: 33186956-33186956 on Build GRCh38
rs941593461 Chr.2: 33187192-33187192 on Build GRCh38
rs778440402 Chr.2: 33187066-33187066 on Build GRCh38
rs549806132 Chr.2: 33187155-33187155 on Build GRCh38
rs762280103 Chr.2: 33186965-33186965 on Build GRCh38
rs748486296 Chr.2: 33187018-33187018 on Build GRCh38
rs544334303 Chr.2: 33187170-33187170 on Build GRCh38
rs745306877 Chr.2: 33187072-33187072 on Build GRCh38
rs574422673 Chr.2: 33186981-33186981 on Build GRCh38
rs866362378 Chr.2: 33186984-33186984 on Build GRCh38
rs866927424 Chr.2: 33186934-33186934 on Build GRCh38
rs118167062 Chr.2: 33186969-33186969 on Build GRCh38
rs554822385 Chr.2: 33186971-33186971 on Build GRCh38
rs1011852761 Chr.2: 33187186-33187186 on Build GRCh38
rs745424717 Chr.2: 33187011-33187011 on Build GRCh38
rs766463358 Chr.2: 33186972-33186972 on Build GRCh38
rs774476886 Chr.2: 33187029-33187029 on Build GRCh38
rs376639670 Chr.2: 33187054-33187054 on Build GRCh38
rs1044639547 Chr.2: 33186959-33186959 on Build GRCh38
rs766577222 Chr.2: 33187026-33187026 on Build GRCh38
rs1018105719 Chr.2: 33187065-33187065 on Build GRCh38
rs992303941 Chr.2: 33187148-33187148 on Build GRCh38
rs144101039 Chr.2: 33186914-33186914 on Build GRCh38
rs369080561 Chr.2: 33187055-33187055 on Build GRCh38
rs139973445 Chr.2: 33187009-33187009 on Build GRCh38
rs907721423 Chr.2: 33187094-33187094 on Build GRCh38
rs758010010 Chr.2: 33187083-33187083 on Build GRCh38
rs967544489 Chr.2: 33187198-33187198 on Build GRCh38
rs2290447 Chr.2: 33186799-33186799 on Build GRCh38
rs146553508 Chr.2: 33187194-33187194 on Build GRCh38
rs765485383 Chr.2: 33186966-33186966 on Build GRCh38
rs773622021 Chr.2: 33187099-33187099 on Build GRCh38
rs756916323 Chr.2: 33187058-33187058 on Build GRCh38

View Full Product Details