Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 43813158-43813663 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCG5
Entrez Gene ID
64240
Gene Name
ATP binding cassette subfamily G member 5
Gene Aliases
STSL
UniGene
Hs.132992
Gene Chromosome Location
Chr.2: 43806154-43838840 on Build GRCh38
Transcripts
NM_022436.2 XM_005264480.3 XM_006712073.3 XM_011533024.2 XM_011533025.2 XM_011533026.2 XM_011533027.2 XM_011533028.2 XM_005264364.4 XM_005264365.4

Gene Symbol
DYNC2LI1
Entrez Gene ID
51626
Gene Name
dynein cytoplasmic 2 light intermediate chain 1
Gene Aliases
CGI-60, D2LIC, LIC3
UniGene
Hs.371597
Gene Chromosome Location
Chr.2: 43774039-43828492 on Build GRCh38
Transcripts
NM_022436.2 XM_005264480.3 XM_006712073.3 XM_011533024.2 XM_011533025.2 XM_011533026.2 XM_011533027.2 XM_011533028.2 XM_005264364.4 XM_005264365.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4531674 c.1814G>A p.G605E Chr.2: 43813258-43813258 on Build GRCh38
1130779 c.1902T>A p.L634L Chr.2: 43813170-43813170 on Build GRCh38
3758242 c.1810C>G p.Q604E Chr.2: 43813262-43813262 on Build GRCh38
6158173 c.1888T>C p.F630L Chr.2: 43813184-43813184 on Build GRCh38
1306815 c.1792C>T p.P598S Chr.2: 43813280-43813280 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs917741139 Chr.2: 43813482-43813482 on Build GRCh38
rs773426923 Chr.2: 43813279-43813279 on Build GRCh38
rs750077360 Chr.2: 43813324-43813324 on Build GRCh38
rs762408008 Chr.2: 43813228-43813228 on Build GRCh38
rs529740762 Chr.2: 43813388-43813388 on Build GRCh38
rs762110858 Chr.2: 43813201-43813201 on Build GRCh38
rs554478312 Chr.2: 43813626-43813626 on Build GRCh38
rs751112708 Chr.2: 43813232-43813232 on Build GRCh38
rs946886036 Chr.2: 43813395-43813395 on Build GRCh38
rs756987849 Chr.2: 43813239-43813239 on Build GRCh38
rs911404180 Chr.2: 43813382-43813382 on Build GRCh38
rs544628876 Chr.2: 43813624-43813625 on Build GRCh38
rs1000953240 Chr.2: 43813556-43813556 on Build GRCh38
rs761337468 Chr.2: 43813318-43813318 on Build GRCh38
rs373162711 Chr.2: 43813325-43813325 on Build GRCh38
rs1052067801 Chr.2: 43813572-43813572 on Build GRCh38
rs150401285 Chr.2: 43813202-43813202 on Build GRCh38
rs183575221 Chr.2: 43813358-43813358 on Build GRCh38
rs768890956 Chr.2: 43813194-43813194 on Build GRCh38
rs369053191 Chr.2: 43813243-43813243 on Build GRCh38
rs780262808 Chr.2: 43813184-43813184 on Build GRCh38
rs947857986 Chr.2: 43813515-43813515 on Build GRCh38
rs199967611 Chr.2: 43813247-43813247 on Build GRCh38
rs6720173 Chr.2: 43813262-43813262 on Build GRCh38
rs763595177 Chr.2: 43813229-43813229 on Build GRCh38
rs753814411 Chr.2: 43813329-43813329 on Build GRCh38
rs528636525 Chr.2: 43813222-43813222 on Build GRCh38
rs754363595 Chr.2: 43813225-43813225 on Build GRCh38
rs368946924 Chr.2: 43813542-43813542 on Build GRCh38
rs779109455 Chr.2: 43813273-43813273 on Build GRCh38
rs1056837256 Chr.2: 43813441-43813441 on Build GRCh38
rs758427977 Chr.2: 43813337-43813337 on Build GRCh38
rs137996263 Chr.2: 43813220-43813220 on Build GRCh38
rs538134481 Chr.2: 43813567-43813567 on Build GRCh38
rs766293298 Chr.2: 43813327-43813327 on Build GRCh38
rs114780578 Chr.2: 43813316-43813316 on Build GRCh38
rs867332645 Chr.2: 43813385-43813385 on Build GRCh38
rs771224255 Chr.2: 43813351-43813351 on Build GRCh38
rs772378718 Chr.2: 43813277-43813277 on Build GRCh38
rs376900842 Chr.2: 43813335-43813335 on Build GRCh38
rs940539323 Chr.2: 43813191-43813191 on Build GRCh38
rs774651183 Chr.2: 43813199-43813199 on Build GRCh38
rs369394704 Chr.2: 43813352-43813352 on Build GRCh38
rs201698182 Chr.2: 43813189-43813189 on Build GRCh38
rs755032783 Chr.2: 43813330-43813330 on Build GRCh38
rs546689078 Chr.2: 43813421-43813421 on Build GRCh38
rs765042157 Chr.2: 43813510-43813510 on Build GRCh38
rs17031651 Chr.2: 43813423-43813423 on Build GRCh38
rs750285438 Chr.2: 43813244-43813244 on Build GRCh38
rs755999941 Chr.2: 43813246-43813246 on Build GRCh38
rs538413682 Chr.2: 43813426-43813426 on Build GRCh38
rs17031655 Chr.2: 43813519-43813519 on Build GRCh38
rs563519234 Chr.2: 43813595-43813595 on Build GRCh38
rs374012053 Chr.2: 43813216-43813216 on Build GRCh38
rs201902491 Chr.2: 43813301-43813301 on Build GRCh38
rs896470981 Chr.2: 43813444-43813444 on Build GRCh38
rs1044315270 Chr.2: 43813479-43813479 on Build GRCh38
rs144063868 Chr.2: 43813286-43813286 on Build GRCh38
rs950030773 Chr.2: 43813468-43813468 on Build GRCh38
rs772295351 Chr.2: 43813186-43813186 on Build GRCh38
rs747235883 Chr.2: 43813347-43813347 on Build GRCh38
rs140374206 Chr.2: 43813208-43813208 on Build GRCh38
rs1019086099 Chr.2: 43813326-43813326 on Build GRCh38
rs921987142 Chr.2: 43813364-43813366 on Build GRCh38
rs935338490 Chr.2: 43813434-43813434 on Build GRCh38
rs763578681 Chr.2: 43813315-43813315 on Build GRCh38
rs777055543 Chr.2: 43813297-43813297 on Build GRCh38
rs748314198 Chr.2: 43813276-43813276 on Build GRCh38
rs769429015 Chr.2: 43813182-43813182 on Build GRCh38
rs999477425 Chr.2: 43813187-43813187 on Build GRCh38
rs758463400 Chr.2: 43813319-43813319 on Build GRCh38
rs1023246862 Chr.2: 43813230-43813230 on Build GRCh38
rs778016002 Chr.2: 43813345-43813345 on Build GRCh38
rs780462357 Chr.2: 43813285-43813285 on Build GRCh38
rs150716811 Chr.2: 43813266-43813266 on Build GRCh38

View Full Product Details